Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133130145-133130637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRA1
Entrez Gene ID
84435
Gene Name
adhesion G protein-coupled receptor A1
Gene Aliases
GPR123
UniGene
Hs.435183
Gene Chromosome Location
Chr.10: 133070929-133131675 on Build GRCh38
Transcripts
NM_001083909.2 NM_001291085.1 XM_005252695.1 XM_011540273.1

SNP Detail

SNP ID Chromosome Location
rs965806292 Chr.10: 133130545-133130545 on Build GRCh38
rs937101713 Chr.10: 133130339-133130339 on Build GRCh38
rs561771711 Chr.10: 133130325-133130325 on Build GRCh38
rs547709545 Chr.10: 133130328-133130328 on Build GRCh38
rs949922381 Chr.10: 133130425-133130425 on Build GRCh38
rs577941269 Chr.10: 133130183-133130183 on Build GRCh38
rs1040198562 Chr.10: 133130518-133130518 on Build GRCh38
rs564357869 Chr.10: 133130180-133130180 on Build GRCh38
rs925638796 Chr.10: 133130394-133130394 on Build GRCh38
rs1018304946 Chr.10: 133130519-133130519 on Build GRCh38
rs543249742 Chr.10: 133130261-133130261 on Build GRCh38
rs527648142 Chr.10: 133130327-133130327 on Build GRCh38
rs945657327 Chr.10: 133130391-133130391 on Build GRCh38
rs1008717975 Chr.10: 133130199-133130199 on Build GRCh38
rs868699493 Chr.10: 133130172-133130172 on Build GRCh38
rs9419123 Chr.10: 133130178-133130178 on Build GRCh38
rs531723886 Chr.10: 133130280-133130280 on Build GRCh38
rs149387253 Chr.10: 133130546-133130546 on Build GRCh38
rs145482387 Chr.10: 133130281-133130281 on Build GRCh38
rs761206683 Chr.10: 133130390-133130390 on Build GRCh38
rs961186850 Chr.10: 133130299-133130299 on Build GRCh38
rs570183139 Chr.10: 133130471-133130471 on Build GRCh38
rs113289335 Chr.10: 133130548-133130548 on Build GRCh38
rs533905290 Chr.10: 133130290-133130290 on Build GRCh38
rs1009183481 Chr.10: 133130497-133130497 on Build GRCh38
rs769530758 Chr.10: 133130194-133130194 on Build GRCh38
rs571351916 Chr.10: 133130247-133130247 on Build GRCh38
rs1027545473 Chr.10: 133130566-133130566 on Build GRCh38
rs888846602 Chr.10: 133130479-133130479 on Build GRCh38
rs1019301462 Chr.10: 133130224-133130224 on Build GRCh38
rs1002189719 Chr.10: 133130615-133130615 on Build GRCh38
rs1036566387 Chr.10: 133130449-133130449 on Build GRCh38
rs980866768 Chr.10: 133130285-133130285 on Build GRCh38
rs73399162 Chr.10: 133130273-133130273 on Build GRCh38
rs952057390 Chr.10: 133130570-133130570 on Build GRCh38
rs184201530 Chr.10: 133130465-133130465 on Build GRCh38
rs9419124 Chr.10: 133130350-133130350 on Build GRCh38
rs533613610 Chr.10: 133130370-133130370 on Build GRCh38
rs777572022 Chr.10: 133130203-133130203 on Build GRCh38
rs935639500 Chr.10: 133130426-133130426 on Build GRCh38
rs750043278 Chr.10: 133130592-133130593 on Build GRCh38
rs555405797 Chr.10: 133130549-133130549 on Build GRCh38
rs1044329680 Chr.10: 133130392-133130392 on Build GRCh38

View Full Product Details