Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64116914-64117412 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLRT1
Entrez Gene ID
23769
Gene Name
fibronectin leucine rich transmembrane protein 1
Gene Aliases
SPG68
UniGene
Hs.584876
Gene Chromosome Location
Chr.11: 64035970-64119183 on Build GRCh38
Transcripts
NM_013280.4 XM_005273861.1 XM_011544883.1 NM_014067.3 XM_005273939.4 XM_006718521.3 XM_011544970.2

Gene Symbol
MACROD1
Entrez Gene ID
28992
Gene Name
MACRO domain containing 1
Gene Aliases
LRP16
UniGene
Hs.602898
Gene Chromosome Location
Chr.11: 63998554-64166652 on Build GRCh38
Transcripts
NM_013280.4 XM_005273861.1 XM_011544883.1 NM_014067.3 XM_005273939.4 XM_006718521.3 XM_011544970.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1207245 c.677G>A p.R226H Chr.11: 64116944-64116944 on Build GRCh38
6433450 c.905C>T p.S302F Chr.11: 64117172-64117172 on Build GRCh38
7207036 c.1110C>A p.D370E Chr.11: 64117377-64117377 on Build GRCh38
6246714 c.1068G>T p.M356I Chr.11: 64117335-64117335 on Build GRCh38
6215184 c.1019G>A p.R340Q Chr.11: 64117286-64117286 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750357914 Chr.11: 64117385-64117385 on Build GRCh38
rs779228423 Chr.11: 64117388-64117388 on Build GRCh38
rs775609758 Chr.11: 64117150-64117150 on Build GRCh38
rs770643761 Chr.11: 64117141-64117141 on Build GRCh38
rs772744996 Chr.11: 64117263-64117263 on Build GRCh38
rs559615919 Chr.11: 64117334-64117334 on Build GRCh38
rs762577383 Chr.11: 64117288-64117288 on Build GRCh38
rs775538870 Chr.11: 64117019-64117019 on Build GRCh38
rs763699368 Chr.11: 64117355-64117355 on Build GRCh38
rs372785993 Chr.11: 64117206-64117206 on Build GRCh38
rs761070130 Chr.11: 64117272-64117272 on Build GRCh38
rs776773158 Chr.11: 64117043-64117043 on Build GRCh38
rs563049161 Chr.11: 64117198-64117198 on Build GRCh38
rs140908634 Chr.11: 64117311-64117311 on Build GRCh38
rs151215662 Chr.11: 64117212-64117212 on Build GRCh38
rs768000465 Chr.11: 64117075-64117075 on Build GRCh38
rs202201413 Chr.11: 64117137-64117137 on Build GRCh38
rs766627575 Chr.11: 64116963-64116963 on Build GRCh38
rs568319026 Chr.11: 64117308-64117308 on Build GRCh38
rs777150139 Chr.11: 64117201-64117201 on Build GRCh38
rs929171159 Chr.11: 64117248-64117248 on Build GRCh38
rs767022154 Chr.11: 64117358-64117358 on Build GRCh38
rs769501659 Chr.11: 64117213-64117213 on Build GRCh38
rs3751120 Chr.11: 64116981-64116981 on Build GRCh38
rs772210831 Chr.11: 64117390-64117390 on Build GRCh38
rs368973071 Chr.11: 64117229-64117229 on Build GRCh38
rs556701976 Chr.11: 64117035-64117035 on Build GRCh38
rs145399902 Chr.11: 64117160-64117160 on Build GRCh38
rs748023643 Chr.11: 64117004-64117004 on Build GRCh38
rs753537051 Chr.11: 64117085-64117085 on Build GRCh38
rs748964357 Chr.11: 64116996-64116996 on Build GRCh38
rs144664182 Chr.11: 64117074-64117074 on Build GRCh38
rs762640509 Chr.11: 64117341-64117341 on Build GRCh38
rs377387995 Chr.11: 64116957-64116957 on Build GRCh38
rs528270608 Chr.11: 64117305-64117305 on Build GRCh38
rs751115779 Chr.11: 64117296-64117296 on Build GRCh38
rs34660800 Chr.11: 64117321-64117321 on Build GRCh38
rs779806631 Chr.11: 64116991-64116991 on Build GRCh38
rs377020952 Chr.11: 64117044-64117044 on Build GRCh38
rs752324400 Chr.11: 64117285-64117285 on Build GRCh38
rs373211331 Chr.11: 64117234-64117234 on Build GRCh38
rs781063452 Chr.11: 64117304-64117304 on Build GRCh38
rs748392464 Chr.11: 64117389-64117389 on Build GRCh38
rs565858250 Chr.11: 64116943-64116943 on Build GRCh38
rs375466804 Chr.11: 64117036-64117036 on Build GRCh38
rs146150625 Chr.11: 64117029-64117029 on Build GRCh38
rs761256926 Chr.11: 64117357-64117357 on Build GRCh38
rs764624707 Chr.11: 64117066-64117066 on Build GRCh38
rs144673578 Chr.11: 64117324-64117324 on Build GRCh38
rs973224653 Chr.11: 64117224-64117224 on Build GRCh38
rs778264587 Chr.11: 64117103-64117103 on Build GRCh38
rs147204811 Chr.11: 64117191-64117191 on Build GRCh38
rs377112197 Chr.11: 64116987-64116987 on Build GRCh38
rs34856996 Chr.11: 64117089-64117089 on Build GRCh38
rs759560338 Chr.11: 64116972-64116972 on Build GRCh38
rs771319324 Chr.11: 64117328-64117328 on Build GRCh38
rs747734134 Chr.11: 64117128-64117128 on Build GRCh38
rs916519862 Chr.11: 64117278-64117278 on Build GRCh38
rs139711112 Chr.11: 64117050-64117050 on Build GRCh38
rs1053939645 Chr.11: 64117329-64117329 on Build GRCh38
rs3751121 Chr.11: 64117002-64117002 on Build GRCh38
rs138445479 Chr.11: 64117387-64117387 on Build GRCh38
rs770733612 Chr.11: 64117037-64117037 on Build GRCh38
rs776473102 Chr.11: 64117147-64117147 on Build GRCh38
rs765311447 Chr.11: 64116980-64116980 on Build GRCh38
rs140497307 Chr.11: 64117199-64117199 on Build GRCh38
rs777982000 Chr.11: 64116985-64116985 on Build GRCh38
rs145828331 Chr.11: 64117053-64117053 on Build GRCh38
rs763641756 Chr.11: 64117294-64117294 on Build GRCh38
rs757865702 Chr.11: 64117200-64117200 on Build GRCh38
rs961463407 Chr.11: 64117219-64117219 on Build GRCh38
rs755874846 Chr.11: 64117306-64117306 on Build GRCh38
rs375237819 Chr.11: 64117134-64117134 on Build GRCh38
rs757875522 Chr.11: 64117133-64117133 on Build GRCh38
rs919080481 Chr.11: 64117247-64117247 on Build GRCh38
rs772300884 Chr.11: 64117322-64117322 on Build GRCh38
rs149882049 Chr.11: 64117244-64117244 on Build GRCh38
rs145808542 Chr.11: 64116940-64116940 on Build GRCh38
rs185115330 Chr.11: 64117182-64117182 on Build GRCh38
rs763041801 Chr.11: 64117159-64117159 on Build GRCh38
rs967595384 Chr.11: 64117167-64117167 on Build GRCh38
rs761090066 Chr.11: 64117325-64117325 on Build GRCh38
rs35247680 Chr.11: 64117275-64117275 on Build GRCh38
rs773886556 Chr.11: 64117269-64117269 on Build GRCh38
rs140370010 Chr.11: 64117218-64117218 on Build GRCh38
rs756527574 Chr.11: 64117003-64117003 on Build GRCh38
rs762478531 Chr.11: 64117166-64117166 on Build GRCh38
rs35250209 Chr.11: 64117266-64117266 on Build GRCh38
rs754088054 Chr.11: 64116966-64116966 on Build GRCh38
rs759754470 Chr.11: 64117045-64117045 on Build GRCh38
rs202104543 Chr.11: 64117307-64117307 on Build GRCh38
rs929198147 Chr.11: 64117051-64117051 on Build GRCh38
rs763341717 Chr.11: 64117060-64117060 on Build GRCh38
rs779105954 Chr.11: 64117312-64117312 on Build GRCh38
rs757729795 Chr.11: 64116988-64116988 on Build GRCh38
rs144550112 Chr.11: 64117210-64117210 on Build GRCh38
rs928607984 Chr.11: 64116990-64116990 on Build GRCh38
rs200363178 Chr.11: 64117251-64117251 on Build GRCh38
rs550169038 Chr.11: 64117356-64117356 on Build GRCh38
rs762388485 Chr.11: 64117245-64117245 on Build GRCh38
rs757240486 Chr.11: 64117209-64117209 on Build GRCh38
rs377711019 Chr.11: 64117303-64117303 on Build GRCh38
rs759006883 Chr.11: 64116984-64116984 on Build GRCh38
rs371817721 Chr.11: 64116941-64116941 on Build GRCh38
rs750867527 Chr.11: 64117190-64117190 on Build GRCh38
rs770098892 Chr.11: 64117148-64117148 on Build GRCh38
rs771760157 Chr.11: 64116944-64116944 on Build GRCh38
rs369580645 Chr.11: 64117164-64117164 on Build GRCh38
rs570176314 Chr.11: 64117386-64117386 on Build GRCh38
rs773116616 Chr.11: 64116948-64116948 on Build GRCh38
rs371858243 Chr.11: 64117106-64117106 on Build GRCh38
rs536649408 Chr.11: 64117320-64117320 on Build GRCh38

View Full Product Details