Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 848003-848551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC11
Entrez Gene ID
79844
Gene Name
zinc finger DHHC-type containing 11
Gene Aliases
ZNF399
UniGene
Hs.654874
Gene Chromosome Location
Chr.5: 795605-863634 on Build GRCh38
Transcripts
NM_024786.2 XM_017009869.1 XM_017009870.1 XM_017009871.1 XM_017009872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1070994 c.339C>T p.F113F Chr.5: 848544-848544 on Build GRCh38
1070993 c.339C>T p.F113F Chr.5: 848544-848544 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs953751941 Chr.5: 848091-848091 on Build GRCh38
rs764818506 Chr.5: 848514-848514 on Build GRCh38
rs879777118 Chr.5: 848047-848047 on Build GRCh38
rs775194138 Chr.5: 848522-848523 on Build GRCh38
rs376637668 Chr.5: 848522-848522 on Build GRCh38
rs772363141 Chr.5: 848462-848462 on Build GRCh38
rs995095191 Chr.5: 848057-848057 on Build GRCh38
rs551895597 Chr.5: 848444-848444 on Build GRCh38
rs537248155 Chr.5: 848351-848351 on Build GRCh38
rs746657259 Chr.5: 848523-848524 on Build GRCh38
rs529275426 Chr.5: 848053-848053 on Build GRCh38
rs200564151 Chr.5: 848456-848456 on Build GRCh38
rs756697985 Chr.5: 848026-848027 on Build GRCh38
rs534519949 Chr.5: 848466-848466 on Build GRCh38
rs539538895 Chr.5: 848352-848352 on Build GRCh38
rs1027871701 Chr.5: 848058-848058 on Build GRCh38
rs962373231 Chr.5: 848055-848055 on Build GRCh38
rs550970432 Chr.5: 848160-848160 on Build GRCh38
rs569265826 Chr.5: 848337-848337 on Build GRCh38
rs778911055 Chr.5: 848465-848465 on Build GRCh38
rs771873378 Chr.5: 848496-848496 on Build GRCh38
rs371976396 Chr.5: 848459-848459 on Build GRCh38
rs773063059 Chr.5: 848523-848523 on Build GRCh38
rs570508705 Chr.5: 848463-848463 on Build GRCh38
rs186566 Chr.5: 848022-848022 on Build GRCh38
rs749439967 Chr.5: 848396-848397 on Build GRCh38
rs761432402 Chr.5: 848484-848484 on Build GRCh38
rs552816107 Chr.5: 848471-848471 on Build GRCh38
rs562382193 Chr.5: 848037-848037 on Build GRCh38
rs768361547 Chr.5: 848525-848527 on Build GRCh38

View Full Product Details