Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 71679491-71680003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
THAP2
Entrez Gene ID
83591
Gene Name
THAP domain containing 2
Gene Aliases
-
UniGene
Hs.245798
Gene Chromosome Location
Chr.12: 71663897-71680648 on Build GRCh38
Transcripts
NM_031435.3

SNP Detail

SNP ID Chromosome Location
rs535464502 Chr.12: 71679564-71679564 on Build GRCh38
rs904951379 Chr.12: 71679651-71679651 on Build GRCh38
rs867022089 Chr.12: 71679645-71679645 on Build GRCh38
rs960566117 Chr.12: 71679751-71679751 on Build GRCh38
rs952025302 Chr.12: 71679861-71679861 on Build GRCh38
rs1040520108 Chr.12: 71679529-71679529 on Build GRCh38
rs1010588514 Chr.12: 71679592-71679592 on Build GRCh38
rs538185514 Chr.12: 71679528-71679530 on Build GRCh38
rs893359519 Chr.12: 71679568-71679568 on Build GRCh38
rs544961545 Chr.12: 71679571-71679571 on Build GRCh38
rs750991935 Chr.12: 71679788-71679788 on Build GRCh38
rs181445783 Chr.12: 71679566-71679566 on Build GRCh38
rs1001915141 Chr.12: 71679696-71679696 on Build GRCh38
rs996731253 Chr.12: 71679565-71679565 on Build GRCh38
rs1043829959 Chr.12: 71679951-71679951 on Build GRCh38
rs927860561 Chr.12: 71679786-71679786 on Build GRCh38
rs1034767893 Chr.12: 71679732-71679732 on Build GRCh38
rs878995010 Chr.12: 71679804-71679804 on Build GRCh38
rs139995450 Chr.12: 71679758-71679758 on Build GRCh38
rs539205814 Chr.12: 71679768-71679768 on Build GRCh38
rs1026256578 Chr.12: 71679844-71679844 on Build GRCh38
rs757515483 Chr.12: 71679853-71679853 on Build GRCh38
rs4558253 Chr.12: 71679665-71679665 on Build GRCh38
rs902057567 Chr.12: 71679538-71679538 on Build GRCh38
rs369538490 Chr.12: 71679871-71679871 on Build GRCh38
rs115385478 Chr.12: 71679890-71679890 on Build GRCh38
rs934816841 Chr.12: 71679548-71679548 on Build GRCh38
rs983760034 Chr.12: 71679634-71679634 on Build GRCh38
rs763828421 Chr.12: 71679547-71679547 on Build GRCh38

View Full Product Details