Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15536627-15537174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC16
Entrez Gene ID
23341
Gene Name
DnaJ heat shock protein family (Hsp40) member C16
Gene Aliases
-
UniGene
Hs.655410
Gene Chromosome Location
Chr.1: 15526694-15571733 on Build GRCh38
Transcripts
NM_001287811.1 NM_015291.3 NR_109898.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
897828 c.417C>T p.D139D Chr.1: 15536657-15536657 on Build GRCh38
5844103 c.542G>A p.W181* Chr.1: 15536782-15536782 on Build GRCh38
6647266 c.505G>A p.D169N Chr.1: 15536745-15536745 on Build GRCh38
1335322 c.571T>G p.L191V Chr.1: 15536811-15536811 on Build GRCh38
1986641 c.504C>T p.S168S Chr.1: 15536744-15536744 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753437690 Chr.1: 15536803-15536803 on Build GRCh38
rs534993449 Chr.1: 15536735-15536735 on Build GRCh38
rs764037269 Chr.1: 15536671-15536671 on Build GRCh38
rs905090234 Chr.1: 15536890-15536890 on Build GRCh38
rs376400359 Chr.1: 15536976-15536976 on Build GRCh38
rs773099239 Chr.1: 15536775-15536775 on Build GRCh38
rs757724911 Chr.1: 15536840-15536840 on Build GRCh38
rs750078791 Chr.1: 15536720-15536720 on Build GRCh38
rs993048276 Chr.1: 15536931-15536931 on Build GRCh38
rs769958348 Chr.1: 15536766-15536766 on Build GRCh38
rs11576719 Chr.1: 15536953-15536953 on Build GRCh38
rs6695141 Chr.1: 15537125-15537125 on Build GRCh38
rs150982165 Chr.1: 15536658-15536658 on Build GRCh38
rs774743191 Chr.1: 15536759-15536760 on Build GRCh38
rs778273278 Chr.1: 15536740-15536740 on Build GRCh38
rs546146398 Chr.1: 15536952-15536952 on Build GRCh38
rs905037858 Chr.1: 15537102-15537102 on Build GRCh38
rs146729141 Chr.1: 15536657-15536657 on Build GRCh38
rs1038195967 Chr.1: 15536881-15536881 on Build GRCh38
rs528813030 Chr.1: 15537001-15537001 on Build GRCh38
rs993377363 Chr.1: 15536998-15536998 on Build GRCh38
rs775593045 Chr.1: 15536767-15536767 on Build GRCh38
rs574975228 Chr.1: 15536902-15536902 on Build GRCh38
rs970661396 Chr.1: 15537121-15537121 on Build GRCh38
rs948913690 Chr.1: 15536932-15536932 on Build GRCh38
rs139382825 Chr.1: 15536747-15536747 on Build GRCh38
rs781683186 Chr.1: 15536855-15536855 on Build GRCh38
rs199649978 Chr.1: 15536676-15536676 on Build GRCh38
rs942959567 Chr.1: 15536741-15536741 on Build GRCh38
rs146679506 Chr.1: 15537103-15537103 on Build GRCh38
rs369643933 Chr.1: 15537007-15537007 on Build GRCh38
rs143627907 Chr.1: 15536919-15536919 on Build GRCh38
rs771285717 Chr.1: 15536652-15536652 on Build GRCh38
rs186338760 Chr.1: 15537070-15537070 on Build GRCh38
rs752081088 Chr.1: 15536836-15536836 on Build GRCh38
rs867623078 Chr.1: 15537101-15537101 on Build GRCh38
rs560017103 Chr.1: 15536991-15536991 on Build GRCh38
rs1037348296 Chr.1: 15536970-15536970 on Build GRCh38
rs761664045 Chr.1: 15536683-15536683 on Build GRCh38
rs112015425 Chr.1: 15536857-15536857 on Build GRCh38
rs61750327 Chr.1: 15536780-15536780 on Build GRCh38
rs76055663 Chr.1: 15536964-15536964 on Build GRCh38
rs142352537 Chr.1: 15536665-15536665 on Build GRCh38
rs150928872 Chr.1: 15536943-15536943 on Build GRCh38
rs747533060 Chr.1: 15536744-15536744 on Build GRCh38
rs140524364 Chr.1: 15536660-15536660 on Build GRCh38
rs774160972 Chr.1: 15536675-15536675 on Build GRCh38
rs561388320 Chr.1: 15537043-15537043 on Build GRCh38
rs539880581 Chr.1: 15537127-15537130 on Build GRCh38
rs746265443 Chr.1: 15536754-15536754 on Build GRCh38
rs767480447 Chr.1: 15536707-15536707 on Build GRCh38
rs563826623 Chr.1: 15536934-15536934 on Build GRCh38
rs554700134 Chr.1: 15536900-15536900 on Build GRCh38
rs968223638 Chr.1: 15536923-15536923 on Build GRCh38
rs983213740 Chr.1: 15537053-15537053 on Build GRCh38
rs1047137176 Chr.1: 15536807-15536807 on Build GRCh38
rs755869317 Chr.1: 15536722-15536722 on Build GRCh38
rs533170434 Chr.1: 15537119-15537119 on Build GRCh38
rs74761515 Chr.1: 15536868-15536868 on Build GRCh38
rs768685930 Chr.1: 15536768-15536768 on Build GRCh38
rs960410642 Chr.1: 15536917-15536917 on Build GRCh38
rs112362156 Chr.1: 15536961-15536961 on Build GRCh38
rs955987642 Chr.1: 15537084-15537084 on Build GRCh38
rs766159134 Chr.1: 15536725-15536725 on Build GRCh38
rs1038145787 Chr.1: 15537082-15537082 on Build GRCh38
rs751790191 Chr.1: 15536749-15536749 on Build GRCh38
rs764708356 Chr.1: 15536811-15536811 on Build GRCh38
rs773744296 Chr.1: 15537041-15537041 on Build GRCh38
rs370142117 Chr.1: 15536788-15536788 on Build GRCh38
rs756011850 Chr.1: 15537023-15537023 on Build GRCh38
rs1002095989 Chr.1: 15536896-15536896 on Build GRCh38
rs896433112 Chr.1: 15537143-15537143 on Build GRCh38
rs55707321 Chr.1: 15537065-15537065 on Build GRCh38
rs767693189 Chr.1: 15536839-15536839 on Build GRCh38
rs796216251 Chr.1: 15536708-15536708 on Build GRCh38
rs1020699968 Chr.1: 15536920-15536920 on Build GRCh38
rs761891997 Chr.1: 15536772-15536772 on Build GRCh38
rs774333530 Chr.1: 15536771-15536771 on Build GRCh38
rs866896226 Chr.1: 15536776-15536776 on Build GRCh38
rs371423001 Chr.1: 15536745-15536745 on Build GRCh38
rs140741623 Chr.1: 15537003-15537003 on Build GRCh38
rs757009565 Chr.1: 15537114-15537114 on Build GRCh38
rs767685755 Chr.1: 15536856-15536857 on Build GRCh38
rs753340096 Chr.1: 15536734-15536734 on Build GRCh38
rs934036354 Chr.1: 15536798-15536798 on Build GRCh38
rs1005366930 Chr.1: 15536867-15536867 on Build GRCh38
rs767390896 Chr.1: 15536774-15536774 on Build GRCh38
rs750439489 Chr.1: 15536857-15536858 on Build GRCh38

View Full Product Details