Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17074764-17075274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PADI2
Entrez Gene ID
11240
Gene Name
peptidyl arginine deiminase 2
Gene Aliases
PAD-H19, PAD2, PDI2
UniGene
Hs.33455
Gene Chromosome Location
Chr.1: 17066761-17119453 on Build GRCh38
Transcripts
NM_007365.2 XM_017000148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2075934 c.1504C>T p.R502* Chr.1: 17074901-17074901 on Build GRCh38
2075933 c.1505G>A p.R502Q Chr.1: 17074900-17074900 on Build GRCh38
1185344 c.1531G>A p.E511K Chr.1: 17074874-17074874 on Build GRCh38
2075931 c.1523G>A p.G508D Chr.1: 17074882-17074882 on Build GRCh38
368396 c.1519G>T p.D507Y Chr.1: 17074886-17074886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780536674 Chr.1: 17074900-17074900 on Build GRCh38
rs997929188 Chr.1: 17074974-17074974 on Build GRCh38
rs961096995 Chr.1: 17075091-17075091 on Build GRCh38
rs376359581 Chr.1: 17074965-17074965 on Build GRCh38
rs1037130187 Chr.1: 17075201-17075201 on Build GRCh38
rs565858861 Chr.1: 17075086-17075086 on Build GRCh38
rs763728279 Chr.1: 17074884-17074884 on Build GRCh38
rs748291869 Chr.1: 17074823-17074823 on Build GRCh38
rs909560363 Chr.1: 17074908-17074908 on Build GRCh38
rs769905126 Chr.1: 17074987-17074987 on Build GRCh38
rs150557436 Chr.1: 17074856-17074856 on Build GRCh38
rs748970650 Chr.1: 17074833-17074833 on Build GRCh38
rs1010534394 Chr.1: 17074976-17074976 on Build GRCh38
rs901336687 Chr.1: 17075180-17075180 on Build GRCh38
rs538682491 Chr.1: 17075068-17075068 on Build GRCh38
rs1048964879 Chr.1: 17074837-17074837 on Build GRCh38
rs867749004 Chr.1: 17074964-17074964 on Build GRCh38
rs752127007 Chr.1: 17074879-17074879 on Build GRCh38
rs867070855 Chr.1: 17074796-17074796 on Build GRCh38
rs750842130 Chr.1: 17074809-17074809 on Build GRCh38
rs923999028 Chr.1: 17074803-17074803 on Build GRCh38
rs148391776 Chr.1: 17074942-17074942 on Build GRCh38
rs758811759 Chr.1: 17074812-17074812 on Build GRCh38
rs1011003403 Chr.1: 17075233-17075233 on Build GRCh38
rs957952439 Chr.1: 17074968-17074968 on Build GRCh38
rs768748616 Chr.1: 17074863-17074863 on Build GRCh38
rs201460964 Chr.1: 17074907-17074907 on Build GRCh38
rs978176035 Chr.1: 17074797-17074797 on Build GRCh38
rs777510618 Chr.1: 17074826-17074826 on Build GRCh38
rs773639092 Chr.1: 17074909-17074909 on Build GRCh38
rs761379107 Chr.1: 17075158-17075158 on Build GRCh38
rs985010738 Chr.1: 17075159-17075159 on Build GRCh38
rs149737469 Chr.1: 17074865-17074865 on Build GRCh38
rs565056808 Chr.1: 17075186-17075186 on Build GRCh38
rs756835685 Chr.1: 17074897-17074897 on Build GRCh38
rs775943689 Chr.1: 17075140-17075140 on Build GRCh38
rs766728709 Chr.1: 17074877-17074877 on Build GRCh38
rs940071924 Chr.1: 17075169-17075169 on Build GRCh38
rs558606165 Chr.1: 17075085-17075085 on Build GRCh38
rs931307898 Chr.1: 17075046-17075046 on Build GRCh38
rs374979922 Chr.1: 17074842-17074842 on Build GRCh38
rs540038246 Chr.1: 17075077-17075077 on Build GRCh38
rs11587313 Chr.1: 17075199-17075199 on Build GRCh38
rs1022184676 Chr.1: 17075020-17075020 on Build GRCh38
rs771887262 Chr.1: 17074845-17074845 on Build GRCh38
rs779905920 Chr.1: 17074920-17074920 on Build GRCh38
rs765556248 Chr.1: 17074807-17074807 on Build GRCh38
rs771341829 Chr.1: 17075019-17075019 on Build GRCh38
rs749975419 Chr.1: 17074937-17074937 on Build GRCh38
rs372374970 Chr.1: 17074901-17074901 on Build GRCh38
rs920489380 Chr.1: 17075036-17075036 on Build GRCh38
rs763443384 Chr.1: 17074872-17074872 on Build GRCh38
rs746982569 Chr.1: 17074921-17074921 on Build GRCh38
rs113957605 Chr.1: 17075006-17075006 on Build GRCh38
rs534476274 Chr.1: 17075107-17075107 on Build GRCh38
rs765712228 Chr.1: 17075090-17075090 on Build GRCh38
rs143992403 Chr.1: 17075176-17075176 on Build GRCh38
rs745721327 Chr.1: 17074843-17074843 on Build GRCh38
rs990689405 Chr.1: 17075035-17075035 on Build GRCh38
rs369407341 Chr.1: 17074950-17074950 on Build GRCh38
rs373758719 Chr.1: 17074866-17074866 on Build GRCh38
rs981940394 Chr.1: 17075014-17075014 on Build GRCh38
rs776797825 Chr.1: 17074864-17074864 on Build GRCh38
rs766969953 Chr.1: 17074822-17074822 on Build GRCh38
rs1041255336 Chr.1: 17075172-17075172 on Build GRCh38
rs372511345 Chr.1: 17075139-17075139 on Build GRCh38
rs966458848 Chr.1: 17075007-17075007 on Build GRCh38
rs753436923 Chr.1: 17074887-17074887 on Build GRCh38
rs757036859 Chr.1: 17074834-17074834 on Build GRCh38
rs968060918 Chr.1: 17075069-17075069 on Build GRCh38
rs902284809 Chr.1: 17075246-17075246 on Build GRCh38
rs368695252 Chr.1: 17074883-17074883 on Build GRCh38
rs760120830 Chr.1: 17075088-17075088 on Build GRCh38
rs3818041 Chr.1: 17074788-17074788 on Build GRCh38
rs3818040 Chr.1: 17075245-17075245 on Build GRCh38
rs550257327 Chr.1: 17074975-17074975 on Build GRCh38
rs775560169 Chr.1: 17074848-17074848 on Build GRCh38
rs768623545 Chr.1: 17074941-17074941 on Build GRCh38

View Full Product Details