Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 17082308-17082818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PADI2
Entrez Gene ID
11240
Gene Name
peptidyl arginine deiminase 2
Gene Aliases
PAD-H19, PAD2, PDI2
UniGene
Hs.33455
Gene Chromosome Location
Chr.1: 17066761-17119453 on Build GRCh38
Transcripts
NM_007365.2 XM_017000148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1336405 c.1090G>T p.G364C Chr.1: 17082613-17082613 on Build GRCh38
4611591 c.1084delC p.H362fs*16 Chr.1: 17082619-17082619 on Build GRCh38
6736854 c.1078G>A p.A360T Chr.1: 17082625-17082625 on Build GRCh38
7245874 c.1158+5G>A p.? Chr.1: 17082540-17082540 on Build GRCh38
899851 c.1140C>A p.F380L Chr.1: 17082563-17082563 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768393170 Chr.1: 17082497-17082497 on Build GRCh38
rs1030158544 Chr.1: 17082612-17082612 on Build GRCh38
rs911227695 Chr.1: 17082412-17082412 on Build GRCh38
rs1013846388 Chr.1: 17082507-17082507 on Build GRCh38
rs749167747 Chr.1: 17082689-17082689 on Build GRCh38
rs935427175 Chr.1: 17082723-17082723 on Build GRCh38
rs973443975 Chr.1: 17082469-17082469 on Build GRCh38
rs1024876323 Chr.1: 17082383-17082383 on Build GRCh38
rs1039812091 Chr.1: 17082516-17082516 on Build GRCh38
rs987680922 Chr.1: 17082711-17082711 on Build GRCh38
rs770951409 Chr.1: 17082608-17082608 on Build GRCh38
rs942662984 Chr.1: 17082420-17082420 on Build GRCh38
rs749150877 Chr.1: 17082605-17082605 on Build GRCh38
rs139824702 Chr.1: 17082626-17082626 on Build GRCh38
rs2235925 Chr.1: 17082746-17082746 on Build GRCh38
rs745993826 Chr.1: 17082618-17082619 on Build GRCh38
rs754266084 Chr.1: 17082650-17082650 on Build GRCh38
rs754553171 Chr.1: 17082663-17082663 on Build GRCh38
rs755871420 Chr.1: 17082677-17082677 on Build GRCh38
rs554495339 Chr.1: 17082665-17082665 on Build GRCh38
rs769791839 Chr.1: 17082509-17082509 on Build GRCh38
rs548579800 Chr.1: 17082568-17082568 on Build GRCh38
rs943626775 Chr.1: 17082730-17082730 on Build GRCh38
rs139580948 Chr.1: 17082373-17082373 on Build GRCh38
rs936313175 Chr.1: 17082787-17082787 on Build GRCh38
rs912156108 Chr.1: 17082499-17082499 on Build GRCh38
rs368253753 Chr.1: 17082494-17082494 on Build GRCh38
rs748323481 Chr.1: 17082617-17082617 on Build GRCh38
rs553352822 Chr.1: 17082706-17082706 on Build GRCh38
rs766845740 Chr.1: 17082486-17082486 on Build GRCh38
rs377082407 Chr.1: 17082628-17082628 on Build GRCh38
rs751215965 Chr.1: 17082722-17082722 on Build GRCh38
rs764470165 Chr.1: 17082649-17082649 on Build GRCh38
rs920948693 Chr.1: 17082752-17082752 on Build GRCh38
rs925382116 Chr.1: 17082707-17082707 on Build GRCh38
rs919442734 Chr.1: 17082476-17082476 on Build GRCh38
rs2076617 Chr.1: 17082522-17082522 on Build GRCh38
rs747833869 Chr.1: 17082673-17082673 on Build GRCh38
rs367875986 Chr.1: 17082704-17082704 on Build GRCh38
rs551094660 Chr.1: 17082629-17082629 on Build GRCh38
rs750971550 Chr.1: 17082659-17082659 on Build GRCh38
rs956500869 Chr.1: 17082370-17082370 on Build GRCh38
rs1032143785 Chr.1: 17082344-17082344 on Build GRCh38
rs766401162 Chr.1: 17082524-17082524 on Build GRCh38
rs757679385 Chr.1: 17082655-17082655 on Build GRCh38
rs754585392 Chr.1: 17082584-17082584 on Build GRCh38
rs963800083 Chr.1: 17082463-17082463 on Build GRCh38
rs765842258 Chr.1: 17082656-17082656 on Build GRCh38
rs369091684 Chr.1: 17082633-17082633 on Build GRCh38
rs35341457 Chr.1: 17082708-17082709 on Build GRCh38
rs753054613 Chr.1: 17082538-17082538 on Build GRCh38
rs756474735 Chr.1: 17082546-17082546 on Build GRCh38
rs535937607 Chr.1: 17082721-17082721 on Build GRCh38
rs746575692 Chr.1: 17082582-17082582 on Build GRCh38
rs546556856 Chr.1: 17082342-17082342 on Build GRCh38
rs144164383 Chr.1: 17082585-17082585 on Build GRCh38
rs1052596011 Chr.1: 17082479-17082479 on Build GRCh38
rs923207371 Chr.1: 17082343-17082344 on Build GRCh38
rs779397644 Chr.1: 17082581-17082581 on Build GRCh38
rs1016052454 Chr.1: 17082559-17082559 on Build GRCh38
rs926636411 Chr.1: 17082439-17082439 on Build GRCh38
rs201659518 Chr.1: 17082572-17082572 on Build GRCh38
rs751701639 Chr.1: 17082529-17082529 on Build GRCh38
rs759701659 Chr.1: 17082779-17082797 on Build GRCh38
rs747897135 Chr.1: 17082586-17082586 on Build GRCh38
rs903918522 Chr.1: 17082437-17082437 on Build GRCh38
rs767782854 Chr.1: 17082536-17082536 on Build GRCh38
rs776557834 Chr.1: 17082500-17082500 on Build GRCh38
rs749189020 Chr.1: 17082658-17082658 on Build GRCh38
rs757882070 Chr.1: 17082574-17082574 on Build GRCh38
rs987241879 Chr.1: 17082404-17082404 on Build GRCh38
rs774283604 Chr.1: 17082624-17082624 on Build GRCh38
rs761770998 Chr.1: 17082502-17082502 on Build GRCh38
rs935526358 Chr.1: 17082464-17082464 on Build GRCh38
rs759657702 Chr.1: 17082530-17082530 on Build GRCh38
rs943633169 Chr.1: 17082513-17082513 on Build GRCh38
rs981218995 Chr.1: 17082435-17082435 on Build GRCh38
rs918095327 Chr.1: 17082749-17082749 on Build GRCh38
rs372245672 Chr.1: 17082521-17082521 on Build GRCh38
rs775636907 Chr.1: 17082631-17082632 on Build GRCh38
rs949560850 Chr.1: 17082775-17082775 on Build GRCh38
rs773244250 Chr.1: 17082595-17082595 on Build GRCh38
rs769982045 Chr.1: 17082627-17082629 on Build GRCh38
rs899621766 Chr.1: 17082338-17082338 on Build GRCh38
rs970650880 Chr.1: 17082409-17082409 on Build GRCh38
rs375682521 Chr.1: 17082682-17082682 on Build GRCh38

View Full Product Details