Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 20149235-20149723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLA2G2F
Entrez Gene ID
64600
Gene Name
phospholipase A2 group IIF
Gene Aliases
-
UniGene
Hs.302034
Gene Chromosome Location
Chr.1: 20139326-20150386 on Build GRCh38
Transcripts
NM_022819.3 XM_011541955.1 XM_011541956.1

SNP Detail

SNP ID Chromosome Location
rs966918131 Chr.1: 20149285-20149285 on Build GRCh38
rs769021920 Chr.1: 20149637-20149637 on Build GRCh38
rs992599911 Chr.1: 20149546-20149546 on Build GRCh38
rs532708596 Chr.1: 20149305-20149305 on Build GRCh38
rs115227696 Chr.1: 20149507-20149507 on Build GRCh38
rs143284373 Chr.1: 20149679-20149679 on Build GRCh38
rs867906065 Chr.1: 20149332-20149332 on Build GRCh38
rs564315710 Chr.1: 20149606-20149606 on Build GRCh38
rs939967645 Chr.1: 20149636-20149636 on Build GRCh38
rs954283847 Chr.1: 20149256-20149256 on Build GRCh38
rs902756158 Chr.1: 20149700-20149700 on Build GRCh38
rs566467986 Chr.1: 20149608-20149608 on Build GRCh38
rs533954814 Chr.1: 20149374-20149374 on Build GRCh38
rs1035632047 Chr.1: 20149539-20149539 on Build GRCh38
rs889751387 Chr.1: 20149676-20149676 on Build GRCh38
rs547415151 Chr.1: 20149403-20149403 on Build GRCh38
rs1022553401 Chr.1: 20149503-20149503 on Build GRCh38
rs186625582 Chr.1: 20149660-20149660 on Build GRCh38
rs74913983 Chr.1: 20149458-20149458 on Build GRCh38
rs566052836 Chr.1: 20149334-20149334 on Build GRCh38
rs915554018 Chr.1: 20149566-20149566 on Build GRCh38
rs558810216 Chr.1: 20149398-20149398 on Build GRCh38
rs915606397 Chr.1: 20149349-20149349 on Build GRCh38
rs939874012 Chr.1: 20149379-20149379 on Build GRCh38
rs776641640 Chr.1: 20149693-20149693 on Build GRCh38
rs992485728 Chr.1: 20149345-20149345 on Build GRCh38
rs1048358401 Chr.1: 20149372-20149372 on Build GRCh38
rs948597887 Chr.1: 20149365-20149365 on Build GRCh38
rs1011137245 Chr.1: 20149278-20149278 on Build GRCh38
rs942298129 Chr.1: 20149604-20149604 on Build GRCh38
rs866938971 Chr.1: 20149517-20149517 on Build GRCh38
rs1031491942 Chr.1: 20149462-20149462 on Build GRCh38
rs1044155290 Chr.1: 20149680-20149680 on Build GRCh38
rs191479337 Chr.1: 20149664-20149664 on Build GRCh38
rs12038747 Chr.1: 20149494-20149494 on Build GRCh38
rs774418496 Chr.1: 20149639-20149639 on Build GRCh38
rs545957391 Chr.1: 20149575-20149575 on Build GRCh38
rs572060830 Chr.1: 20149528-20149528 on Build GRCh38
rs924386388 Chr.1: 20149306-20149306 on Build GRCh38
rs1049699668 Chr.1: 20149443-20149443 on Build GRCh38
rs1021464424 Chr.1: 20149447-20149447 on Build GRCh38
rs968640554 Chr.1: 20149451-20149451 on Build GRCh38
rs1022396188 Chr.1: 20149284-20149284 on Build GRCh38
rs998256183 Chr.1: 20149430-20149430 on Build GRCh38
rs999695822 Chr.1: 20149538-20149538 on Build GRCh38
rs191379750 Chr.1: 20149387-20149387 on Build GRCh38
rs984002745 Chr.1: 20149595-20149595 on Build GRCh38
rs112318978 Chr.1: 20149397-20149397 on Build GRCh38
rs41307880 Chr.1: 20149429-20149429 on Build GRCh38
rs757530069 Chr.1: 20149298-20149298 on Build GRCh38
rs749340702 Chr.1: 20149545-20149545 on Build GRCh38
rs574371096 Chr.1: 20149493-20149493 on Build GRCh38
rs569221289 Chr.1: 20149441-20149441 on Build GRCh38

View Full Product Details