Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24136935-24137390 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL22RA1
Entrez Gene ID
58985
Gene Name
interleukin 22 receptor subunit alpha 1
Gene Aliases
CRF2-9, IL22R, IL22R1
UniGene
Hs.110915
Gene Chromosome Location
Chr.1: 24119771-24143285 on Build GRCh38
Transcripts
NM_021258.3 XM_011541882.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4692291 c.299G>A p.S100N Chr.1: 24137187-24137187 on Build GRCh38
3486388 c.182G>A p.G61E Chr.1: 24137304-24137304 on Build GRCh38
6455212 c.245C>T p.T82M Chr.1: 24137241-24137241 on Build GRCh38
7372063 c.212G>A p.C71Y Chr.1: 24137274-24137274 on Build GRCh38
1719147 c.189G>A p.R63R Chr.1: 24137297-24137297 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767286786 Chr.1: 24137300-24137300 on Build GRCh38
rs368021525 Chr.1: 24137105-24137105 on Build GRCh38
rs762941665 Chr.1: 24137128-24137128 on Build GRCh38
rs908996778 Chr.1: 24136974-24136974 on Build GRCh38
rs1053784222 Chr.1: 24137070-24137070 on Build GRCh38
rs767121534 Chr.1: 24136970-24136970 on Build GRCh38
rs41268135 Chr.1: 24137335-24137335 on Build GRCh38
rs369494000 Chr.1: 24137358-24137358 on Build GRCh38
rs201213086 Chr.1: 24137104-24137104 on Build GRCh38
rs376646024 Chr.1: 24137253-24137253 on Build GRCh38
rs984445596 Chr.1: 24137186-24137186 on Build GRCh38
rs937324364 Chr.1: 24137296-24137296 on Build GRCh38
rs752365268 Chr.1: 24137160-24137160 on Build GRCh38
rs754198545 Chr.1: 24137228-24137275 on Build GRCh38
rs751012724 Chr.1: 24137138-24137138 on Build GRCh38
rs147985725 Chr.1: 24137184-24137184 on Build GRCh38
rs751637875 Chr.1: 24137321-24137321 on Build GRCh38
rs950857675 Chr.1: 24137174-24137174 on Build GRCh38
rs891526189 Chr.1: 24137067-24137067 on Build GRCh38
rs200289901 Chr.1: 24137337-24137337 on Build GRCh38
rs768005511 Chr.1: 24137346-24137346 on Build GRCh38
rs760156645 Chr.1: 24137270-24137270 on Build GRCh38
rs780836581 Chr.1: 24137085-24137085 on Build GRCh38
rs759499053 Chr.1: 24137211-24137211 on Build GRCh38
rs758970972 Chr.1: 24137310-24137310 on Build GRCh38
rs763774627 Chr.1: 24137290-24137290 on Build GRCh38
rs374892261 Chr.1: 24137176-24137176 on Build GRCh38
rs556183942 Chr.1: 24137279-24137279 on Build GRCh38
rs750914539 Chr.1: 24137241-24137241 on Build GRCh38
rs943105620 Chr.1: 24136986-24136986 on Build GRCh38
rs187213669 Chr.1: 24137074-24137074 on Build GRCh38
rs148516044 Chr.1: 24137255-24137255 on Build GRCh38
rs769607269 Chr.1: 24137115-24137115 on Build GRCh38
rs144035143 Chr.1: 24137215-24137215 on Build GRCh38
rs758989459 Chr.1: 24137244-24137244 on Build GRCh38
rs905969269 Chr.1: 24137084-24137084 on Build GRCh38
rs200815886 Chr.1: 24137260-24137260 on Build GRCh38
rs147173863 Chr.1: 24137204-24137204 on Build GRCh38
rs761583215 Chr.1: 24137305-24137305 on Build GRCh38
rs1052963065 Chr.1: 24137066-24137066 on Build GRCh38
rs767118225 Chr.1: 24137153-24137153 on Build GRCh38
rs754225897 Chr.1: 24137167-24137167 on Build GRCh38
rs984417616 Chr.1: 24136971-24136971 on Build GRCh38
rs992270728 Chr.1: 24137101-24137101 on Build GRCh38
rs373981247 Chr.1: 24137269-24137269 on Build GRCh38
rs747003110 Chr.1: 24137345-24137345 on Build GRCh38
rs191965661 Chr.1: 24137086-24137086 on Build GRCh38
rs772991905 Chr.1: 24137195-24137195 on Build GRCh38
rs754558371 Chr.1: 24137083-24137083 on Build GRCh38
rs762795201 Chr.1: 24137197-24137197 on Build GRCh38
rs748319635 Chr.1: 24137332-24137332 on Build GRCh38
rs776386465 Chr.1: 24137297-24137297 on Build GRCh38
rs923447529 Chr.1: 24137006-24137006 on Build GRCh38
rs761493184 Chr.1: 24137353-24137353 on Build GRCh38
rs561539401 Chr.1: 24137020-24137020 on Build GRCh38
rs770672223 Chr.1: 24137263-24137263 on Build GRCh38
rs376252940 Chr.1: 24137329-24137329 on Build GRCh38
rs766854264 Chr.1: 24137318-24137318 on Build GRCh38
rs529790576 Chr.1: 24136997-24136997 on Build GRCh38
rs755773960 Chr.1: 24137161-24137161 on Build GRCh38
rs772936211 Chr.1: 24137123-24137123 on Build GRCh38
rs1026086732 Chr.1: 24137351-24137351 on Build GRCh38
rs761658544 Chr.1: 24137240-24137240 on Build GRCh38
rs770833663 Chr.1: 24137198-24137198 on Build GRCh38
rs745953437 Chr.1: 24137100-24137100 on Build GRCh38
rs776297440 Chr.1: 24137352-24137352 on Build GRCh38
rs371446595 Chr.1: 24137139-24137139 on Build GRCh38
rs574308339 Chr.1: 24137306-24137306 on Build GRCh38
rs542890009 Chr.1: 24137019-24137019 on Build GRCh38
rs933526125 Chr.1: 24137225-24137225 on Build GRCh38
rs775333840 Chr.1: 24137338-24137338 on Build GRCh38
rs201058183 Chr.1: 24137231-24137231 on Build GRCh38
rs142566825 Chr.1: 24137219-24137219 on Build GRCh38
rs376950768 Chr.1: 24137327-24137327 on Build GRCh38
rs149856921 Chr.1: 24137247-24137247 on Build GRCh38
rs768825346 Chr.1: 24137194-24137194 on Build GRCh38
rs4486393 Chr.1: 24137073-24137073 on Build GRCh38
rs377040917 Chr.1: 24137357-24137357 on Build GRCh38
rs747238659 Chr.1: 24137193-24137193 on Build GRCh38
rs905873042 Chr.1: 24137276-24137276 on Build GRCh38
rs778041754 Chr.1: 24137088-24137088 on Build GRCh38
rs41268137 Chr.1: 24137364-24137364 on Build GRCh38
rs760304840 Chr.1: 24137227-24137227 on Build GRCh38
rs749783128 Chr.1: 24137099-24137099 on Build GRCh38
rs757740132 Chr.1: 24137234-24137234 on Build GRCh38
rs200680853 Chr.1: 24137268-24137268 on Build GRCh38
rs201480321 Chr.1: 24137135-24137135 on Build GRCh38
rs757609834 Chr.1: 24137324-24137324 on Build GRCh38
rs5773069 Chr.1: 24137110-24137110 on Build GRCh38
rs370292845 Chr.1: 24137333-24137333 on Build GRCh38
rs760844229 Chr.1: 24137110-24137110 on Build GRCh38
rs143430940 Chr.1: 24137175-24137175 on Build GRCh38
rs745899530 Chr.1: 24137178-24137178 on Build GRCh38
rs536741554 Chr.1: 24137183-24137183 on Build GRCh38
rs1011360149 Chr.1: 24137071-24137071 on Build GRCh38
rs886250629 Chr.1: 24137355-24137355 on Build GRCh38
rs573375692 Chr.1: 24137232-24137232 on Build GRCh38
rs1042821423 Chr.1: 24137281-24137281 on Build GRCh38
rs781318907 Chr.1: 24137259-24137259 on Build GRCh38

View Full Product Details