Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 73927370-73927878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOXL1
Entrez Gene ID
4016
Gene Name
lysyl oxidase like 1
Gene Aliases
LOL, LOXL
UniGene
Hs.65436
Gene Chromosome Location
Chr.15: 73926451-73952141 on Build GRCh38
Transcripts
NM_005576.3 XM_011521555.1 NR_040066.1 NR_040067.1 NR_040068.1 NR_040069.1 NR_040070.1

Gene Symbol
LOXL1-AS1
Entrez Gene ID
100287616
Gene Name
LOXL1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.599785
Gene Chromosome Location
Chr.15: 73917468-73928248 on Build GRCh38
Transcripts
NM_005576.3 XM_011521555.1 NR_040066.1 NR_040067.1 NR_040068.1 NR_040069.1 NR_040070.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5461570 c.960G>T p.A320A Chr.15: 73927743-73927743 on Build GRCh38
4389685 c.929A>C p.Y310S Chr.15: 73927712-73927712 on Build GRCh38
4554983 c.638G>A p.G213D Chr.15: 73927421-73927421 on Build GRCh38
6050244 c.780_781insGCC p.V260_P261insA Chr.15: 73927563-73927564 on Build GRCh38
4128388 c.1066G>C p.V356L Chr.15: 73927849-73927849 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1050051646 Chr.15: 73927454-73927454 on Build GRCh38
rs750341881 Chr.15: 73927716-73927716 on Build GRCh38
rs898480433 Chr.15: 73927580-73927580 on Build GRCh38
rs535750269 Chr.15: 73927421-73927421 on Build GRCh38
rs749461688 Chr.15: 73927470-73927470 on Build GRCh38
rs779143433 Chr.15: 73927770-73927770 on Build GRCh38
rs773814901 Chr.15: 73927598-73927598 on Build GRCh38
rs761303432 Chr.15: 73927618-73927618 on Build GRCh38
rs374905538 Chr.15: 73927417-73927417 on Build GRCh38
rs931901934 Chr.15: 73927749-73927778 on Build GRCh38
rs932336600 Chr.15: 73927425-73927425 on Build GRCh38
rs932328615 Chr.15: 73927663-73927663 on Build GRCh38
rs539520823 Chr.15: 73927685-73927685 on Build GRCh38
rs750976975 Chr.15: 73927405-73927405 on Build GRCh38
rs557922714 Chr.15: 73927733-73927733 on Build GRCh38
rs1034097940 Chr.15: 73927857-73927857 on Build GRCh38
rs961081955 Chr.15: 73927697-73927697 on Build GRCh38
rs933068120 Chr.15: 73927763-73927763 on Build GRCh38
rs756159948 Chr.15: 73927443-73927443 on Build GRCh38
rs925228666 Chr.15: 73927742-73927742 on Build GRCh38
rs923012250 Chr.15: 73927748-73927748 on Build GRCh38
rs943069608 Chr.15: 73927412-73927412 on Build GRCh38
rs1051209864 Chr.15: 73927691-73927691 on Build GRCh38
rs1038261108 Chr.15: 73927775-73927775 on Build GRCh38
rs769666713 Chr.15: 73927563-73927565 on Build GRCh38
rs1023501921 Chr.15: 73927541-73927541 on Build GRCh38
rs1011677129 Chr.15: 73927515-73927515 on Build GRCh38
rs767996872 Chr.15: 73927398-73927398 on Build GRCh38
rs768986175 Chr.15: 73927503-73927503 on Build GRCh38
rs982539921 Chr.15: 73927600-73927600 on Build GRCh38
rs768274373 Chr.15: 73927404-73927415 on Build GRCh38
rs764871428 Chr.15: 73927411-73927411 on Build GRCh38
rs772342644 Chr.15: 73927583-73927583 on Build GRCh38
rs555933908 Chr.15: 73927782-73927782 on Build GRCh38
rs1021227970 Chr.15: 73927729-73927729 on Build GRCh38
rs748482862 Chr.15: 73927549-73927549 on Build GRCh38
rs369266200 Chr.15: 73927390-73927390 on Build GRCh38
rs185499294 Chr.15: 73927781-73927781 on Build GRCh38
rs781059487 Chr.15: 73927434-73927434 on Build GRCh38
rs974415887 Chr.15: 73927744-73927744 on Build GRCh38
rs1002168743 Chr.15: 73927820-73927820 on Build GRCh38
rs1017226210 Chr.15: 73927530-73927530 on Build GRCh38
rs767343358 Chr.15: 73927715-73927715 on Build GRCh38
rs774834031 Chr.15: 73927395-73927395 on Build GRCh38
rs762375758 Chr.15: 73927681-73927681 on Build GRCh38
rs908401428 Chr.15: 73927642-73927642 on Build GRCh38
rs893204461 Chr.15: 73927491-73927491 on Build GRCh38
rs893297665 Chr.15: 73927747-73927747 on Build GRCh38
rs745840832 Chr.15: 73927442-73927442 on Build GRCh38
rs780159202 Chr.15: 73927445-73927445 on Build GRCh38
rs1044481786 Chr.15: 73927753-73927753 on Build GRCh38
rs973836022 Chr.15: 73927660-73927660 on Build GRCh38
rs889646643 Chr.15: 73927520-73927520 on Build GRCh38
rs960618258 Chr.15: 73927595-73927595 on Build GRCh38
rs751194373 Chr.15: 73927686-73927686 on Build GRCh38
rs41435250 Chr.15: 73927743-73927743 on Build GRCh38
rs1038629327 Chr.15: 73927787-73927787 on Build GRCh38
rs780106036 Chr.15: 73927731-73927731 on Build GRCh38
rs777426965 Chr.15: 73927422-73927422 on Build GRCh38
rs979306571 Chr.15: 73927734-73927734 on Build GRCh38
rs898782284 Chr.15: 73927811-73927811 on Build GRCh38
rs940194335 Chr.15: 73927783-73927783 on Build GRCh38
rs887343852 Chr.15: 73927495-73927495 on Build GRCh38
rs759067104 Chr.15: 73927410-73927410 on Build GRCh38
rs550588716 Chr.15: 73927624-73927624 on Build GRCh38
rs747730813 Chr.15: 73927563-73927571 on Build GRCh38
rs757017225 Chr.15: 73927432-73927432 on Build GRCh38
rs1130133 Chr.15: 73927851-73927851 on Build GRCh38
rs1038742601 Chr.15: 73927413-73927413 on Build GRCh38
rs998990011 Chr.15: 73927438-73927438 on Build GRCh38
rs1051709582 Chr.15: 73927474-73927474 on Build GRCh38
rs921053371 Chr.15: 73927662-73927662 on Build GRCh38
rs934167970 Chr.15: 73927471-73927471 on Build GRCh38
rs756104720 Chr.15: 73927723-73927723 on Build GRCh38
rs12906373 Chr.15: 73927719-73927719 on Build GRCh38
rs774438838 Chr.15: 73927518-73927518 on Build GRCh38
rs991965130 Chr.15: 73927700-73927700 on Build GRCh38
rs776481735 Chr.15: 73927562-73927563 on Build GRCh38
rs970068089 Chr.15: 73927555-73927555 on Build GRCh38
rs751304280 Chr.15: 73927429-73927429 on Build GRCh38
rs41429348 Chr.15: 73927659-73927659 on Build GRCh38

View Full Product Details