Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 81002485-81002981 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MESDC1
Entrez Gene ID
59274
Gene Name
mesoderm development candidate 1
Gene Aliases
-
UniGene
Hs.513071
Gene Chromosome Location
Chr.15: 81000954-81004004 on Build GRCh38
Transcripts
NM_022566.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6711533 c.250G>A p.A84T Chr.15: 81002521-81002521 on Build GRCh38
4612347 c.324delG p.D110fs*13 Chr.15: 81002595-81002595 on Build GRCh38
6711535 c.510G>A p.T170T Chr.15: 81002781-81002781 on Build GRCh38
4616750 c.564G>T p.S188S Chr.15: 81002835-81002835 on Build GRCh38
4617614 c.214G>A p.A72T Chr.15: 81002485-81002485 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753404857 Chr.15: 81002614-81002614 on Build GRCh38
rs757845374 Chr.15: 81002643-81002643 on Build GRCh38
rs911513174 Chr.15: 81002758-81002758 on Build GRCh38
rs554668980 Chr.15: 81002793-81002793 on Build GRCh38
rs768156679 Chr.15: 81002938-81002938 on Build GRCh38
rs916608448 Chr.15: 81002607-81002607 on Build GRCh38
rs770195738 Chr.15: 81002584-81002584 on Build GRCh38
rs751806417 Chr.15: 81002841-81002841 on Build GRCh38
rs896284428 Chr.15: 81002853-81002853 on Build GRCh38
rs372943666 Chr.15: 81002538-81002538 on Build GRCh38
rs776050269 Chr.15: 81002688-81002688 on Build GRCh38
rs749880685 Chr.15: 81002536-81002536 on Build GRCh38
rs760516118 Chr.15: 81002681-81002681 on Build GRCh38
rs535656571 Chr.15: 81002964-81002964 on Build GRCh38
rs748528050 Chr.15: 81002553-81002553 on Build GRCh38
rs371053006 Chr.15: 81002640-81002640 on Build GRCh38
rs758105022 Chr.15: 81002721-81002721 on Build GRCh38
rs140101445 Chr.15: 81002573-81002573 on Build GRCh38
rs759075710 Chr.15: 81002703-81002703 on Build GRCh38
rs1044339843 Chr.15: 81002646-81002646 on Build GRCh38
rs142209157 Chr.15: 81002895-81002895 on Build GRCh38
rs750885763 Chr.15: 81002961-81002961 on Build GRCh38
rs771147747 Chr.15: 81002508-81002508 on Build GRCh38
rs774076838 Chr.15: 81002942-81002942 on Build GRCh38
rs935460729 Chr.15: 81002682-81002682 on Build GRCh38
rs372825596 Chr.15: 81002773-81002773 on Build GRCh38
rs962026373 Chr.15: 81002767-81002767 on Build GRCh38
rs776568988 Chr.15: 81002818-81002818 on Build GRCh38
rs955546793 Chr.15: 81002739-81002739 on Build GRCh38
rs146088440 Chr.15: 81002564-81002564 on Build GRCh38
rs905141259 Chr.15: 81002941-81002941 on Build GRCh38
rs995904130 Chr.15: 81002706-81002706 on Build GRCh38
rs753965117 Chr.15: 81002859-81002859 on Build GRCh38
rs779225082 Chr.15: 81002881-81002881 on Build GRCh38
rs768090252 Chr.15: 81002846-81002846 on Build GRCh38
rs990879914 Chr.15: 81002808-81002808 on Build GRCh38
rs367956130 Chr.15: 81002571-81002571 on Build GRCh38
rs1015216082 Chr.15: 81002766-81002766 on Build GRCh38
rs375988914 Chr.15: 81002605-81002605 on Build GRCh38
rs763947378 Chr.15: 81002529-81002529 on Build GRCh38
rs146488661 Chr.15: 81002595-81002595 on Build GRCh38
rs746823055 Chr.15: 81002812-81002812 on Build GRCh38
rs773126503 Chr.15: 81002670-81002670 on Build GRCh38
rs761703741 Chr.15: 81002587-81002587 on Build GRCh38
rs375724238 Chr.15: 81002957-81002957 on Build GRCh38
rs758715467 Chr.15: 81002634-81002634 on Build GRCh38
rs747927954 Chr.15: 81002665-81002665 on Build GRCh38
rs766352666 Chr.15: 81002684-81002684 on Build GRCh38
rs773044637 Chr.15: 81002599-81002599 on Build GRCh38
rs201461230 Chr.15: 81002531-81002531 on Build GRCh38
rs984648370 Chr.15: 81002533-81002533 on Build GRCh38
rs756285359 Chr.15: 81002785-81002785 on Build GRCh38
rs775052186 Chr.15: 81002829-81002829 on Build GRCh38
rs775888015 Chr.15: 81002658-81002658 on Build GRCh38
rs745961400 Chr.15: 81002649-81002649 on Build GRCh38
rs746730101 Chr.15: 81002882-81002882 on Build GRCh38
rs750944779 Chr.15: 81002851-81002851 on Build GRCh38
rs749777713 Chr.15: 81002513-81002513 on Build GRCh38
rs770688555 Chr.15: 81002884-81002884 on Build GRCh38
rs769785232 Chr.15: 81002900-81002900 on Build GRCh38
rs370565639 Chr.15: 81002839-81002839 on Build GRCh38
rs762502720 Chr.15: 81002830-81002830 on Build GRCh38
rs745905529 Chr.15: 81002783-81002783 on Build GRCh38
rs778177126 Chr.15: 81002557-81002557 on Build GRCh38
rs753023700 Chr.15: 81002629-81002629 on Build GRCh38
rs767367199 Chr.15: 81002535-81002535 on Build GRCh38
rs759882211 Chr.15: 81002527-81002527 on Build GRCh38
rs774227920 Chr.15: 81002586-81002586 on Build GRCh38
rs754568773 Chr.15: 81002622-81002622 on Build GRCh38
rs1005849846 Chr.15: 81002756-81002756 on Build GRCh38
rs755087507 Chr.15: 81002865-81002865 on Build GRCh38
rs770076998 Chr.15: 81002651-81002651 on Build GRCh38
rs762532190 Chr.15: 81002932-81002932 on Build GRCh38
rs780267135 Chr.15: 81002856-81002856 on Build GRCh38
rs781766377 Chr.15: 81002776-81002776 on Build GRCh38
rs572653982 Chr.15: 81002817-81002817 on Build GRCh38
rs764876946 Chr.15: 81002625-81002625 on Build GRCh38
rs138274571 Chr.15: 81002887-81002887 on Build GRCh38
rs151231136 Chr.15: 81002943-81002943 on Build GRCh38
rs949241070 Chr.15: 81002871-81002871 on Build GRCh38
rs769259302 Chr.15: 81002828-81002828 on Build GRCh38
rs759515265 Chr.15: 81002823-81002823 on Build GRCh38
rs762270733 Chr.15: 81002842-81002842 on Build GRCh38
rs772565870 Chr.15: 81002556-81002556 on Build GRCh38
rs935631853 Chr.15: 81002861-81002861 on Build GRCh38
rs774808064 Chr.15: 81002901-81002901 on Build GRCh38
rs1016320861 Chr.15: 81002958-81002958 on Build GRCh38
rs539190409 Chr.15: 81002685-81002685 on Build GRCh38
rs901586607 Chr.15: 81002701-81002701 on Build GRCh38
rs942697537 Chr.15: 81002763-81002763 on Build GRCh38
rs371377128 Chr.15: 81002960-81002960 on Build GRCh38
rs764817896 Chr.15: 81002710-81002710 on Build GRCh38
rs1006324901 Chr.15: 81002521-81002521 on Build GRCh38
rs761826442 Chr.15: 81002534-81002534 on Build GRCh38
rs34802477 Chr.15: 81002544-81002544 on Build GRCh38
rs780330390 Chr.15: 81002789-81002789 on Build GRCh38
rs752320822 Chr.15: 81002718-81002718 on Build GRCh38
rs759825144 Chr.15: 81002581-81002581 on Build GRCh38
rs781405318 Chr.15: 81002648-81002648 on Build GRCh38
rs760716073 Chr.15: 81002600-81002600 on Build GRCh38
rs776745069 Chr.15: 81002515-81002515 on Build GRCh38
rs904859745 Chr.15: 81002506-81002506 on Build GRCh38
rs368204597 Chr.15: 81002745-81002745 on Build GRCh38
rs765803859 Chr.15: 81002610-81002610 on Build GRCh38
rs778253161 Chr.15: 81002635-81002635 on Build GRCh38
rs772824200 Chr.15: 81002799-81002799 on Build GRCh38
rs752022115 Chr.15: 81002751-81002751 on Build GRCh38
rs756726952 Chr.15: 81002855-81002855 on Build GRCh38
rs779237573 Chr.15: 81002552-81002552 on Build GRCh38
rs962149775 Chr.15: 81002565-81002565 on Build GRCh38

View Full Product Details