Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7613962-7614464 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FXR2
Entrez Gene ID
9513
Gene Name
FMR1 autosomal homolog 2
Gene Aliases
FMR1L2, FXR2P
UniGene
Hs.52788
Gene Chromosome Location
Chr.17: 7591230-7614897 on Build GRCh38
Transcripts
NM_004860.3 NM_001289114.1

Gene Symbol
SHBG
Entrez Gene ID
6462
Gene Name
sex hormone binding globulin
Gene Aliases
ABP, SBP, TEBG
UniGene
Hs.632235
Gene Chromosome Location
Chr.17: 7614064-7633383 on Build GRCh38
Transcripts
NM_004860.3 NM_001289114.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5387410 c.75C>T p.F25F Chr.17: 7614458-7614458 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1001096819 Chr.17: 7614285-7614285 on Build GRCh38
rs529609974 Chr.17: 7614223-7614223 on Build GRCh38
rs896573851 Chr.17: 7614235-7614235 on Build GRCh38
rs550484358 Chr.17: 7614212-7614212 on Build GRCh38
rs749773723 Chr.17: 7614428-7614428 on Build GRCh38
rs777522966 Chr.17: 7614434-7614444 on Build GRCh38
rs536658596 Chr.17: 7614045-7614045 on Build GRCh38
rs944576773 Chr.17: 7614095-7614095 on Build GRCh38
rs901920948 Chr.17: 7614361-7614362 on Build GRCh38
rs921724270 Chr.17: 7614147-7614147 on Build GRCh38
rs189746071 Chr.17: 7614104-7614104 on Build GRCh38
rs756801287 Chr.17: 7614430-7614430 on Build GRCh38
rs762330205 Chr.17: 7614293-7614293 on Build GRCh38
rs889080753 Chr.17: 7614079-7614079 on Build GRCh38
rs533623556 Chr.17: 7614370-7614370 on Build GRCh38
rs896449474 Chr.17: 7614427-7614427 on Build GRCh38
rs930557839 Chr.17: 7614148-7614148 on Build GRCh38
rs1038869042 Chr.17: 7614142-7614142 on Build GRCh38
rs965830615 Chr.17: 7614334-7614334 on Build GRCh38
rs778205181 Chr.17: 7614423-7614423 on Build GRCh38
rs1023818353 Chr.17: 7614265-7614265 on Build GRCh38
rs544043356 Chr.17: 7614426-7614426 on Build GRCh38
rs977270082 Chr.17: 7614156-7614156 on Build GRCh38
rs753041831 Chr.17: 7614158-7614158 on Build GRCh38
rs1032040530 Chr.17: 7614174-7614174 on Build GRCh38
rs777080412 Chr.17: 7614264-7614264 on Build GRCh38
rs939512871 Chr.17: 7614005-7614005 on Build GRCh38
rs939542858 Chr.17: 7614274-7614275 on Build GRCh38
rs1027651808 Chr.17: 7614074-7614074 on Build GRCh38
rs555442557 Chr.17: 7614375-7614375 on Build GRCh38
rs915588559 Chr.17: 7614224-7614224 on Build GRCh38
rs968599205 Chr.17: 7614266-7614266 on Build GRCh38
rs930443157 Chr.17: 7614379-7614379 on Build GRCh38
rs988547507 Chr.17: 7614061-7614061 on Build GRCh38
rs1040511326 Chr.17: 7614354-7614354 on Build GRCh38
rs979626474 Chr.17: 7613997-7613997 on Build GRCh38
rs557500822 Chr.17: 7614324-7614324 on Build GRCh38
rs981177158 Chr.17: 7614238-7614238 on Build GRCh38
rs1031506414 Chr.17: 7614388-7614388 on Build GRCh38
rs750789430 Chr.17: 7614346-7614346 on Build GRCh38
rs115150806 Chr.17: 7614406-7614406 on Build GRCh38
rs180679429 Chr.17: 7614386-7614386 on Build GRCh38
rs1004880110 Chr.17: 7614187-7614187 on Build GRCh38
rs564012928 Chr.17: 7614046-7614046 on Build GRCh38
rs928387194 Chr.17: 7614241-7614241 on Build GRCh38
rs1031201455 Chr.17: 7614286-7614286 on Build GRCh38
rs1056603608 Chr.17: 7614222-7614222 on Build GRCh38
rs111226773 Chr.17: 7614342-7614342 on Build GRCh38
rs562441636 Chr.17: 7614436-7614436 on Build GRCh38
rs907792958 Chr.17: 7614412-7614412 on Build GRCh38
rs886461066 Chr.17: 7614182-7614182 on Build GRCh38
rs1019010770 Chr.17: 7614105-7614105 on Build GRCh38
rs966289826 Chr.17: 7614121-7614121 on Build GRCh38
rs988432536 Chr.17: 7614311-7614311 on Build GRCh38
rs375920561 Chr.17: 7614022-7614022 on Build GRCh38
rs921804993 Chr.17: 7614348-7614348 on Build GRCh38
rs540325106 Chr.17: 7614231-7614232 on Build GRCh38
rs539416486 Chr.17: 7614277-7614277 on Build GRCh38
rs995278639 Chr.17: 7614054-7614054 on Build GRCh38
rs911637051 Chr.17: 7614077-7614077 on Build GRCh38
rs751765397 Chr.17: 7614011-7614011 on Build GRCh38
rs538446789 Chr.17: 7614186-7614186 on Build GRCh38
rs940551705 Chr.17: 7614422-7614422 on Build GRCh38
rs1007541498 Chr.17: 7614343-7614343 on Build GRCh38
rs773899662 Chr.17: 7613989-7613989 on Build GRCh38
rs1048982858 Chr.17: 7614177-7614177 on Build GRCh38

View Full Product Details