Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 38852930-38853417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL23
Entrez Gene ID
9349
Gene Name
ribosomal protein L23
Gene Aliases
L23, rpL17
UniGene
Hs.406300
Gene Chromosome Location
Chr.17: 38850068-38853800 on Build GRCh38
Transcripts
NM_000978.3 NR_002576.1

Gene Symbol
SNORA21
Entrez Gene ID
619505
Gene Name
small nucleolar RNA, H/ACA box 21
Gene Aliases
ACA21
UniGene
Hs.738742
Gene Chromosome Location
Chr.17: 38852863-38852995 on Build GRCh38
Transcripts
NM_000978.3 NR_002576.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1736218 c.83G>T p.C28F Chr.17: 38853036-38853036 on Build GRCh38
4065925 c.44G>A p.R15Q Chr.17: 38853075-38853075 on Build GRCh38
1224209 c.88G>A p.D30N Chr.17: 38853031-38853031 on Build GRCh38
3370694 c.45G>C p.R15R Chr.17: 38853074-38853074 on Build GRCh38
6220692 c.78C>T p.I26I Chr.17: 38853041-38853041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1029170067 Chr.17: 38853319-38853319 on Build GRCh38
rs977734528 Chr.17: 38853154-38853154 on Build GRCh38
rs748407262 Chr.17: 38853176-38853177 on Build GRCh38
rs371656936 Chr.17: 38852991-38852991 on Build GRCh38
rs780663707 Chr.17: 38853061-38853061 on Build GRCh38
rs774851297 Chr.17: 38852993-38852993 on Build GRCh38
rs187374217 Chr.17: 38853204-38853204 on Build GRCh38
rs779749441 Chr.17: 38853149-38853149 on Build GRCh38
rs745351161 Chr.17: 38852953-38852953 on Build GRCh38
rs966100608 Chr.17: 38853303-38853303 on Build GRCh38
rs1030668316 Chr.17: 38853293-38853293 on Build GRCh38
rs371838445 Chr.17: 38853012-38853012 on Build GRCh38
rs143537161 Chr.17: 38853057-38853057 on Build GRCh38
rs867947497 Chr.17: 38853269-38853269 on Build GRCh38
rs756964133 Chr.17: 38853139-38853139 on Build GRCh38
rs796774396 Chr.17: 38853281-38853281 on Build GRCh38
rs141792721 Chr.17: 38853291-38853291 on Build GRCh38
rs112538197 Chr.17: 38853020-38853020 on Build GRCh38
rs772367688 Chr.17: 38853110-38853110 on Build GRCh38
rs928466778 Chr.17: 38853207-38853207 on Build GRCh38
rs769979297 Chr.17: 38853181-38853181 on Build GRCh38
rs749869874 Chr.17: 38853069-38853069 on Build GRCh38
rs538144057 Chr.17: 38853009-38853009 on Build GRCh38
rs921576707 Chr.17: 38853077-38853077 on Build GRCh38
rs763414359 Chr.17: 38852976-38852976 on Build GRCh38
rs767633241 Chr.17: 38853198-38853198 on Build GRCh38
rs770356733 Chr.17: 38852961-38852961 on Build GRCh38
rs34920345 Chr.17: 38853282-38853283 on Build GRCh38
rs546393811 Chr.17: 38853140-38853140 on Build GRCh38
rs774870348 Chr.17: 38852978-38852978 on Build GRCh38
rs976275089 Chr.17: 38853188-38853188 on Build GRCh38
rs759987104 Chr.17: 38853117-38853117 on Build GRCh38
rs932148095 Chr.17: 38853202-38853202 on Build GRCh38
rs769254792 Chr.17: 38853075-38853075 on Build GRCh38
rs749133337 Chr.17: 38853339-38853339 on Build GRCh38
rs34227215 Chr.17: 38853245-38853246 on Build GRCh38
rs765081769 Chr.17: 38853388-38853388 on Build GRCh38
rs751381482 Chr.17: 38853123-38853123 on Build GRCh38
rs777615381 Chr.17: 38853029-38853029 on Build GRCh38
rs376234763 Chr.17: 38853119-38853119 on Build GRCh38
rs780797285 Chr.17: 38852952-38852952 on Build GRCh38
rs745417060 Chr.17: 38853064-38853064 on Build GRCh38
rs909167827 Chr.17: 38853148-38853148 on Build GRCh38
rs1034318357 Chr.17: 38852972-38852972 on Build GRCh38
rs773238508 Chr.17: 38853113-38853113 on Build GRCh38
rs761023930 Chr.17: 38853221-38853221 on Build GRCh38
rs752569951 Chr.17: 38853023-38853023 on Build GRCh38
rs751328610 Chr.17: 38853039-38853039 on Build GRCh38
rs755779594 Chr.17: 38853146-38853146 on Build GRCh38
rs1055909018 Chr.17: 38853232-38853232 on Build GRCh38
rs764206144 Chr.17: 38853222-38853222 on Build GRCh38
rs115701245 Chr.17: 38853208-38853208 on Build GRCh38
rs769121390 Chr.17: 38853068-38853068 on Build GRCh38
rs755564906 Chr.17: 38853156-38853156 on Build GRCh38
rs940977796 Chr.17: 38853215-38853216 on Build GRCh38
rs1021913440 Chr.17: 38852996-38852996 on Build GRCh38
rs191010322 Chr.17: 38853373-38853373 on Build GRCh38
rs753019421 Chr.17: 38853144-38853145 on Build GRCh38
rs879470503 Chr.17: 38853179-38853180 on Build GRCh38
rs940519225 Chr.17: 38853182-38853182 on Build GRCh38
rs748782781 Chr.17: 38853150-38853150 on Build GRCh38
rs985029054 Chr.17: 38853206-38853206 on Build GRCh38
rs563160861 Chr.17: 38853270-38853270 on Build GRCh38
rs760818652 Chr.17: 38853115-38853115 on Build GRCh38
rs180973711 Chr.17: 38853386-38853386 on Build GRCh38
rs368570213 Chr.17: 38853011-38853011 on Build GRCh38
rs894950470 Chr.17: 38853280-38853280 on Build GRCh38
rs116204671 Chr.17: 38853332-38853332 on Build GRCh38
rs1011617449 Chr.17: 38853282-38853282 on Build GRCh38
rs1019749571 Chr.17: 38853336-38853336 on Build GRCh38
rs367956826 Chr.17: 38853084-38853084 on Build GRCh38
rs903758776 Chr.17: 38853287-38853287 on Build GRCh38
rs182421097 Chr.17: 38852975-38852975 on Build GRCh38
rs74889814 Chr.17: 38853310-38853310 on Build GRCh38
rs977656573 Chr.17: 38853016-38853016 on Build GRCh38
rs767205327 Chr.17: 38853141-38853141 on Build GRCh38
rs965777034 Chr.17: 38853179-38853179 on Build GRCh38
rs909421278 Chr.17: 38853352-38853352 on Build GRCh38
rs965946294 Chr.17: 38853349-38853349 on Build GRCh38
rs770570176 Chr.17: 38852949-38852949 on Build GRCh38
rs767610446 Chr.17: 38853000-38853000 on Build GRCh38
rs763999661 Chr.17: 38853120-38853120 on Build GRCh38
rs34160491 Chr.17: 38853168-38853169 on Build GRCh38
rs374599622 Chr.17: 38853157-38853157 on Build GRCh38
rs531947694 Chr.17: 38853014-38853014 on Build GRCh38

View Full Product Details