Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 10927523-10928024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
YIPF2
Entrez Gene ID
78992
Gene Name
Yip1 domain family member 2
Gene Aliases
FinGER2
UniGene
Hs.164026
Gene Chromosome Location
Chr.19: 10922185-10929012 on Build GRCh38
Transcripts
NM_001321439.1 NM_001321440.1 NM_024029.4 XM_011528270.2 XM_011528271.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3796584 c.264C>T p.D88D Chr.19: 10927645-10927645 on Build GRCh38
1390238 c.111A>G p.Q37Q Chr.19: 10927880-10927880 on Build GRCh38
6447848 c.231C>T p.F77F Chr.19: 10927678-10927678 on Build GRCh38
3422391 c.86G>C p.S29T Chr.19: 10927905-10927905 on Build GRCh38
3672724 c.225G>T p.P75P Chr.19: 10927684-10927684 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774497445 Chr.19: 10927908-10927908 on Build GRCh38
rs762394442 Chr.19: 10927755-10927755 on Build GRCh38
rs751777054 Chr.19: 10927799-10927799 on Build GRCh38
rs143382661 Chr.19: 10927919-10927919 on Build GRCh38
rs549120826 Chr.19: 10927874-10927874 on Build GRCh38
rs201902152 Chr.19: 10927729-10927729 on Build GRCh38
rs766276992 Chr.19: 10927722-10927722 on Build GRCh38
rs750728770 Chr.19: 10927710-10927710 on Build GRCh38
rs761275425 Chr.19: 10927674-10927674 on Build GRCh38
rs561710353 Chr.19: 10927691-10927691 on Build GRCh38
rs899850047 Chr.19: 10927942-10927942 on Build GRCh38
rs770531545 Chr.19: 10927741-10927741 on Build GRCh38
rs568855346 Chr.19: 10927996-10927996 on Build GRCh38
rs370490313 Chr.19: 10927585-10927585 on Build GRCh38
rs764788939 Chr.19: 10927988-10927988 on Build GRCh38
rs760629274 Chr.19: 10927928-10927928 on Build GRCh38
rs747663909 Chr.19: 10927624-10927624 on Build GRCh38
rs563349006 Chr.19: 10927737-10927737 on Build GRCh38
rs749423544 Chr.19: 10927898-10927898 on Build GRCh38
rs748471843 Chr.19: 10927998-10927998 on Build GRCh38
rs775650745 Chr.19: 10927661-10927661 on Build GRCh38
rs899993351 Chr.19: 10927637-10927637 on Build GRCh38
rs539633883 Chr.19: 10927556-10927556 on Build GRCh38
rs763443041 Chr.19: 10927830-10927830 on Build GRCh38
rs772916796 Chr.19: 10927643-10927643 on Build GRCh38
rs754784751 Chr.19: 10927727-10927727 on Build GRCh38
rs775225909 Chr.19: 10927921-10927921 on Build GRCh38
rs372511424 Chr.19: 10927859-10927859 on Build GRCh38
rs917594741 Chr.19: 10927839-10927839 on Build GRCh38
rs752591720 Chr.19: 10927818-10927818 on Build GRCh38
rs748944970 Chr.19: 10927739-10927739 on Build GRCh38
rs866221107 Chr.19: 10927547-10927547 on Build GRCh38
rs773355112 Chr.19: 10927766-10927766 on Build GRCh38
rs528380469 Chr.19: 10927829-10927829 on Build GRCh38
rs763205677 Chr.19: 10927717-10927717 on Build GRCh38
rs766562986 Chr.19: 10927790-10927790 on Build GRCh38
rs137987403 Chr.19: 10927851-10927851 on Build GRCh38
rs758332601 Chr.19: 10927589-10927589 on Build GRCh38
rs750640809 Chr.19: 10927588-10927588 on Build GRCh38
rs367776422 Chr.19: 10927705-10927705 on Build GRCh38
rs530775371 Chr.19: 10927745-10927745 on Build GRCh38
rs975469763 Chr.19: 10927683-10927683 on Build GRCh38
rs201625119 Chr.19: 10927582-10927582 on Build GRCh38
rs779980034 Chr.19: 10927600-10927600 on Build GRCh38
rs1027380347 Chr.19: 10927936-10927936 on Build GRCh38
rs759054672 Chr.19: 10927723-10927723 on Build GRCh38
rs374758020 Chr.19: 10927608-10927608 on Build GRCh38
rs199654122 Chr.19: 10927728-10927728 on Build GRCh38
rs1052684867 Chr.19: 10927889-10927889 on Build GRCh38
rs550690440 Chr.19: 10927989-10927989 on Build GRCh38
rs374465630 Chr.19: 10927645-10927645 on Build GRCh38
rs777171737 Chr.19: 10927701-10927701 on Build GRCh38
rs777296949 Chr.19: 10927828-10927828 on Build GRCh38
rs747381992 Chr.19: 10927746-10927746 on Build GRCh38
rs758410119 Chr.19: 10927686-10927688 on Build GRCh38
rs771041294 Chr.19: 10927903-10927903 on Build GRCh38
rs774993759 Chr.19: 10927764-10927764 on Build GRCh38
rs759746896 Chr.19: 10927913-10927913 on Build GRCh38
rs903693646 Chr.19: 10927587-10927587 on Build GRCh38
rs781194632 Chr.19: 10927698-10927699 on Build GRCh38
rs892270191 Chr.19: 10927906-10927906 on Build GRCh38
rs200663379 Chr.19: 10927801-10927801 on Build GRCh38
rs769028170 Chr.19: 10927544-10927544 on Build GRCh38
rs768956125 Chr.19: 10927854-10927854 on Build GRCh38
rs757998375 Chr.19: 10927994-10927994 on Build GRCh38
rs755979844 Chr.19: 10927820-10927820 on Build GRCh38
rs370185248 Chr.19: 10927598-10927598 on Build GRCh38
rs1014583749 Chr.19: 10927740-10927740 on Build GRCh38
rs746099189 Chr.19: 10927935-10927937 on Build GRCh38
rs763907477 Chr.19: 10927685-10927686 on Build GRCh38
rs371411035 Chr.19: 10927807-10927807 on Build GRCh38
rs1019586568 Chr.19: 10927667-10927667 on Build GRCh38
rs756827634 Chr.19: 10927980-10927980 on Build GRCh38
rs200242606 Chr.19: 10927616-10927616 on Build GRCh38
rs756290255 Chr.19: 10927988-10927989 on Build GRCh38
rs963990129 Chr.19: 10927682-10927682 on Build GRCh38
rs773837355 Chr.19: 10927891-10927891 on Build GRCh38
rs757352556 Chr.19: 10927686-10927694 on Build GRCh38
rs763296502 Chr.19: 10927779-10927779 on Build GRCh38
rs761838015 Chr.19: 10927704-10927704 on Build GRCh38
rs777070541 Chr.19: 10927750-10927750 on Build GRCh38
rs769032823 Chr.19: 10927747-10927747 on Build GRCh38
rs754834978 Chr.19: 10927611-10927611 on Build GRCh38
rs558474175 Chr.19: 10927560-10927560 on Build GRCh38
rs947667908 Chr.19: 10927842-10927842 on Build GRCh38
rs552085351 Chr.19: 10927760-10927760 on Build GRCh38
rs748665976 Chr.19: 10927873-10927873 on Build GRCh38
rs753741583 Chr.19: 10927973-10927973 on Build GRCh38
rs372330459 Chr.19: 10927636-10927636 on Build GRCh38
rs200178760 Chr.19: 10927566-10927566 on Build GRCh38
rs764066952 Chr.19: 10927954-10927954 on Build GRCh38
rs767331965 Chr.19: 10927803-10927803 on Build GRCh38
rs903599167 Chr.19: 10927856-10927856 on Build GRCh38

View Full Product Details