Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 35666734-35667241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UPK1A
Entrez Gene ID
11045
Gene Name
uroplakin 1A
Gene Aliases
TSPAN21, UP1A, UPIA, UPKA
UniGene
Hs.159309
Gene Chromosome Location
Chr.19: 35666127-35678485 on Build GRCh38
Transcripts
NM_001281443.1 NM_007000.3 XM_017026225.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3756673 c.26C>T p.A9V Chr.19: 35666838-35666838 on Build GRCh38
1304446 c.38C>T p.S13F Chr.19: 35666850-35666850 on Build GRCh38
6817651 c.27C>T p.A9A Chr.19: 35666839-35666839 on Build GRCh38
1392915 c.6G>A p.A2A Chr.19: 35666818-35666818 on Build GRCh38
3198311 c.5C>T p.A2V Chr.19: 35666817-35666817 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759565315 Chr.19: 35666833-35666833 on Build GRCh38
rs764736278 Chr.19: 35666902-35666902 on Build GRCh38
rs375733481 Chr.19: 35666861-35666861 on Build GRCh38
rs371411157 Chr.19: 35666846-35666846 on Build GRCh38
rs200550573 Chr.19: 35666912-35666912 on Build GRCh38
rs202068352 Chr.19: 35666860-35666860 on Build GRCh38
rs779573409 Chr.19: 35667080-35667081 on Build GRCh38
rs772358911 Chr.19: 35666818-35666818 on Build GRCh38
rs143562593 Chr.19: 35667184-35667184 on Build GRCh38
rs781595526 Chr.19: 35666787-35666787 on Build GRCh38
rs912071015 Chr.19: 35666994-35666994 on Build GRCh38
rs111334074 Chr.19: 35667119-35667119 on Build GRCh38
rs946335153 Chr.19: 35667011-35667011 on Build GRCh38
rs1022181804 Chr.19: 35666819-35666819 on Build GRCh38
rs373014974 Chr.19: 35666910-35666910 on Build GRCh38
rs553637067 Chr.19: 35666903-35666903 on Build GRCh38
rs1052170175 Chr.19: 35666997-35666997 on Build GRCh38
rs533602270 Chr.19: 35666824-35666824 on Build GRCh38
rs758997258 Chr.19: 35666883-35666883 on Build GRCh38
rs561171344 Chr.19: 35667045-35667045 on Build GRCh38
rs375310334 Chr.19: 35666775-35666775 on Build GRCh38
rs756397617 Chr.19: 35666839-35666839 on Build GRCh38
rs145078164 Chr.19: 35666762-35666762 on Build GRCh38
rs1002559116 Chr.19: 35667173-35667173 on Build GRCh38
rs374293776 Chr.19: 35666940-35666940 on Build GRCh38
rs772484208 Chr.19: 35666858-35666858 on Build GRCh38
rs749870495 Chr.19: 35666808-35666808 on Build GRCh38
rs748635844 Chr.19: 35666853-35666853 on Build GRCh38
rs200335323 Chr.19: 35666830-35666830 on Build GRCh38
rs776296013 Chr.19: 35666866-35666866 on Build GRCh38
rs751364943 Chr.19: 35666909-35666909 on Build GRCh38
rs575367949 Chr.19: 35666944-35666944 on Build GRCh38
rs200117343 Chr.19: 35666924-35666924 on Build GRCh38
rs866681434 Chr.19: 35667022-35667022 on Build GRCh38
rs905120856 Chr.19: 35667163-35667163 on Build GRCh38
rs370994769 Chr.19: 35667030-35667030 on Build GRCh38
rs1007169394 Chr.19: 35666761-35666761 on Build GRCh38
rs552372624 Chr.19: 35666804-35666804 on Build GRCh38
rs148884350 Chr.19: 35667182-35667182 on Build GRCh38
rs772868690 Chr.19: 35666820-35666820 on Build GRCh38
rs746705155 Chr.19: 35666862-35666862 on Build GRCh38
rs367904056 Chr.19: 35666817-35666817 on Build GRCh38
rs774971270 Chr.19: 35666885-35666887 on Build GRCh38
rs1014824932 Chr.19: 35667032-35667047 on Build GRCh38
rs769010885 Chr.19: 35666810-35666810 on Build GRCh38
rs374611091 Chr.19: 35666794-35666794 on Build GRCh38
rs770002659 Chr.19: 35666799-35666799 on Build GRCh38
rs993363210 Chr.19: 35666856-35666856 on Build GRCh38
rs199645666 Chr.19: 35666837-35666837 on Build GRCh38
rs774874919 Chr.19: 35666811-35666811 on Build GRCh38
rs185751948 Chr.19: 35666906-35666906 on Build GRCh38
rs935185177 Chr.19: 35666971-35666971 on Build GRCh38
rs138472065 Chr.19: 35666823-35666823 on Build GRCh38
rs1025660962 Chr.19: 35667164-35667164 on Build GRCh38
rs944470260 Chr.19: 35666995-35666995 on Build GRCh38
rs544670673 Chr.19: 35666961-35666961 on Build GRCh38
rs762892355 Chr.19: 35666967-35666968 on Build GRCh38
rs780280536 Chr.19: 35666982-35666982 on Build GRCh38
rs544024742 Chr.19: 35667175-35667175 on Build GRCh38
rs768768667 Chr.19: 35667217-35667217 on Build GRCh38
rs138580482 Chr.19: 35667063-35667063 on Build GRCh38
rs755030100 Chr.19: 35666941-35666941 on Build GRCh38
rs762966896 Chr.19: 35666905-35666905 on Build GRCh38
rs61741212 Chr.19: 35666838-35666838 on Build GRCh38
rs780220744 Chr.19: 35666840-35666840 on Build GRCh38
rs373513519 Chr.19: 35666832-35666832 on Build GRCh38
rs888805434 Chr.19: 35667059-35667059 on Build GRCh38
rs969449370 Chr.19: 35667214-35667214 on Build GRCh38
rs770549976 Chr.19: 35666863-35666863 on Build GRCh38
rs932115960 Chr.19: 35666967-35666967 on Build GRCh38
rs962069590 Chr.19: 35666872-35666872 on Build GRCh38
rs560418486 Chr.19: 35667127-35667127 on Build GRCh38
rs1041335645 Chr.19: 35667071-35667071 on Build GRCh38
rs1050604633 Chr.19: 35666988-35666988 on Build GRCh38
rs538839986 Chr.19: 35666769-35666769 on Build GRCh38
rs752409965 Chr.19: 35666776-35666776 on Build GRCh38
rs768744929 Chr.19: 35666850-35666850 on Build GRCh38
rs766544281 Chr.19: 35666923-35666923 on Build GRCh38

View Full Product Details