Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38409841-38410378 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASGRP4
Entrez Gene ID
115727
Gene Name
RAS guanyl releasing protein 4
Gene Aliases
-
UniGene
Hs.130434
Gene Chromosome Location
Chr.19: 38409055-38426305 on Build GRCh38
Transcripts
NM_001146202.1 NM_001146203.1 NM_001146204.1 NM_001146205.1 NM_001146206.1 NM_001146207.1 NM_170604.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7191558 c.1996A>T p.T666S Chr.19: 38410066-38410066 on Build GRCh38
21866 c.1969C>T p.P657S Chr.19: 38410093-38410093 on Build GRCh38
6023121 c.1974C>G p.C658W Chr.19: 38410088-38410088 on Build GRCh38
3388950 c.1976C>T p.P659L Chr.19: 38410086-38410086 on Build GRCh38
7191557 c.1996A>T p.T666S Chr.19: 38410066-38410066 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs896507335 Chr.19: 38410201-38410201 on Build GRCh38
rs999653027 Chr.19: 38409980-38409980 on Build GRCh38
rs186770690 Chr.19: 38410252-38410252 on Build GRCh38
rs1055747338 Chr.19: 38409893-38409893 on Build GRCh38
rs527898566 Chr.19: 38410160-38410160 on Build GRCh38
rs143241039 Chr.19: 38410353-38410353 on Build GRCh38
rs775930746 Chr.19: 38410275-38410275 on Build GRCh38
rs569997501 Chr.19: 38410029-38410029 on Build GRCh38
rs772395265 Chr.19: 38410162-38410162 on Build GRCh38
rs751644039 Chr.19: 38410014-38410014 on Build GRCh38
rs1016537118 Chr.19: 38410326-38410326 on Build GRCh38
rs746396484 Chr.19: 38410079-38410079 on Build GRCh38
rs771612485 Chr.19: 38410071-38410071 on Build GRCh38
rs370999460 Chr.19: 38410138-38410138 on Build GRCh38
rs1028624147 Chr.19: 38410301-38410301 on Build GRCh38
rs577036360 Chr.19: 38410291-38410291 on Build GRCh38
rs1036306677 Chr.19: 38410348-38410348 on Build GRCh38
rs968213778 Chr.19: 38409941-38409941 on Build GRCh38
rs1017344173 Chr.19: 38409948-38409948 on Build GRCh38
rs775534220 Chr.19: 38410001-38410001 on Build GRCh38
rs377701983 Chr.19: 38410086-38410086 on Build GRCh38
rs753657118 Chr.19: 38410143-38410143 on Build GRCh38
rs770325724 Chr.19: 38410228-38410228 on Build GRCh38
rs903690439 Chr.19: 38409961-38409961 on Build GRCh38
rs553564676 Chr.19: 38410112-38410112 on Build GRCh38
rs767723934 Chr.19: 38410124-38410124 on Build GRCh38
rs763239978 Chr.19: 38410005-38410005 on Build GRCh38
rs530924409 Chr.19: 38410048-38410048 on Build GRCh38
rs182815429 Chr.19: 38410223-38410223 on Build GRCh38
rs761862766 Chr.19: 38410028-38410028 on Build GRCh38
rs754798166 Chr.19: 38410146-38410146 on Build GRCh38
rs565525070 Chr.19: 38410137-38410137 on Build GRCh38
rs530406122 Chr.19: 38409981-38409981 on Build GRCh38
rs1056524655 Chr.19: 38410177-38410177 on Build GRCh38
rs1021150258 Chr.19: 38409934-38409934 on Build GRCh38
rs1043499267 Chr.19: 38409919-38409919 on Build GRCh38
rs369216237 Chr.19: 38409992-38409992 on Build GRCh38
rs768916629 Chr.19: 38410092-38410092 on Build GRCh38
rs755920174 Chr.19: 38410032-38410032 on Build GRCh38
rs979993543 Chr.19: 38410318-38410318 on Build GRCh38
rs964440269 Chr.19: 38409976-38409977 on Build GRCh38
rs773240089 Chr.19: 38410125-38410125 on Build GRCh38
rs1021204286 Chr.19: 38410215-38410215 on Build GRCh38
rs541648612 Chr.19: 38409899-38409899 on Build GRCh38
rs191452942 Chr.19: 38409935-38409935 on Build GRCh38
rs947937386 Chr.19: 38410214-38410214 on Build GRCh38
rs374894240 Chr.19: 38410044-38410044 on Build GRCh38
rs935159107 Chr.19: 38410256-38410256 on Build GRCh38
rs867282636 Chr.19: 38410273-38410273 on Build GRCh38
rs909126138 Chr.19: 38410111-38410111 on Build GRCh38
rs781649595 Chr.19: 38410076-38410076 on Build GRCh38
rs559959115 Chr.19: 38409903-38409903 on Build GRCh38
rs778489895 Chr.19: 38410053-38410053 on Build GRCh38
rs762378587 Chr.19: 38410149-38410149 on Build GRCh38
rs373991533 Chr.19: 38410161-38410161 on Build GRCh38
rs938716136 Chr.19: 38409881-38409881 on Build GRCh38
rs530450268 Chr.19: 38409904-38409904 on Build GRCh38
rs991445079 Chr.19: 38410168-38410168 on Build GRCh38
rs575997362 Chr.19: 38410217-38410217 on Build GRCh38
rs141622048 Chr.19: 38410088-38410088 on Build GRCh38
rs370177675 Chr.19: 38410066-38410066 on Build GRCh38
rs979569521 Chr.19: 38409943-38409943 on Build GRCh38
rs77113475 Chr.19: 38410308-38410308 on Build GRCh38
rs766617529 Chr.19: 38410309-38410309 on Build GRCh38
rs552244915 Chr.19: 38410077-38410077 on Build GRCh38
rs1010247611 Chr.19: 38409933-38409933 on Build GRCh38
rs371538688 Chr.19: 38410274-38410274 on Build GRCh38
rs1043659580 Chr.19: 38410230-38410230 on Build GRCh38
rs114190185 Chr.19: 38410338-38410338 on Build GRCh38
rs750431942 Chr.19: 38410030-38410030 on Build GRCh38
rs74387292 Chr.19: 38410307-38410307 on Build GRCh38
rs1009552995 Chr.19: 38410209-38410209 on Build GRCh38
rs754676327 Chr.19: 38410052-38410052 on Build GRCh38
rs972246349 Chr.19: 38410347-38410347 on Build GRCh38
rs761959276 Chr.19: 38410118-38410118 on Build GRCh38
rs915912789 Chr.19: 38410184-38410184 on Build GRCh38
rs373183766 Chr.19: 38410012-38410012 on Build GRCh38
rs373491454 Chr.19: 38410085-38410085 on Build GRCh38
rs903661290 Chr.19: 38410240-38410240 on Build GRCh38
rs747047014 Chr.19: 38410070-38410072 on Build GRCh38
rs115241994 Chr.19: 38410290-38410290 on Build GRCh38
rs964877291 Chr.19: 38410267-38410267 on Build GRCh38
rs1052726324 Chr.19: 38410282-38410282 on Build GRCh38
rs757799904 Chr.19: 38410018-38410018 on Build GRCh38

View Full Product Details