Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 38889601-38890100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SIRT2
Entrez Gene ID
22933
Gene Name
sirtuin 2
Gene Aliases
SIR2, SIR2L, SIR2L2
UniGene
Hs.466693
Gene Chromosome Location
Chr.19: 38878555-38899862 on Build GRCh38
Transcripts
NM_001193286.1 NM_012237.3 NM_030593.2 NR_034146.1 XM_006723111.1 XM_011526654.1 XM_011526655.1 XM_017026500.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4485748 c.296C>T p.P99L Chr.19: 38889934-38889934 on Build GRCh38
3223136 c.389C>T p.P130L Chr.19: 38889732-38889732 on Build GRCh38
996184 c.383C>T p.P128L Chr.19: 38889738-38889738 on Build GRCh38
996183 c.419C>A p.P140H Chr.19: 38889702-38889702 on Build GRCh38
7389981 c.358G>C p.E120Q Chr.19: 38889872-38889872 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374519216 Chr.19: 38889714-38889714 on Build GRCh38
rs768882921 Chr.19: 38889811-38889811 on Build GRCh38
rs750693215 Chr.19: 38889805-38889805 on Build GRCh38
rs780421129 Chr.19: 38889892-38889892 on Build GRCh38
rs776194765 Chr.19: 38889845-38889845 on Build GRCh38
rs143889239 Chr.19: 38889718-38889718 on Build GRCh38
rs748298105 Chr.19: 38889919-38889919 on Build GRCh38
rs761744596 Chr.19: 38889683-38889683 on Build GRCh38
rs202062280 Chr.19: 38890075-38890075 on Build GRCh38
rs757149967 Chr.19: 38889999-38889999 on Build GRCh38
rs201674443 Chr.19: 38889841-38889841 on Build GRCh38
rs200654695 Chr.19: 38889661-38889661 on Build GRCh38
rs562679361 Chr.19: 38890049-38890049 on Build GRCh38
rs752989664 Chr.19: 38890074-38890074 on Build GRCh38
rs762318031 Chr.19: 38889864-38889864 on Build GRCh38
rs764061956 Chr.19: 38889665-38889665 on Build GRCh38
rs1019519021 Chr.19: 38889637-38889637 on Build GRCh38
rs201226287 Chr.19: 38889819-38889819 on Build GRCh38
rs767454073 Chr.19: 38889758-38889758 on Build GRCh38
rs376377803 Chr.19: 38889831-38889831 on Build GRCh38
rs202102362 Chr.19: 38889791-38889791 on Build GRCh38
rs760390403 Chr.19: 38889779-38889779 on Build GRCh38
rs991561866 Chr.19: 38889816-38889816 on Build GRCh38
rs76265208 Chr.19: 38889926-38889926 on Build GRCh38
rs200279657 Chr.19: 38889737-38889737 on Build GRCh38
rs374491281 Chr.19: 38889940-38889940 on Build GRCh38
rs199610453 Chr.19: 38889870-38889870 on Build GRCh38
rs761795880 Chr.19: 38889752-38889752 on Build GRCh38
rs1028588523 Chr.19: 38889674-38889674 on Build GRCh38
rs949405140 Chr.19: 38889906-38889906 on Build GRCh38
rs779149028 Chr.19: 38889911-38889911 on Build GRCh38
rs572446780 Chr.19: 38889797-38889797 on Build GRCh38
rs199533948 Chr.19: 38889988-38889988 on Build GRCh38
rs764704680 Chr.19: 38889787-38889787 on Build GRCh38
rs764756007 Chr.19: 38889854-38889854 on Build GRCh38
rs747483458 Chr.19: 38889719-38889719 on Build GRCh38
rs369519515 Chr.19: 38890067-38890067 on Build GRCh38
rs990901201 Chr.19: 38890070-38890070 on Build GRCh38
rs780975758 Chr.19: 38890002-38890002 on Build GRCh38
rs200544410 Chr.19: 38889983-38889983 on Build GRCh38
rs372171025 Chr.19: 38890009-38890009 on Build GRCh38
rs199959727 Chr.19: 38889995-38889995 on Build GRCh38
rs756483984 Chr.19: 38889808-38889808 on Build GRCh38
rs202073777 Chr.19: 38889663-38889663 on Build GRCh38
rs202027151 Chr.19: 38890071-38890071 on Build GRCh38
rs768087087 Chr.19: 38890011-38890011 on Build GRCh38
rs200159438 Chr.19: 38889898-38889898 on Build GRCh38
rs1006649779 Chr.19: 38889948-38889948 on Build GRCh38
rs201717531 Chr.19: 38889960-38889960 on Build GRCh38
rs200366538 Chr.19: 38889858-38889858 on Build GRCh38
rs200307334 Chr.19: 38889886-38889886 on Build GRCh38
rs757719196 Chr.19: 38889796-38889796 on Build GRCh38
rs765920385 Chr.19: 38889707-38889707 on Build GRCh38
rs371291429 Chr.19: 38889970-38889970 on Build GRCh38
rs375447974 Chr.19: 38890053-38890053 on Build GRCh38
rs759138961 Chr.19: 38889848-38889848 on Build GRCh38
rs199693194 Chr.19: 38889927-38889927 on Build GRCh38
rs755684272 Chr.19: 38889702-38889702 on Build GRCh38
rs200424464 Chr.19: 38889823-38889823 on Build GRCh38
rs757666044 Chr.19: 38889723-38889723 on Build GRCh38
rs772817974 Chr.19: 38889772-38889772 on Build GRCh38
rs199856206 Chr.19: 38889738-38889738 on Build GRCh38
rs761102526 Chr.19: 38890058-38890058 on Build GRCh38
rs750115754 Chr.19: 38889689-38889689 on Build GRCh38
rs34700059 Chr.19: 38889981-38889982 on Build GRCh38
rs551071804 Chr.19: 38889908-38889908 on Build GRCh38
rs180706698 Chr.19: 38889664-38889664 on Build GRCh38
rs776877877 Chr.19: 38889651-38889651 on Build GRCh38
rs748926052 Chr.19: 38890010-38890010 on Build GRCh38
rs749503258 Chr.19: 38889810-38889810 on Build GRCh38
rs767053214 Chr.19: 38890060-38890060 on Build GRCh38
rs769376141 Chr.19: 38889941-38889941 on Build GRCh38
rs750933845 Chr.19: 38889879-38889879 on Build GRCh38
rs998701862 Chr.19: 38889623-38889623 on Build GRCh38
rs755382259 Chr.19: 38889971-38889971 on Build GRCh38
rs934465189 Chr.19: 38889840-38889840 on Build GRCh38
rs190524849 Chr.19: 38889792-38889792 on Build GRCh38
rs769837989 Chr.19: 38889725-38889725 on Build GRCh38
rs1029032321 Chr.19: 38889692-38889692 on Build GRCh38
rs766005923 Chr.19: 38889785-38889785 on Build GRCh38
rs200971034 Chr.19: 38889830-38889830 on Build GRCh38
rs774413721 Chr.19: 38889682-38889682 on Build GRCh38
rs776368661 Chr.19: 38889935-38889935 on Build GRCh38
rs769569029 Chr.19: 38890007-38890007 on Build GRCh38
rs959080832 Chr.19: 38890015-38890015 on Build GRCh38
rs769937497 Chr.19: 38889656-38889656 on Build GRCh38
rs763097606 Chr.19: 38889824-38889824 on Build GRCh38
rs991722119 Chr.19: 38890037-38890037 on Build GRCh38
rs749382036 Chr.19: 38889730-38889730 on Build GRCh38
rs770654234 Chr.19: 38889931-38889931 on Build GRCh38
rs768714344 Chr.19: 38889946-38889947 on Build GRCh38
rs150309258 Chr.19: 38889724-38889724 on Build GRCh38
rs554192730 Chr.19: 38889795-38889795 on Build GRCh38
rs753231190 Chr.19: 38889712-38889712 on Build GRCh38
rs916162179 Chr.19: 38890047-38890047 on Build GRCh38
rs765623042 Chr.19: 38889975-38889975 on Build GRCh38
rs775649914 Chr.19: 38889728-38889728 on Build GRCh38
rs200814071 Chr.19: 38889959-38889959 on Build GRCh38
rs11551762 Chr.19: 38889881-38889881 on Build GRCh38
rs201156500 Chr.19: 38889865-38889865 on Build GRCh38
rs200980881 Chr.19: 38890073-38890073 on Build GRCh38
rs767401052 Chr.19: 38889688-38889688 on Build GRCh38
rs563953734 Chr.19: 38889786-38889786 on Build GRCh38
rs750920243 Chr.19: 38889961-38889961 on Build GRCh38
rs201762886 Chr.19: 38889989-38889989 on Build GRCh38
rs201311892 Chr.19: 38889762-38889762 on Build GRCh38
rs764083176 Chr.19: 38890077-38890077 on Build GRCh38
rs745718254 Chr.19: 38890006-38890006 on Build GRCh38
rs373892878 Chr.19: 38890039-38890039 on Build GRCh38
rs779099527 Chr.19: 38889815-38889815 on Build GRCh38
rs761208240 Chr.19: 38889967-38889967 on Build GRCh38

View Full Product Details