Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39515024-39515511 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SELV
Entrez Gene ID
348303
Gene Name
selenoprotein V
Gene Aliases
-
UniGene
Hs.319284
Gene Chromosome Location
Chr.19: 39515113-39520686 on Build GRCh38
Transcripts
NM_182704.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5687638 c.123C>A p.T41T Chr.19: 39515335-39515335 on Build GRCh38
7389170 c.287C>A p.P96Q Chr.19: 39515499-39515499 on Build GRCh38
4138229 c.287C>G p.P96R Chr.19: 39515499-39515499 on Build GRCh38
4646597 c.270G>T p.P90P Chr.19: 39515482-39515482 on Build GRCh38
1742013 c.232C>G p.L78V Chr.19: 39515444-39515444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs908988946 Chr.19: 39515101-39515101 on Build GRCh38
rs1018391496 Chr.19: 39515436-39515436 on Build GRCh38
rs984467551 Chr.19: 39515335-39515335 on Build GRCh38
rs77624079 Chr.19: 39515450-39515450 on Build GRCh38
rs781148764 Chr.19: 39515317-39515317 on Build GRCh38
rs868676769 Chr.19: 39515232-39515232 on Build GRCh38
rs867280500 Chr.19: 39515487-39515487 on Build GRCh38
rs758468639 Chr.19: 39515349-39515349 on Build GRCh38
rs966323275 Chr.19: 39515275-39515275 on Build GRCh38
rs765427470 Chr.19: 39515261-39515261 on Build GRCh38
rs773929555 Chr.19: 39515184-39515184 on Build GRCh38
rs957175437 Chr.19: 39515473-39515473 on Build GRCh38
rs548145137 Chr.19: 39515069-39515069 on Build GRCh38
rs942649143 Chr.19: 39515347-39515358 on Build GRCh38
rs746007504 Chr.19: 39515294-39515311 on Build GRCh38
rs763089947 Chr.19: 39515284-39515284 on Build GRCh38
rs1025477924 Chr.19: 39515402-39515402 on Build GRCh38
rs759310775 Chr.19: 39515474-39515474 on Build GRCh38
rs939554396 Chr.19: 39515421-39515421 on Build GRCh38
rs897359612 Chr.19: 39515161-39515161 on Build GRCh38
rs868047287 Chr.19: 39515325-39515325 on Build GRCh38
rs748950350 Chr.19: 39515364-39515364 on Build GRCh38
rs746961689 Chr.19: 39515118-39515118 on Build GRCh38
rs768364046 Chr.19: 39515380-39515380 on Build GRCh38
rs372238324 Chr.19: 39515072-39515072 on Build GRCh38
rs576629288 Chr.19: 39515216-39515216 on Build GRCh38
rs999575452 Chr.19: 39515461-39515461 on Build GRCh38
rs918754803 Chr.19: 39515408-39515408 on Build GRCh38
rs979686658 Chr.19: 39515245-39515245 on Build GRCh38
rs775255754 Chr.19: 39515483-39515483 on Build GRCh38
rs748862322 Chr.19: 39515173-39515173 on Build GRCh38
rs1045484794 Chr.19: 39515337-39515337 on Build GRCh38
rs949577030 Chr.19: 39515485-39515485 on Build GRCh38
rs1001445444 Chr.19: 39515111-39515112 on Build GRCh38
rs779049444 Chr.19: 39515208-39515208 on Build GRCh38
rs376284154 Chr.19: 39515295-39515295 on Build GRCh38
rs942720937 Chr.19: 39515227-39515227 on Build GRCh38
rs761098265 Chr.19: 39515206-39515206 on Build GRCh38
rs1025697377 Chr.19: 39515200-39515200 on Build GRCh38
rs759914209 Chr.19: 39515242-39515242 on Build GRCh38
rs770140977 Chr.19: 39515448-39515448 on Build GRCh38
rs921624422 Chr.19: 39515044-39515044 on Build GRCh38
rs973277019 Chr.19: 39515397-39515397 on Build GRCh38
rs536413308 Chr.19: 39515197-39515197 on Build GRCh38
rs868416059 Chr.19: 39515471-39515471 on Build GRCh38
rs541106632 Chr.19: 39515302-39515302 on Build GRCh38
rs1016821762 Chr.19: 39515378-39515378 on Build GRCh38
rs757168476 Chr.19: 39515312-39515312 on Build GRCh38
rs867877308 Chr.19: 39515183-39515183 on Build GRCh38
rs903961393 Chr.19: 39515209-39515209 on Build GRCh38
rs988386520 Chr.19: 39515474-39515474 on Build GRCh38
rs767345410 Chr.19: 39515477-39515477 on Build GRCh38
rs1056701001 Chr.19: 39515438-39515438 on Build GRCh38
rs755921170 Chr.19: 39515331-39515331 on Build GRCh38
rs925540786 Chr.19: 39515254-39515254 on Build GRCh38
rs771419379 Chr.19: 39515207-39515207 on Build GRCh38
rs768274360 Chr.19: 39515182-39515182 on Build GRCh38
rs766883662 Chr.19: 39515068-39515068 on Build GRCh38
rs868763877 Chr.19: 39515369-39515369 on Build GRCh38
rs778586906 Chr.19: 39515411-39515411 on Build GRCh38
rs755348079 Chr.19: 39515114-39515114 on Build GRCh38
rs865841510 Chr.19: 39515469-39515469 on Build GRCh38
rs565919445 Chr.19: 39515096-39515096 on Build GRCh38
rs764165889 Chr.19: 39515287-39515287 on Build GRCh38
rs543769850 Chr.19: 39515226-39515226 on Build GRCh38
rs865967755 Chr.19: 39515244-39515244 on Build GRCh38
rs375570045 Chr.19: 39515444-39515444 on Build GRCh38
rs187415319 Chr.19: 39515296-39515296 on Build GRCh38
rs747653055 Chr.19: 39515441-39515441 on Build GRCh38
rs76140888 Chr.19: 39515263-39515263 on Build GRCh38
rs375911338 Chr.19: 39515320-39515320 on Build GRCh38
rs898413439 Chr.19: 39515385-39515385 on Build GRCh38
rs777774862 Chr.19: 39515387-39515387 on Build GRCh38
rs1037220197 Chr.19: 39515146-39515146 on Build GRCh38
rs553874104 Chr.19: 39515055-39515055 on Build GRCh38
rs566071020 Chr.19: 39515424-39515424 on Build GRCh38
rs746278000 Chr.19: 39515445-39515445 on Build GRCh38
rs1012227806 Chr.19: 39515190-39515190 on Build GRCh38
rs554886244 Chr.19: 39515215-39515215 on Build GRCh38
rs372527012 Chr.19: 39515341-39515341 on Build GRCh38
rs760688919 Chr.19: 39515491-39515491 on Build GRCh38

View Full Product Details