Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 19765621-19766121 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TBX1
Entrez Gene ID
6899
Gene Name
T-box 1
Gene Aliases
CAFS, CATCH22, CTHM, DGCR, DGS, DORV, TBX1C, TGA, VCF, VCFS
UniGene
Hs.173984
Gene Chromosome Location
Chr.22: 19756703-19783593 on Build GRCh38
Transcripts
NM_005992.1 NM_080646.1 NM_080647.1 XM_006724312.2 XM_017028925.1 XM_017028926.1 XM_017028927.1 XM_017028928.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6241610 c.971C>A p.P324H Chr.22: 19765964-19765964 on Build GRCh38
6431802 c.956C>T p.A319V Chr.22: 19765949-19765949 on Build GRCh38
7288723 c.847C>T p.Q283* Chr.22: 19765764-19765764 on Build GRCh38
6241609 c.971C>A p.P324H Chr.22: 19765964-19765964 on Build GRCh38
4135151 c.933A>G p.A311A Chr.22: 19765926-19765926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752278002 Chr.22: 19765912-19765912 on Build GRCh38
rs1028497136 Chr.22: 19766051-19766051 on Build GRCh38
rs1004508627 Chr.22: 19765895-19765895 on Build GRCh38
rs766377918 Chr.22: 19765763-19765763 on Build GRCh38
rs763127510 Chr.22: 19765891-19765891 on Build GRCh38
rs780894151 Chr.22: 19765713-19765713 on Build GRCh38
rs921515181 Chr.22: 19765689-19765691 on Build GRCh38
rs773338175 Chr.22: 19765752-19765752 on Build GRCh38
rs900936631 Chr.22: 19766028-19766028 on Build GRCh38
rs558072690 Chr.22: 19765923-19765923 on Build GRCh38
rs920347875 Chr.22: 19765753-19765753 on Build GRCh38
rs892098892 Chr.22: 19765816-19765816 on Build GRCh38
rs550907643 Chr.22: 19765857-19765857 on Build GRCh38
rs764610884 Chr.22: 19765986-19765986 on Build GRCh38
rs778151658 Chr.22: 19766041-19766041 on Build GRCh38
rs766324089 Chr.22: 19765953-19765953 on Build GRCh38
rs200879129 Chr.22: 19765841-19765841 on Build GRCh38
rs1019354356 Chr.22: 19765849-19765849 on Build GRCh38
rs543755910 Chr.22: 19766067-19766067 on Build GRCh38
rs779209396 Chr.22: 19765838-19765838 on Build GRCh38
rs775788782 Chr.22: 19765940-19765940 on Build GRCh38
rs201448674 Chr.22: 19765710-19765710 on Build GRCh38
rs199883948 Chr.22: 19765851-19765851 on Build GRCh38
rs745370928 Chr.22: 19765717-19765717 on Build GRCh38
rs41298834 Chr.22: 19765692-19765693 on Build GRCh38
rs746457113 Chr.22: 19765738-19765739 on Build GRCh38
rs753660073 Chr.22: 19765958-19765958 on Build GRCh38
rs72646963 Chr.22: 19766085-19766085 on Build GRCh38
rs754623522 Chr.22: 19765834-19765834 on Build GRCh38
rs779659218 Chr.22: 19765818-19765818 on Build GRCh38
rs961356403 Chr.22: 19765876-19765876 on Build GRCh38
rs769708851 Chr.22: 19765931-19765931 on Build GRCh38
rs773107514 Chr.22: 19765951-19765951 on Build GRCh38
rs754644643 Chr.22: 19765903-19765903 on Build GRCh38
rs41298836 Chr.22: 19765856-19765856 on Build GRCh38
rs569704302 Chr.22: 19765878-19765878 on Build GRCh38
rs1005796787 Chr.22: 19766060-19766060 on Build GRCh38
rs765842871 Chr.22: 19765886-19765886 on Build GRCh38
rs751339103 Chr.22: 19765803-19765803 on Build GRCh38
rs775970680 Chr.22: 19765854-19765855 on Build GRCh38
rs962099082 Chr.22: 19765705-19765705 on Build GRCh38
rs578007841 Chr.22: 19765662-19765662 on Build GRCh38
rs1024698516 Chr.22: 19766091-19766091 on Build GRCh38
rs761402868 Chr.22: 19765734-19765734 on Build GRCh38
rs1014624067 Chr.22: 19765700-19765700 on Build GRCh38
rs992823960 Chr.22: 19766087-19766087 on Build GRCh38
rs112979119 Chr.22: 19766094-19766094 on Build GRCh38
rs774752069 Chr.22: 19765745-19765745 on Build GRCh38
rs992906695 Chr.22: 19765889-19765889 on Build GRCh38
rs555522870 Chr.22: 19765907-19765907 on Build GRCh38
rs748639207 Chr.22: 19765746-19765746 on Build GRCh38
rs748939569 Chr.22: 19765821-19765821 on Build GRCh38
rs772562231 Chr.22: 19765749-19765749 on Build GRCh38
rs564116728 Chr.22: 19765644-19765644 on Build GRCh38
rs767553992 Chr.22: 19765809-19765809 on Build GRCh38
rs755891567 Chr.22: 19765723-19765723 on Build GRCh38
rs748928162 Chr.22: 19765736-19765736 on Build GRCh38
rs13054135 Chr.22: 19766075-19766075 on Build GRCh38
rs747565599 Chr.22: 19765922-19765922 on Build GRCh38
rs41298840 Chr.22: 19765926-19765926 on Build GRCh38
rs535210581 Chr.22: 19765897-19765897 on Build GRCh38
rs757196973 Chr.22: 19765711-19765711 on Build GRCh38
rs763843987 Chr.22: 19765793-19765793 on Build GRCh38
rs1009360453 Chr.22: 19765842-19765842 on Build GRCh38
rs776751131 Chr.22: 19765774-19765774 on Build GRCh38
rs529375396 Chr.22: 19765695-19765695 on Build GRCh38
rs760755800 Chr.22: 19765760-19765760 on Build GRCh38
rs761716175 Chr.22: 19765858-19765858 on Build GRCh38
rs763231000 Chr.22: 19765942-19765942 on Build GRCh38
rs72646961 Chr.22: 19765663-19765663 on Build GRCh38
rs768822245 Chr.22: 19766031-19766031 on Build GRCh38
rs563765156 Chr.22: 19765669-19765669 on Build GRCh38
rs888573903 Chr.22: 19766058-19766058 on Build GRCh38
rs779869702 Chr.22: 19765730-19765730 on Build GRCh38
rs771023775 Chr.22: 19765853-19765853 on Build GRCh38
rs758025693 Chr.22: 19766031-19766031 on Build GRCh38
rs1056678428 Chr.22: 19765877-19765877 on Build GRCh38
rs398088161 Chr.22: 19765694-19765695 on Build GRCh38
rs397897337 Chr.22: 19765693-19765694 on Build GRCh38
rs1041218128 Chr.22: 19766007-19766007 on Build GRCh38
rs768923000 Chr.22: 19765742-19765742 on Build GRCh38
rs770044781 Chr.22: 19765842-19765843 on Build GRCh38
rs1014592830 Chr.22: 19765904-19765904 on Build GRCh38
rs745951558 Chr.22: 19765927-19765927 on Build GRCh38
rs755692915 Chr.22: 19765814-19765814 on Build GRCh38
rs201607803 Chr.22: 19765920-19765920 on Build GRCh38
rs911796187 Chr.22: 19765804-19765804 on Build GRCh38
rs753380966 Chr.22: 19765894-19765894 on Build GRCh38
rs549514380 Chr.22: 19765866-19765866 on Build GRCh38
rs772362864 Chr.22: 19765847-19765847 on Build GRCh38
rs1028884131 Chr.22: 19765852-19765852 on Build GRCh38
rs752369951 Chr.22: 19766016-19766016 on Build GRCh38
rs41298838 Chr.22: 19765921-19765921 on Build GRCh38
rs146109202 Chr.22: 19765805-19765805 on Build GRCh38
rs543545408 Chr.22: 19765668-19765668 on Build GRCh38
rs759090563 Chr.22: 19765979-19765979 on Build GRCh38
rs917326981 Chr.22: 19765891-19765891 on Build GRCh38
rs764891073 Chr.22: 19765780-19765780 on Build GRCh38
rs760664903 Chr.22: 19765872-19765872 on Build GRCh38
rs772039142 Chr.22: 19765944-19765944 on Build GRCh38
rs892172437 Chr.22: 19765969-19765969 on Build GRCh38
rs750330152 Chr.22: 19765812-19765812 on Build GRCh38

View Full Product Details