Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 76738683-76739179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SHROOM3
Entrez Gene ID
57619
Gene Name
shroom family member 3
Gene Aliases
APXL3, MSTP013, SHRM, ShrmL
UniGene
Hs.683808
Gene Chromosome Location
Chr.4: 76435100-76783253 on Build GRCh38
Transcripts
NM_020859.3 XM_005263163.4 XM_005263164.4 XM_005263165.4 XM_006714282.3 XM_011532157.2 XM_011532158.2 XM_011532159.1 XM_011532160.1 XM_011532161.2 XM_017008488.1 XM_017008489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4644619 c.725C>A p.S242Y Chr.4: 76738898-76738898 on Build GRCh38
233359 c.744G>C p.Q248H Chr.4: 76738917-76738917 on Build GRCh38
7101510 c.715C>G p.P239A Chr.4: 76738888-76738888 on Build GRCh38
4625342 c.985C>T p.L329L Chr.4: 76739158-76739158 on Build GRCh38
5481901 c.845G>A p.R282H Chr.4: 76739018-76739018 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004266045 Chr.4: 76739127-76739127 on Build GRCh38
rs769701354 Chr.4: 76739087-76739087 on Build GRCh38
rs771453964 Chr.4: 76738778-76738778 on Build GRCh38
rs760768267 Chr.4: 76739112-76739112 on Build GRCh38
rs374543213 Chr.4: 76739040-76739040 on Build GRCh38
rs776605073 Chr.4: 76739017-76739017 on Build GRCh38
rs140625853 Chr.4: 76738849-76738849 on Build GRCh38
rs374710074 Chr.4: 76738995-76738995 on Build GRCh38
rs752570250 Chr.4: 76738902-76738902 on Build GRCh38
rs764886278 Chr.4: 76739020-76739020 on Build GRCh38
rs764378677 Chr.4: 76738947-76738947 on Build GRCh38
rs1046155029 Chr.4: 76738864-76738864 on Build GRCh38
rs747696289 Chr.4: 76739000-76739000 on Build GRCh38
rs754121390 Chr.4: 76739105-76739105 on Build GRCh38
rs748998507 Chr.4: 76738858-76738858 on Build GRCh38
rs111521571 Chr.4: 76739067-76739067 on Build GRCh38
rs201586601 Chr.4: 76739091-76739091 on Build GRCh38
rs139587539 Chr.4: 76738754-76738754 on Build GRCh38
rs763313294 Chr.4: 76738946-76738946 on Build GRCh38
rs1001861897 Chr.4: 76738880-76738880 on Build GRCh38
rs377199268 Chr.4: 76738870-76738870 on Build GRCh38
rs377324397 Chr.4: 76739025-76739025 on Build GRCh38
rs760622199 Chr.4: 76738913-76738913 on Build GRCh38
rs761805227 Chr.4: 76738726-76738726 on Build GRCh38
rs759628103 Chr.4: 76738830-76738830 on Build GRCh38
rs780285364 Chr.4: 76738838-76738839 on Build GRCh38
rs202185868 Chr.4: 76739047-76739047 on Build GRCh38
rs752918223 Chr.4: 76738752-76738752 on Build GRCh38
rs777319771 Chr.4: 76739139-76739139 on Build GRCh38
rs777388129 Chr.4: 76739061-76739061 on Build GRCh38
rs142581367 Chr.4: 76739053-76739053 on Build GRCh38
rs762477288 Chr.4: 76738716-76738716 on Build GRCh38
rs773899698 Chr.4: 76738807-76738807 on Build GRCh38
rs763540413 Chr.4: 76738721-76738721 on Build GRCh38
rs754292324 Chr.4: 76739039-76739039 on Build GRCh38
rs144220275 Chr.4: 76738809-76738809 on Build GRCh38
rs147825182 Chr.4: 76738993-76738993 on Build GRCh38
rs752977945 Chr.4: 76739118-76739118 on Build GRCh38
rs773776536 Chr.4: 76738898-76738898 on Build GRCh38
rs758685837 Chr.4: 76738836-76738836 on Build GRCh38
rs746996342 Chr.4: 76739011-76739011 on Build GRCh38
rs199685557 Chr.4: 76739113-76739113 on Build GRCh38
rs891989362 Chr.4: 76738731-76738731 on Build GRCh38
rs781294814 Chr.4: 76739072-76739072 on Build GRCh38
rs775139022 Chr.4: 76739092-76739092 on Build GRCh38
rs747055218 Chr.4: 76738841-76738841 on Build GRCh38
rs149557511 Chr.4: 76738799-76738799 on Build GRCh38
rs1057208436 Chr.4: 76738725-76738725 on Build GRCh38
rs752971457 Chr.4: 76738831-76738831 on Build GRCh38
rs750480368 Chr.4: 76739031-76739031 on Build GRCh38
rs777929615 Chr.4: 76738762-76738762 on Build GRCh38
rs76406459 Chr.4: 76738805-76738805 on Build GRCh38
rs367773952 Chr.4: 76739086-76739086 on Build GRCh38
rs757246509 Chr.4: 76738848-76738848 on Build GRCh38
rs962270438 Chr.4: 76739138-76739138 on Build GRCh38
rs778647684 Chr.4: 76738999-76738999 on Build GRCh38
rs184494467 Chr.4: 76738803-76738803 on Build GRCh38
rs553897462 Chr.4: 76738812-76738812 on Build GRCh38
rs376382276 Chr.4: 76738854-76738854 on Build GRCh38
rs931394479 Chr.4: 76738973-76738973 on Build GRCh38
rs757969587 Chr.4: 76739056-76739056 on Build GRCh38
rs751633853 Chr.4: 76738845-76738845 on Build GRCh38
rs1010390029 Chr.4: 76738733-76738733 on Build GRCh38
rs149686763 Chr.4: 76738940-76738940 on Build GRCh38
rs759433745 Chr.4: 76739018-76739018 on Build GRCh38
rs776550091 Chr.4: 76738930-76738930 on Build GRCh38
rs950241062 Chr.4: 76738832-76738832 on Build GRCh38
rs371504856 Chr.4: 76739048-76739048 on Build GRCh38
rs766340002 Chr.4: 76738915-76738915 on Build GRCh38
rs747533046 Chr.4: 76738768-76738768 on Build GRCh38
rs765237474 Chr.4: 76738785-76738785 on Build GRCh38
rs932668101 Chr.4: 76738783-76738783 on Build GRCh38
rs747723693 Chr.4: 76738903-76738903 on Build GRCh38
rs765691133 Chr.4: 76739117-76739117 on Build GRCh38
rs531797026 Chr.4: 76738966-76738966 on Build GRCh38
rs750211887 Chr.4: 76738737-76738737 on Build GRCh38
rs542556063 Chr.4: 76738808-76738808 on Build GRCh38
rs965261503 Chr.4: 76738980-76738980 on Build GRCh38
rs751685681 Chr.4: 76738962-76738962 on Build GRCh38
rs976849558 Chr.4: 76738712-76738712 on Build GRCh38
rs778053278 Chr.4: 76738844-76738844 on Build GRCh38
rs755193779 Chr.4: 76738743-76738743 on Build GRCh38
rs766905572 Chr.4: 76739038-76739038 on Build GRCh38
rs771433228 Chr.4: 76738907-76738907 on Build GRCh38
rs61999292 Chr.4: 76739146-76739146 on Build GRCh38
rs560258820 Chr.4: 76739022-76739022 on Build GRCh38
rs767275356 Chr.4: 76738729-76738729 on Build GRCh38
rs758919326 Chr.4: 76738831-76738836 on Build GRCh38
rs372730468 Chr.4: 76739103-76739103 on Build GRCh38
rs1015900752 Chr.4: 76738978-76738978 on Build GRCh38
rs770153135 Chr.4: 76738853-76738853 on Build GRCh38
rs756654865 Chr.4: 76739032-76739032 on Build GRCh38
rs771615558 Chr.4: 76739008-76739008 on Build GRCh38
rs577193004 Chr.4: 76738856-76738856 on Build GRCh38
rs770966223 Chr.4: 76739012-76739012 on Build GRCh38
rs200211229 Chr.4: 76739028-76739028 on Build GRCh38
rs779306431 Chr.4: 76738751-76738751 on Build GRCh38
rs370967544 Chr.4: 76738910-76738910 on Build GRCh38
rs751009311 Chr.4: 76738988-76738988 on Build GRCh38
rs551747532 Chr.4: 76739128-76739128 on Build GRCh38
rs344140 Chr.4: 76739009-76739009 on Build GRCh38
rs139272770 Chr.4: 76738792-76738792 on Build GRCh38
rs762145571 Chr.4: 76739102-76739102 on Build GRCh38
rs766219406 Chr.4: 76738820-76738820 on Build GRCh38
rs757374651 Chr.4: 76738965-76738965 on Build GRCh38
rs201796092 Chr.4: 76738724-76738724 on Build GRCh38
rs746483211 Chr.4: 76739062-76739062 on Build GRCh38
rs554304598 Chr.4: 76738782-76738782 on Build GRCh38
rs140762344 Chr.4: 76739114-76739114 on Build GRCh38

View Full Product Details