Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 173888640-173889142 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPEB4
Entrez Gene ID
80315
Gene Name
cytoplasmic polyadenylation element binding protein 4
Gene Aliases
CPE-BP4, hCPEB-4
UniGene
Hs.127126
Gene Chromosome Location
Chr.5: 173888328-173960991 on Build GRCh38
Transcripts
NM_001308189.1 NM_001308191.1 NM_030627.3 XM_005265994.1 XM_011534660.2 XM_011534661.2

SNP Detail

SNP ID Chromosome Location
rs988015576 Chr.5: 173888814-173888814 on Build GRCh38
rs544324505 Chr.5: 173889061-173889061 on Build GRCh38
rs556072209 Chr.5: 173889086-173889086 on Build GRCh38
rs191828634 Chr.5: 173889022-173889022 on Build GRCh38
rs558172795 Chr.5: 173888937-173888937 on Build GRCh38
rs948293499 Chr.5: 173888834-173888834 on Build GRCh38
rs993435915 Chr.5: 173888926-173888926 on Build GRCh38
rs1008769140 Chr.5: 173888973-173888973 on Build GRCh38
rs573100851 Chr.5: 173888880-173888880 on Build GRCh38
rs1008986227 Chr.5: 173888986-173888986 on Build GRCh38
rs926224653 Chr.5: 173888857-173888857 on Build GRCh38
rs535757959 Chr.5: 173888831-173888831 on Build GRCh38
rs550102065 Chr.5: 173888668-173888668 on Build GRCh38
rs1026285322 Chr.5: 173888979-173888979 on Build GRCh38
rs775869639 Chr.5: 173888837-173888837 on Build GRCh38
rs1005376335 Chr.5: 173888900-173888900 on Build GRCh38
rs955556437 Chr.5: 173888894-173888925 on Build GRCh38
rs359466 Chr.5: 173888863-173888863 on Build GRCh38
rs934300650 Chr.5: 173888861-173888861 on Build GRCh38
rs145803277 Chr.5: 173888771-173888771 on Build GRCh38
rs1038238901 Chr.5: 173888903-173888903 on Build GRCh38
rs967460075 Chr.5: 173889004-173889004 on Build GRCh38
rs916959133 Chr.5: 173888864-173888864 on Build GRCh38
rs954923932 Chr.5: 173888985-173888985 on Build GRCh38
rs994668194 Chr.5: 173888894-173888895 on Build GRCh38
rs55943834 Chr.5: 173889044-173889044 on Build GRCh38
rs1016847053 Chr.5: 173888742-173888742 on Build GRCh38
rs897102438 Chr.5: 173888909-173888909 on Build GRCh38
rs1018587895 Chr.5: 173888982-173888982 on Build GRCh38
rs371717356 Chr.5: 173888997-173888997 on Build GRCh38
rs769466984 Chr.5: 173888730-173888730 on Build GRCh38
rs372309008 Chr.5: 173888739-173888739 on Build GRCh38
rs949838778 Chr.5: 173888866-173888866 on Build GRCh38
rs1044586035 Chr.5: 173888877-173888877 on Build GRCh38
rs888997017 Chr.5: 173888972-173888972 on Build GRCh38
rs1026339294 Chr.5: 173888773-173888773 on Build GRCh38
rs187831468 Chr.5: 173888858-173888858 on Build GRCh38
rs940757441 Chr.5: 173888879-173888879 on Build GRCh38
rs963339946 Chr.5: 173888769-173888769 on Build GRCh38
rs954834738 Chr.5: 173888797-173888797 on Build GRCh38
rs910728230 Chr.5: 173888826-173888826 on Build GRCh38
rs978953213 Chr.5: 173889025-173889025 on Build GRCh38

View Full Product Details