Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2366228-2366737 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3B
Entrez Gene ID
8662
Gene Name
eukaryotic translation initiation factor 3 subunit B
Gene Aliases
EIF3-ETA, EIF3-P110, EIF3-P116, EIF3S9, PRT1
UniGene
Hs.371001
Gene Chromosome Location
Chr.7: 2354540-2380745 on Build GRCh38
Transcripts
NM_001037283.1 NM_003751.3 XM_011515599.1 XM_011515600.1 XM_011515601.1 XM_017012752.1 XM_017012753.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4817917 c.1300G>A p.D434N Chr.7: 2366535-2366535 on Build GRCh38
7079987 c.1311C>G p.F437L Chr.7: 2366546-2366546 on Build GRCh38
6880822 c.1233G>A p.T411T Chr.7: 2366392-2366392 on Build GRCh38
4461945 c.1227C>T p.I409I Chr.7: 2366386-2366386 on Build GRCh38
5872503 c.1202_1203delCT p.Q402fs*19 Chr.7: 2366361-2366362 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs147656962 Chr.7: 2366317-2366317 on Build GRCh38
rs961772962 Chr.7: 2366694-2366694 on Build GRCh38
rs750176840 Chr.7: 2366374-2366374 on Build GRCh38
rs767629839 Chr.7: 2366471-2366471 on Build GRCh38
rs373938496 Chr.7: 2366306-2366306 on Build GRCh38
rs11551171 Chr.7: 2366427-2366427 on Build GRCh38
rs892617969 Chr.7: 2366618-2366618 on Build GRCh38
rs752031549 Chr.7: 2366617-2366617 on Build GRCh38
rs752320741 Chr.7: 2366604-2366604 on Build GRCh38
rs1050412444 Chr.7: 2366676-2366678 on Build GRCh38
rs779435465 Chr.7: 2366319-2366319 on Build GRCh38
rs781417389 Chr.7: 2366622-2366622 on Build GRCh38
rs747386278 Chr.7: 2366454-2366454 on Build GRCh38
rs746924223 Chr.7: 2366643-2366643 on Build GRCh38
rs746096206 Chr.7: 2366512-2366512 on Build GRCh38
rs755665780 Chr.7: 2366308-2366308 on Build GRCh38
rs766156242 Chr.7: 2366479-2366479 on Build GRCh38
rs757161477 Chr.7: 2366302-2366302 on Build GRCh38
rs761797817 Chr.7: 2366368-2366368 on Build GRCh38
rs757597236 Chr.7: 2366507-2366507 on Build GRCh38
rs534158691 Chr.7: 2366254-2366257 on Build GRCh38
rs773507723 Chr.7: 2366336-2366336 on Build GRCh38
rs374733150 Chr.7: 2366626-2366626 on Build GRCh38
rs760611208 Chr.7: 2366560-2366560 on Build GRCh38
rs1025251092 Chr.7: 2366285-2366285 on Build GRCh38
rs113667884 Chr.7: 2366502-2366502 on Build GRCh38
rs765332786 Chr.7: 2366355-2366355 on Build GRCh38
rs367860910 Chr.7: 2366642-2366642 on Build GRCh38
rs780659010 Chr.7: 2366471-2366472 on Build GRCh38
rs761341411 Chr.7: 2366389-2366389 on Build GRCh38
rs376357368 Chr.7: 2366467-2366467 on Build GRCh38
rs768835970 Chr.7: 2366536-2366536 on Build GRCh38
rs1032511383 Chr.7: 2366259-2366259 on Build GRCh38
rs922516153 Chr.7: 2366434-2366434 on Build GRCh38
rs144904326 Chr.7: 2366416-2366416 on Build GRCh38
rs537722744 Chr.7: 2366688-2366688 on Build GRCh38
rs772585084 Chr.7: 2366421-2366421 on Build GRCh38
rs1030770963 Chr.7: 2366251-2366251 on Build GRCh38
rs780420259 Chr.7: 2366519-2366519 on Build GRCh38
rs890448130 Chr.7: 2366679-2366679 on Build GRCh38
rs551561961 Chr.7: 2366696-2366696 on Build GRCh38
rs749492873 Chr.7: 2366635-2366635 on Build GRCh38
rs922429370 Chr.7: 2366700-2366700 on Build GRCh38
rs759653106 Chr.7: 2366275-2366275 on Build GRCh38
rs947670377 Chr.7: 2366303-2366303 on Build GRCh38
rs754477038 Chr.7: 2366392-2366392 on Build GRCh38
rs374204646 Chr.7: 2366503-2366503 on Build GRCh38
rs969101244 Chr.7: 2366525-2366525 on Build GRCh38
rs549332269 Chr.7: 2366569-2366569 on Build GRCh38
rs778245646 Chr.7: 2366441-2366441 on Build GRCh38
rs545603601 Chr.7: 2366263-2366263 on Build GRCh38
rs201449328 Chr.7: 2366350-2366350 on Build GRCh38
rs757728481 Chr.7: 2366621-2366621 on Build GRCh38
rs760953443 Chr.7: 2366340-2366340 on Build GRCh38
rs576256095 Chr.7: 2366257-2366257 on Build GRCh38
rs2286204 Chr.7: 2366250-2366250 on Build GRCh38
rs1030826702 Chr.7: 2366480-2366481 on Build GRCh38
rs370757117 Chr.7: 2366484-2366484 on Build GRCh38
rs779620649 Chr.7: 2366312-2366312 on Build GRCh38
rs369567364 Chr.7: 2366305-2366305 on Build GRCh38
rs376326730 Chr.7: 2366391-2366391 on Build GRCh38
rs764731272 Chr.7: 2366496-2366496 on Build GRCh38
rs1037801741 Chr.7: 2366663-2366663 on Build GRCh38
rs142377401 Chr.7: 2366335-2366335 on Build GRCh38
rs1002873700 Chr.7: 2366664-2366664 on Build GRCh38
rs1008808515 Chr.7: 2366685-2366685 on Build GRCh38
rs760436774 Chr.7: 2366478-2366478 on Build GRCh38
rs748612094 Chr.7: 2366320-2366320 on Build GRCh38
rs932064752 Chr.7: 2366675-2366675 on Build GRCh38
rs999950513 Chr.7: 2366256-2366256 on Build GRCh38
rs774464816 Chr.7: 2366360-2366360 on Build GRCh38
rs371006035 Chr.7: 2366349-2366349 on Build GRCh38
rs756534645 Chr.7: 2366625-2366625 on Build GRCh38
rs758834764 Chr.7: 2366424-2366424 on Build GRCh38
rs763552523 Chr.7: 2366616-2366616 on Build GRCh38
rs759321115 Chr.7: 2366578-2366578 on Build GRCh38
rs530108384 Chr.7: 2366448-2366448 on Build GRCh38
rs770644085 Chr.7: 2366567-2366567 on Build GRCh38
rs571306218 Chr.7: 2366703-2366703 on Build GRCh38
rs370608315 Chr.7: 2366515-2366515 on Build GRCh38
rs768201055 Chr.7: 2366318-2366318 on Build GRCh38
rs1033638900 Chr.7: 2366669-2366669 on Build GRCh38
rs952749651 Chr.7: 2366673-2366673 on Build GRCh38
rs892306467 Chr.7: 2366707-2366707 on Build GRCh38
rs751517862 Chr.7: 2366298-2366298 on Build GRCh38
rs776755336 Chr.7: 2366462-2366462 on Build GRCh38
rs376014325 Chr.7: 2366510-2366510 on Build GRCh38
rs375624527 Chr.7: 2366276-2366276 on Build GRCh38
rs773064856 Chr.7: 2366543-2366543 on Build GRCh38
rs770278009 Chr.7: 2366273-2366273 on Build GRCh38
rs749437686 Chr.7: 2366533-2366533 on Build GRCh38
rs749669315 Chr.7: 2366301-2366301 on Build GRCh38
rs940823083 Chr.7: 2366656-2366656 on Build GRCh38
rs773189958 Chr.7: 2366475-2366475 on Build GRCh38
rs746066124 Chr.7: 2366466-2366466 on Build GRCh38
rs764924085 Chr.7: 2366594-2366594 on Build GRCh38
rs771120755 Chr.7: 2366343-2366343 on Build GRCh38
rs984157586 Chr.7: 2366686-2366686 on Build GRCh38
rs755933619 Chr.7: 2366386-2366386 on Build GRCh38
rs760763167 Chr.7: 2366468-2366468 on Build GRCh38
rs771302842 Chr.7: 2366459-2366459 on Build GRCh38
rs199814936 Chr.7: 2366372-2366372 on Build GRCh38

View Full Product Details