Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 12686696-12687194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL4A
Entrez Gene ID
10124
Gene Name
ADP ribosylation factor like GTPase 4A
Gene Aliases
ARL4
UniGene
Hs.245540
Gene Chromosome Location
Chr.7: 12686827-12690934 on Build GRCh38
Transcripts
NM_001037164.2 NM_212460.3

SNP Detail

SNP ID Chromosome Location
rs938290719 Chr.7: 12686952-12686952 on Build GRCh38
rs917471861 Chr.7: 12686831-12686841 on Build GRCh38
rs975437363 Chr.7: 12686739-12686739 on Build GRCh38
rs1042051009 Chr.7: 12686799-12686799 on Build GRCh38
rs893305964 Chr.7: 12686996-12686996 on Build GRCh38
rs546959731 Chr.7: 12686970-12686970 on Build GRCh38
rs540115829 Chr.7: 12686858-12686858 on Build GRCh38
rs1020940111 Chr.7: 12687013-12687013 on Build GRCh38
rs906855808 Chr.7: 12686820-12686820 on Build GRCh38
rs989567502 Chr.7: 12687054-12687054 on Build GRCh38
rs531038447 Chr.7: 12686928-12686928 on Build GRCh38
rs997089197 Chr.7: 12687009-12687009 on Build GRCh38
rs528466913 Chr.7: 12687061-12687069 on Build GRCh38
rs921364935 Chr.7: 12686749-12686749 on Build GRCh38
rs983333715 Chr.7: 12687158-12687158 on Build GRCh38
rs193271226 Chr.7: 12686760-12686760 on Build GRCh38
rs1018193614 Chr.7: 12687092-12687092 on Build GRCh38
rs1045376464 Chr.7: 12686947-12686947 on Build GRCh38
rs920052092 Chr.7: 12687137-12687137 on Build GRCh38
rs13227823 Chr.7: 12686825-12686825 on Build GRCh38
rs560432508 Chr.7: 12686860-12686860 on Build GRCh38
rs1055479307 Chr.7: 12686957-12686957 on Build GRCh38
rs778821231 Chr.7: 12686768-12686768 on Build GRCh38
rs968811907 Chr.7: 12687060-12687060 on Build GRCh38
rs899023117 Chr.7: 12686772-12686772 on Build GRCh38
rs374620591 Chr.7: 12686757-12686757 on Build GRCh38
rs577727102 Chr.7: 12686881-12686881 on Build GRCh38
rs975386659 Chr.7: 12686890-12686890 on Build GRCh38
rs969825996 Chr.7: 12686836-12686836 on Build GRCh38
rs987475725 Chr.7: 12687070-12687070 on Build GRCh38
rs574355548 Chr.7: 12686832-12686832 on Build GRCh38
rs928504234 Chr.7: 12687143-12687143 on Build GRCh38
rs1002553098 Chr.7: 12686823-12686823 on Build GRCh38
rs955620038 Chr.7: 12687067-12687067 on Build GRCh38
rs13227930 Chr.7: 12686883-12686883 on Build GRCh38
rs965377698 Chr.7: 12686728-12686728 on Build GRCh38
rs913190156 Chr.7: 12686719-12686719 on Build GRCh38
rs150622801 Chr.7: 12686929-12686929 on Build GRCh38
rs955536130 Chr.7: 12686882-12686882 on Build GRCh38
rs1001298906 Chr.7: 12686843-12686843 on Build GRCh38
rs907715645 Chr.7: 12687162-12687162 on Build GRCh38
rs1047650308 Chr.7: 12686797-12686797 on Build GRCh38
rs911224793 Chr.7: 12686893-12686893 on Build GRCh38
rs552153789 Chr.7: 12687153-12687153 on Build GRCh38
rs1004804317 Chr.7: 12686993-12686993 on Build GRCh38
rs958302516 Chr.7: 12687051-12687051 on Build GRCh38
rs887252478 Chr.7: 12686977-12686977 on Build GRCh38
rs894182355 Chr.7: 12686962-12686962 on Build GRCh38
rs930514034 Chr.7: 12686909-12686909 on Build GRCh38
rs945981860 Chr.7: 12686781-12686781 on Build GRCh38
rs1054303747 Chr.7: 12686770-12686770 on Build GRCh38
rs886099628 Chr.7: 12686812-12686812 on Build GRCh38
rs1033139235 Chr.7: 12686856-12686856 on Build GRCh38
rs567192414 Chr.7: 12687091-12687091 on Build GRCh38
rs79178345 Chr.7: 12687129-12687129 on Build GRCh38
rs950291701 Chr.7: 12686890-12686915 on Build GRCh38
rs957533479 Chr.7: 12686866-12686866 on Build GRCh38
rs911331336 Chr.7: 12686745-12686745 on Build GRCh38
rs117002576 Chr.7: 12686934-12686934 on Build GRCh38
rs994676010 Chr.7: 12686778-12686778 on Build GRCh38
rs1010424930 Chr.7: 12687001-12687001 on Build GRCh38
rs974133263 Chr.7: 12686905-12686905 on Build GRCh38
rs530344018 Chr.7: 12686936-12686936 on Build GRCh38
rs912543226 Chr.7: 12686720-12686720 on Build GRCh38
rs989460269 Chr.7: 12686870-12686870 on Build GRCh38
rs967213941 Chr.7: 12687073-12687073 on Build GRCh38
rs1013960718 Chr.7: 12686819-12686819 on Build GRCh38
rs934040765 Chr.7: 12686730-12686730 on Build GRCh38
rs1048103589 Chr.7: 12686923-12686923 on Build GRCh38
rs964022963 Chr.7: 12687119-12687119 on Build GRCh38
rs768343962 Chr.7: 12686949-12686949 on Build GRCh38
rs1032796154 Chr.7: 12686975-12686975 on Build GRCh38
rs928555305 Chr.7: 12686948-12686948 on Build GRCh38
rs1028277493 Chr.7: 12687030-12687030 on Build GRCh38
rs183276608 Chr.7: 12686900-12686900 on Build GRCh38
rs920395806 Chr.7: 12686908-12686908 on Build GRCh38
rs866463437 Chr.7: 12686998-12686998 on Build GRCh38
rs930455245 Chr.7: 12687146-12687146 on Build GRCh38
rs958139741 Chr.7: 12686902-12686902 on Build GRCh38
rs914423383 Chr.7: 12686777-12686777 on Build GRCh38
rs370671246 Chr.7: 12686731-12686731 on Build GRCh38

View Full Product Details