Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 138804677-138805200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM213
Entrez Gene ID
155006
Gene Name
transmembrane protein 213
Gene Aliases
-
UniGene
Hs.567729
Gene Chromosome Location
Chr.7: 138797487-138806759 on Build GRCh38
Transcripts
NM_001085429.1 XM_005250167.3

SNP Detail

SNP ID Chromosome Location
rs901319164 Chr.7: 138804709-138804709 on Build GRCh38
rs973983916 Chr.7: 138804716-138804716 on Build GRCh38
rs757711423 Chr.7: 138804862-138804862 on Build GRCh38
rs946529666 Chr.7: 138804936-138804936 on Build GRCh38
rs1041109431 Chr.7: 138805142-138805142 on Build GRCh38
rs779497301 Chr.7: 138804958-138804958 on Build GRCh38
rs1049917565 Chr.7: 138805171-138805171 on Build GRCh38
rs1049955354 Chr.7: 138804788-138804788 on Build GRCh38
rs1011187760 Chr.7: 138804858-138804858 on Build GRCh38
rs913733126 Chr.7: 138804934-138804934 on Build GRCh38
rs967448648 Chr.7: 138804901-138804901 on Build GRCh38
rs998590211 Chr.7: 138804920-138804920 on Build GRCh38
rs1050406206 Chr.7: 138805044-138805044 on Build GRCh38
rs577655055 Chr.7: 138804821-138804821 on Build GRCh38
rs537981717 Chr.7: 138804704-138804704 on Build GRCh38
rs763875311 Chr.7: 138804773-138804773 on Build GRCh38
rs35062638 Chr.7: 138804758-138804759 on Build GRCh38
rs922708929 Chr.7: 138805149-138805149 on Build GRCh38
rs768334384 Chr.7: 138805143-138805143 on Build GRCh38
rs908500019 Chr.7: 138805134-138805134 on Build GRCh38
rs370178388 Chr.7: 138804998-138804998 on Build GRCh38
rs939090010 Chr.7: 138804886-138804886 on Build GRCh38
rs991342956 Chr.7: 138805058-138805058 on Build GRCh38
rs970057582 Chr.7: 138804787-138804787 on Build GRCh38
rs562565651 Chr.7: 138805087-138805087 on Build GRCh38
rs754139943 Chr.7: 138804801-138804801 on Build GRCh38
rs558116460 Chr.7: 138804739-138804739 on Build GRCh38
rs962246007 Chr.7: 138804711-138804711 on Build GRCh38
rs564866776 Chr.7: 138805155-138805155 on Build GRCh38
rs897913036 Chr.7: 138805139-138805139 on Build GRCh38
rs1035860775 Chr.7: 138804967-138804967 on Build GRCh38
rs867794019 Chr.7: 138805071-138805071 on Build GRCh38
rs553601339 Chr.7: 138804974-138804974 on Build GRCh38
rs987883819 Chr.7: 138804913-138804913 on Build GRCh38
rs1021254822 Chr.7: 138804883-138804883 on Build GRCh38
rs190054494 Chr.7: 138805116-138805116 on Build GRCh38
rs994988251 Chr.7: 138805168-138805168 on Build GRCh38
rs185594118 Chr.7: 138804850-138804850 on Build GRCh38
rs542775184 Chr.7: 138805007-138805007 on Build GRCh38
rs932805779 Chr.7: 138805156-138805156 on Build GRCh38
rs952642678 Chr.7: 138805114-138805114 on Build GRCh38
rs2034507 Chr.7: 138805138-138805138 on Build GRCh38
rs894087762 Chr.7: 138804844-138804844 on Build GRCh38
rs932586707 Chr.7: 138805009-138805009 on Build GRCh38

View Full Product Details