Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179843862-179844395 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOR1AIP2
Entrez Gene ID
163590
Gene Name
torsin 1A interacting protein 2
Gene Aliases
IFRG15, LULL1, NET9
UniGene
Hs.571797
Gene Chromosome Location
Chr.1: 179839967-179878614 on Build GRCh38
Transcripts
NM_001199260.1 NM_145034.4 XM_005244939.4 XM_017000474.1 XM_017000475.1 XM_017000476.1

SNP Detail

SNP ID Chromosome Location
rs76645336 Chr.1: 179843984-179843984 on Build GRCh38
rs932064971 Chr.1: 179844320-179844320 on Build GRCh38
rs931984620 Chr.1: 179844118-179844118 on Build GRCh38
rs143325909 Chr.1: 179844365-179844365 on Build GRCh38
rs577575788 Chr.1: 179844181-179844181 on Build GRCh38
rs1022291703 Chr.1: 179843894-179843894 on Build GRCh38
rs796364047 Chr.1: 179844317-179844317 on Build GRCh38
rs200870209 Chr.1: 179843902-179843902 on Build GRCh38
rs907672317 Chr.1: 179844039-179844039 on Build GRCh38
rs1037723428 Chr.1: 179844082-179844082 on Build GRCh38
rs763104271 Chr.1: 179843892-179843892 on Build GRCh38
rs929605011 Chr.1: 179844229-179844229 on Build GRCh38
rs1281408 Chr.1: 179844364-179844364 on Build GRCh38
rs968398476 Chr.1: 179843891-179843891 on Build GRCh38
rs547982993 Chr.1: 179844335-179844335 on Build GRCh38
rs752743615 Chr.1: 179844119-179844119 on Build GRCh38
rs999769275 Chr.1: 179843919-179843919 on Build GRCh38
rs960883670 Chr.1: 179844027-179844027 on Build GRCh38
rs573488507 Chr.1: 179844228-179844228 on Build GRCh38
rs1010017420 Chr.1: 179844362-179844362 on Build GRCh38
rs974170322 Chr.1: 179844216-179844216 on Build GRCh38
rs996611773 Chr.1: 179843888-179843888 on Build GRCh38
rs940799154 Chr.1: 179844050-179844050 on Build GRCh38
rs562210604 Chr.1: 179844303-179844303 on Build GRCh38
rs529588611 Chr.1: 179844026-179844026 on Build GRCh38
rs2794583 Chr.1: 179844217-179844217 on Build GRCh38
rs766258882 Chr.1: 179844280-179844280 on Build GRCh38
rs373647753 Chr.1: 179843907-179843907 on Build GRCh38
rs537824737 Chr.1: 179844037-179844037 on Build GRCh38
rs772856443 Chr.1: 179844127-179844127 on Build GRCh38
rs557500707 Chr.1: 179844114-179844114 on Build GRCh38
rs938546566 Chr.1: 179844012-179844012 on Build GRCh38
rs1023424329 Chr.1: 179844113-179844113 on Build GRCh38
rs540160821 Chr.1: 179844186-179844186 on Build GRCh38
rs1044144075 Chr.1: 179844343-179844343 on Build GRCh38
rs943879931 Chr.1: 179844301-179844301 on Build GRCh38
rs190229393 Chr.1: 179844279-179844279 on Build GRCh38
rs992294380 Chr.1: 179844100-179844100 on Build GRCh38
rs754986193 Chr.1: 179844327-179844327 on Build GRCh38

View Full Product Details