Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 179850758-179851252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TOR1AIP2
Entrez Gene ID
163590
Gene Name
torsin 1A interacting protein 2
Gene Aliases
IFRG15, LULL1, NET9
UniGene
Hs.571797
Gene Chromosome Location
Chr.1: 179839967-179878614 on Build GRCh38
Transcripts
NM_001199260.1 NM_145034.4 XM_005244939.4 XM_017000474.1 XM_017000475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2095638 c.428C>T p.A143V Chr.1: 179850970-179850970 on Build GRCh38
6872748 c.237G>A p.G79G Chr.1: 179851161-179851161 on Build GRCh38
900321 c.538C>T p.R180* Chr.1: 179850860-179850860 on Build GRCh38
3863740 c.293G>A p.G98E Chr.1: 179851105-179851105 on Build GRCh38
5485517 c.510G>A p.G170G Chr.1: 179850888-179850888 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759557017 Chr.1: 179851117-179851117 on Build GRCh38
rs752491114 Chr.1: 179851126-179851126 on Build GRCh38
rs746341717 Chr.1: 179851167-179851167 on Build GRCh38
rs752007143 Chr.1: 179851112-179851113 on Build GRCh38
rs774666160 Chr.1: 179850842-179850842 on Build GRCh38
rs756568716 Chr.1: 179851179-179851179 on Build GRCh38
rs566526582 Chr.1: 179850968-179850968 on Build GRCh38
rs780574843 Chr.1: 179851199-179851199 on Build GRCh38
rs1009276802 Chr.1: 179850845-179850845 on Build GRCh38
rs756750047 Chr.1: 179850891-179850891 on Build GRCh38
rs202192648 Chr.1: 179851115-179851115 on Build GRCh38
rs369907841 Chr.1: 179850963-179850963 on Build GRCh38
rs749891313 Chr.1: 179850970-179850970 on Build GRCh38
rs747016311 Chr.1: 179851030-179851030 on Build GRCh38
rs772856426 Chr.1: 179851106-179851106 on Build GRCh38
rs769022500 Chr.1: 179851204-179851204 on Build GRCh38
rs143074105 Chr.1: 179850984-179850984 on Build GRCh38
rs374345176 Chr.1: 179850801-179850801 on Build GRCh38
rs773952986 Chr.1: 179850924-179850924 on Build GRCh38
rs761838923 Chr.1: 179850788-179850788 on Build GRCh38
rs781038841 Chr.1: 179850827-179850827 on Build GRCh38
rs763610484 Chr.1: 179850862-179850862 on Build GRCh38
rs1038341854 Chr.1: 179851060-179851060 on Build GRCh38
rs115350827 Chr.1: 179850896-179850896 on Build GRCh38
rs760263804 Chr.1: 179850977-179850977 on Build GRCh38
rs754807579 Chr.1: 179850922-179850922 on Build GRCh38
rs781375352 Chr.1: 179851155-179851155 on Build GRCh38
rs114937221 Chr.1: 179851208-179851208 on Build GRCh38
rs919778741 Chr.1: 179851063-179851063 on Build GRCh38
rs953726788 Chr.1: 179850863-179850863 on Build GRCh38
rs748173529 Chr.1: 179850847-179850847 on Build GRCh38
rs757268800 Chr.1: 179851148-179851148 on Build GRCh38
rs78506845 Chr.1: 179851008-179851008 on Build GRCh38
rs756789076 Chr.1: 179850824-179850824 on Build GRCh38
rs386637213 Chr.1: 179850853-179850854 on Build GRCh38
rs768712464 Chr.1: 179850841-179850841 on Build GRCh38
rs751817866 Chr.1: 179851136-179851136 on Build GRCh38
rs749508191 Chr.1: 179850835-179850835 on Build GRCh38
rs770205390 Chr.1: 179850962-179850962 on Build GRCh38
rs61826282 Chr.1: 179851198-179851198 on Build GRCh38
rs1047814906 Chr.1: 179851092-179851092 on Build GRCh38
rs375587727 Chr.1: 179851018-179851018 on Build GRCh38
rs779222171 Chr.1: 179850923-179850923 on Build GRCh38
rs375297372 Chr.1: 179850895-179850895 on Build GRCh38
rs766995676 Chr.1: 179850969-179850969 on Build GRCh38
rs568756687 Chr.1: 179850993-179850993 on Build GRCh38
rs200428893 Chr.1: 179851140-179851140 on Build GRCh38
rs139945443 Chr.1: 179851228-179851228 on Build GRCh38
rs772272447 Chr.1: 179850929-179850929 on Build GRCh38
rs138587725 Chr.1: 179851108-179851108 on Build GRCh38
rs749587597 Chr.1: 179851200-179851200 on Build GRCh38
rs767083823 Chr.1: 179850893-179850893 on Build GRCh38
rs367898216 Chr.1: 179850881-179850881 on Build GRCh38
rs377600126 Chr.1: 179851094-179851094 on Build GRCh38
rs199500621 Chr.1: 179851028-179851028 on Build GRCh38
rs373555927 Chr.1: 179851006-179851006 on Build GRCh38
rs115892642 Chr.1: 179850795-179850795 on Build GRCh38
rs775886374 Chr.1: 179851097-179851097 on Build GRCh38
rs369872700 Chr.1: 179851026-179851026 on Build GRCh38
rs370469437 Chr.1: 179850822-179850822 on Build GRCh38
rs929769544 Chr.1: 179851073-179851073 on Build GRCh38
rs778071605 Chr.1: 179851133-179851133 on Build GRCh38
rs548916342 Chr.1: 179851036-179851036 on Build GRCh38
rs763334353 Chr.1: 179851104-179851104 on Build GRCh38
rs759963550 Chr.1: 179850859-179850859 on Build GRCh38
rs748996935 Chr.1: 179850916-179850916 on Build GRCh38
rs758295893 Chr.1: 179851132-179851132 on Build GRCh38
rs201930264 Chr.1: 179850833-179850833 on Build GRCh38
rs752934673 Chr.1: 179851016-179851016 on Build GRCh38
rs375003867 Chr.1: 179850832-179850832 on Build GRCh38
rs773839681 Chr.1: 179850848-179850848 on Build GRCh38
rs747496738 Chr.1: 179850940-179850940 on Build GRCh38
rs200558019 Chr.1: 179851107-179851107 on Build GRCh38
rs770026524 Chr.1: 179851096-179851096 on Build GRCh38
rs138617443 Chr.1: 179850996-179850996 on Build GRCh38
rs771289980 Chr.1: 179850948-179850948 on Build GRCh38
rs761331608 Chr.1: 179850853-179850853 on Build GRCh38
rs914441089 Chr.1: 179850941-179850941 on Build GRCh38
rs763635134 Chr.1: 179850816-179850816 on Build GRCh38
rs201392581 Chr.1: 179850831-179850831 on Build GRCh38
rs766915015 Chr.1: 179851090-179851091 on Build GRCh38
rs765100630 Chr.1: 179851122-179851122 on Build GRCh38
rs149853908 Chr.1: 179850961-179850961 on Build GRCh38
rs766735054 Chr.1: 179850854-179850854 on Build GRCh38
rs112711003 Chr.1: 179851050-179851050 on Build GRCh38
rs776905107 Chr.1: 179850858-179850858 on Build GRCh38
rs758026520 Chr.1: 179850808-179850808 on Build GRCh38
rs780062708 Chr.1: 179850897-179850897 on Build GRCh38
rs771673669 Chr.1: 179851105-179851105 on Build GRCh38
rs542174680 Chr.1: 179850783-179850783 on Build GRCh38
rs141174223 Chr.1: 179851125-179851125 on Build GRCh38
rs752389847 Chr.1: 179850805-179850805 on Build GRCh38
rs777222778 Chr.1: 179850955-179850955 on Build GRCh38
rs778038876 Chr.1: 179850932-179850932 on Build GRCh38

View Full Product Details