Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 110201099-110201598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C9orf152
Entrez Gene ID
401546
Gene Name
chromosome 9 open reading frame 152
Gene Aliases
bA470J20.2
UniGene
Hs.125608
Gene Chromosome Location
Chr.9: 110199561-110208133 on Build GRCh38
Transcripts
NM_001012993.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3652794 c.443G>A p.G148E Chr.9: 110201162-110201162 on Build GRCh38
3925883 c.276A>G p.E92E Chr.9: 110201392-110201392 on Build GRCh38
4498323 c.517C>T p.P173S Chr.9: 110201151-110201151 on Build GRCh38
3652798 c.243G>A p.L81L Chr.9: 110201362-110201362 on Build GRCh38
6991538 c.467A>G p.D156G Chr.9: 110201201-110201201 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775767635 Chr.9: 110201142-110201142 on Build GRCh38
rs752870762 Chr.9: 110201516-110201516 on Build GRCh38
rs368910774 Chr.9: 110201524-110201524 on Build GRCh38
rs372134658 Chr.9: 110201172-110201172 on Build GRCh38
rs780933917 Chr.9: 110201125-110201125 on Build GRCh38
rs372684740 Chr.9: 110201522-110201522 on Build GRCh38
rs774648485 Chr.9: 110201440-110201440 on Build GRCh38
rs772416281 Chr.9: 110201141-110201141 on Build GRCh38
rs779668852 Chr.9: 110201198-110201198 on Build GRCh38
rs745670115 Chr.9: 110201398-110201398 on Build GRCh38
rs760610085 Chr.9: 110201156-110201156 on Build GRCh38
rs752224113 Chr.9: 110201304-110201304 on Build GRCh38
rs150839166 Chr.9: 110201225-110201225 on Build GRCh38
rs755614512 Chr.9: 110201374-110201374 on Build GRCh38
rs769440312 Chr.9: 110201128-110201128 on Build GRCh38
rs780345431 Chr.9: 110201312-110201312 on Build GRCh38
rs144259716 Chr.9: 110201178-110201178 on Build GRCh38
rs898971211 Chr.9: 110201369-110201369 on Build GRCh38
rs920733553 Chr.9: 110201365-110201365 on Build GRCh38
rs201531341 Chr.9: 110201264-110201264 on Build GRCh38
rs747244822 Chr.9: 110201318-110201318 on Build GRCh38
rs756175021 Chr.9: 110201308-110201308 on Build GRCh38
rs970449279 Chr.9: 110201253-110201253 on Build GRCh38
rs575706795 Chr.9: 110201257-110201257 on Build GRCh38
rs115367896 Chr.9: 110201335-110201335 on Build GRCh38
rs1033310910 Chr.9: 110201566-110201566 on Build GRCh38
rs769434291 Chr.9: 110201317-110201317 on Build GRCh38
rs527359075 Chr.9: 110201540-110201540 on Build GRCh38
rs201662135 Chr.9: 110201129-110201129 on Build GRCh38
rs754543502 Chr.9: 110201285-110201285 on Build GRCh38
rs758115589 Chr.9: 110201195-110201195 on Build GRCh38
rs774194242 Chr.9: 110201350-110201350 on Build GRCh38
rs369380119 Chr.9: 110201488-110201488 on Build GRCh38
rs199516049 Chr.9: 110201459-110201459 on Build GRCh38
rs778610464 Chr.9: 110201397-110201397 on Build GRCh38
rs529821968 Chr.9: 110201450-110201450 on Build GRCh38
rs776748221 Chr.9: 110201266-110201266 on Build GRCh38
rs899248810 Chr.9: 110201138-110201138 on Build GRCh38
rs554733063 Chr.9: 110201274-110201274 on Build GRCh38
rs564934493 Chr.9: 110201364-110201364 on Build GRCh38
rs765206256 Chr.9: 110201482-110201482 on Build GRCh38
rs766685025 Chr.9: 110201344-110201344 on Build GRCh38
rs749079859 Chr.9: 110201228-110201228 on Build GRCh38
rs149848546 Chr.9: 110201446-110201446 on Build GRCh38
rs762260259 Chr.9: 110201171-110201171 on Build GRCh38
rs975968092 Chr.9: 110201515-110201515 on Build GRCh38
rs752158560 Chr.9: 110201371-110201371 on Build GRCh38
rs778278588 Chr.9: 110201417-110201417 on Build GRCh38
rs370004312 Chr.9: 110201319-110201319 on Build GRCh38
rs759883794 Chr.9: 110201441-110201441 on Build GRCh38
rs763245177 Chr.9: 110201276-110201276 on Build GRCh38
rs911537829 Chr.9: 110201569-110201569 on Build GRCh38
rs78835206 Chr.9: 110201179-110201179 on Build GRCh38
rs765730779 Chr.9: 110201275-110201275 on Build GRCh38
rs753367387 Chr.9: 110201462-110201462 on Build GRCh38
rs957588533 Chr.9: 110201492-110201492 on Build GRCh38
rs757238941 Chr.9: 110201393-110201393 on Build GRCh38
rs769826122 Chr.9: 110201411-110201411 on Build GRCh38
rs768697667 Chr.9: 110201124-110201124 on Build GRCh38
rs926082326 Chr.9: 110201246-110201246 on Build GRCh38
rs1009527018 Chr.9: 110201223-110201223 on Build GRCh38
rs781157372 Chr.9: 110201188-110201188 on Build GRCh38
rs764154176 Chr.9: 110201381-110201381 on Build GRCh38
rs747142567 Chr.9: 110201261-110201261 on Build GRCh38
rs76016983 Chr.9: 110201523-110201523 on Build GRCh38
rs573275088 Chr.9: 110201327-110201327 on Build GRCh38
rs967593372 Chr.9: 110201212-110201212 on Build GRCh38
rs200937336 Chr.9: 110201442-110201442 on Build GRCh38
rs770680296 Chr.9: 110201300-110201300 on Build GRCh38
rs759890058 Chr.9: 110201140-110201140 on Build GRCh38
rs1036216231 Chr.9: 110201345-110201345 on Build GRCh38
rs755214234 Chr.9: 110201400-110201400 on Build GRCh38
rs772432788 Chr.9: 110201249-110201249 on Build GRCh38
rs776642989 Chr.9: 110201326-110201326 on Build GRCh38
rs1003222768 Chr.9: 110201564-110201564 on Build GRCh38
rs146599802 Chr.9: 110201419-110201419 on Build GRCh38
rs10120707 Chr.9: 110201224-110201224 on Build GRCh38
rs773642922 Chr.9: 110201334-110201334 on Build GRCh38
rs138966973 Chr.9: 110201157-110201157 on Build GRCh38
rs149319009 Chr.9: 110201158-110201158 on Build GRCh38
rs117858678 Chr.9: 110201217-110201217 on Build GRCh38
rs745826686 Chr.9: 110201273-110201274 on Build GRCh38
rs761906385 Chr.9: 110201465-110201465 on Build GRCh38
rs779362819 Chr.9: 110201315-110201315 on Build GRCh38
rs111567458 Chr.9: 110201401-110201401 on Build GRCh38
rs753779115 Chr.9: 110201391-110201391 on Build GRCh38
rs1028781766 Chr.9: 110201518-110201518 on Build GRCh38
rs774758275 Chr.9: 110201354-110201354 on Build GRCh38
rs751094160 Chr.9: 110201214-110201214 on Build GRCh38
rs759154481 Chr.9: 110201359-110201359 on Build GRCh38
rs376699609 Chr.9: 110201311-110201311 on Build GRCh38
rs1054947635 Chr.9: 110201498-110201498 on Build GRCh38
rs758591848 Chr.9: 110201548-110201548 on Build GRCh38
rs775357899 Chr.9: 110201408-110201410 on Build GRCh38
rs1032143477 Chr.9: 110201192-110201192 on Build GRCh38
rs751135399 Chr.9: 110201282-110201282 on Build GRCh38
rs781225120 Chr.9: 110201219-110201219 on Build GRCh38
rs548379585 Chr.9: 110201514-110201514 on Build GRCh38
rs765060902 Chr.9: 110201518-110201518 on Build GRCh38
rs766025731 Chr.9: 110201279-110201279 on Build GRCh38
rs766987153 Chr.9: 110201294-110201294 on Build GRCh38
rs1038984113 Chr.9: 110201438-110201438 on Build GRCh38
rs4978888 Chr.9: 110201422-110201422 on Build GRCh38
rs750200274 Chr.9: 110201508-110201508 on Build GRCh38

View Full Product Details