Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72751757-72752303 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC1
Entrez Gene ID
117531
Gene Name
transmembrane channel like 1
Gene Aliases
DFNA36, DFNB11, DFNB7
UniGene
Hs.670211
Gene Chromosome Location
Chr.9: 72521801-72836351 on Build GRCh38
Transcripts
NM_138691.2 XM_017014256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3746037 c.602T>C p.V201A Chr.9: 72751916-72751916 on Build GRCh38
150617 c.640G>A p.E214K Chr.9: 72751954-72751954 on Build GRCh38
4734074 c.546C>T p.G182G Chr.9: 72751860-72751860 on Build GRCh38
5730654 c.556G>T p.A186S Chr.9: 72751870-72751870 on Build GRCh38
5411224 c.582G>A p.W194* Chr.9: 72751896-72751896 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs34157686 Chr.9: 72752127-72752128 on Build GRCh38
rs936541030 Chr.9: 72752109-72752109 on Build GRCh38
rs762575823 Chr.9: 72752059-72752059 on Build GRCh38
rs372449375 Chr.9: 72751917-72751917 on Build GRCh38
rs773838098 Chr.9: 72751858-72751858 on Build GRCh38
rs774994689 Chr.9: 72751965-72751965 on Build GRCh38
rs1036361471 Chr.9: 72751788-72751788 on Build GRCh38
rs760128641 Chr.9: 72751967-72751967 on Build GRCh38
rs770508485 Chr.9: 72752190-72752190 on Build GRCh38
rs368496447 Chr.9: 72752044-72752044 on Build GRCh38
rs748580616 Chr.9: 72751941-72751941 on Build GRCh38
rs555485816 Chr.9: 72752048-72752048 on Build GRCh38
rs889628028 Chr.9: 72752022-72752022 on Build GRCh38
rs747576868 Chr.9: 72751810-72751810 on Build GRCh38
rs549448535 Chr.9: 72751819-72751819 on Build GRCh38
rs553402359 Chr.9: 72752229-72752229 on Build GRCh38
rs1050905891 Chr.9: 72752256-72752256 on Build GRCh38
rs752255917 Chr.9: 72752009-72752009 on Build GRCh38
rs1030610346 Chr.9: 72752015-72752015 on Build GRCh38
rs199560971 Chr.9: 72751859-72751859 on Build GRCh38
rs371578516 Chr.9: 72751961-72751961 on Build GRCh38
rs557483327 Chr.9: 72752150-72752150 on Build GRCh38
rs368246679 Chr.9: 72751899-72751899 on Build GRCh38
rs750662616 Chr.9: 72751913-72751913 on Build GRCh38
rs774972622 Chr.9: 72751870-72751870 on Build GRCh38
rs375128682 Chr.9: 72751863-72751863 on Build GRCh38
rs573450985 Chr.9: 72752276-72752276 on Build GRCh38
rs201051103 Chr.9: 72751826-72751826 on Build GRCh38
rs577361343 Chr.9: 72752173-72752173 on Build GRCh38
rs535224088 Chr.9: 72751793-72751794 on Build GRCh38
rs773609760 Chr.9: 72751946-72751946 on Build GRCh38
rs765001441 Chr.9: 72751979-72751979 on Build GRCh38
rs761025965 Chr.9: 72751829-72751830 on Build GRCh38
rs1038149335 Chr.9: 72752041-72752041 on Build GRCh38
rs961947747 Chr.9: 72752073-72752073 on Build GRCh38
rs192851206 Chr.9: 72752093-72752093 on Build GRCh38
rs967374875 Chr.9: 72752246-72752246 on Build GRCh38
rs727504470 Chr.9: 72751871-72751871 on Build GRCh38
rs529821299 Chr.9: 72751791-72751791 on Build GRCh38
rs773573869 Chr.9: 72751805-72751806 on Build GRCh38
rs753687760 Chr.9: 72751903-72751903 on Build GRCh38
rs1005369861 Chr.9: 72752033-72752033 on Build GRCh38
rs780260889 Chr.9: 72751809-72751809 on Build GRCh38
rs780348810 Chr.9: 72751919-72751919 on Build GRCh38
rs1001705124 Chr.9: 72752113-72752113 on Build GRCh38
rs1042818751 Chr.9: 72751901-72751901 on Build GRCh38
rs376768305 Chr.9: 72751920-72751920 on Build GRCh38
rs978327175 Chr.9: 72752252-72752252 on Build GRCh38
rs554722114 Chr.9: 72752001-72752001 on Build GRCh38
rs865985561 Chr.9: 72751895-72751895 on Build GRCh38
rs758699963 Chr.9: 72751997-72751997 on Build GRCh38
rs745500816 Chr.9: 72752014-72752015 on Build GRCh38
rs368042336 Chr.9: 72751933-72751933 on Build GRCh38
rs922610007 Chr.9: 72752272-72752272 on Build GRCh38
rs751716573 Chr.9: 72752004-72752004 on Build GRCh38
rs989635871 Chr.9: 72751857-72751857 on Build GRCh38
rs565926145 Chr.9: 72751942-72751942 on Build GRCh38
rs568140027 Chr.9: 72752058-72752058 on Build GRCh38
rs770640925 Chr.9: 72751850-72751850 on Build GRCh38
rs777142875 Chr.9: 72751843-72751843 on Build GRCh38
rs903441299 Chr.9: 72752003-72752003 on Build GRCh38
rs927821762 Chr.9: 72752063-72752064 on Build GRCh38
rs781747541 Chr.9: 72751938-72751938 on Build GRCh38
rs957749192 Chr.9: 72752036-72752036 on Build GRCh38
rs199499836 Chr.9: 72751932-72751932 on Build GRCh38
rs750228353 Chr.9: 72751992-72751992 on Build GRCh38
rs200755455 Chr.9: 72751835-72751835 on Build GRCh38
rs761790416 Chr.9: 72751974-72751974 on Build GRCh38
rs756960425 Chr.9: 72751910-72751910 on Build GRCh38
rs745569653 Chr.9: 72751951-72751951 on Build GRCh38
rs534533412 Chr.9: 72751971-72751971 on Build GRCh38
rs766347593 Chr.9: 72751834-72751835 on Build GRCh38
rs755641942 Chr.9: 72752057-72752057 on Build GRCh38
rs376222732 Chr.9: 72752202-72752202 on Build GRCh38
rs773823438 Chr.9: 72752046-72752046 on Build GRCh38

View Full Product Details