Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 134433984-134434487 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RXRA
Entrez Gene ID
6256
Gene Name
retinoid X receptor alpha
Gene Aliases
NR2B1
UniGene
Hs.590886
Gene Chromosome Location
Chr.9: 134326463-134440586 on Build GRCh38
Transcripts
NM_001291920.1 NM_001291921.1 NM_002957.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3214209 c.1173G>A p.A391A Chr.9: 134434139-134434139 on Build GRCh38
4609833 c.1157_1158insCGG p.P386_A387insG Chr.9: 134434123-134434124 on Build GRCh38
1106575 c.1235C>A p.P412Q Chr.9: 134434201-134434201 on Build GRCh38
5977126 c.1161C>G p.A387A Chr.9: 134434127-134434127 on Build GRCh38
1106574 c.1159G>T p.A387S Chr.9: 134434125-134434125 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs927420933 Chr.9: 134434014-134434014 on Build GRCh38
rs183422344 Chr.9: 134434184-134434184 on Build GRCh38
rs908331808 Chr.9: 134434374-134434374 on Build GRCh38
rs1014465227 Chr.9: 134434278-134434278 on Build GRCh38
rs965179038 Chr.9: 134434381-134434381 on Build GRCh38
rs748679602 Chr.9: 134434406-134434406 on Build GRCh38
rs767455588 Chr.9: 134434224-134434224 on Build GRCh38
rs776877361 Chr.9: 134434202-134434202 on Build GRCh38
rs748715081 Chr.9: 134434062-134434062 on Build GRCh38
rs1031813507 Chr.9: 134434011-134434011 on Build GRCh38
rs759690021 Chr.9: 134434091-134434091 on Build GRCh38
rs919605924 Chr.9: 134434085-134434085 on Build GRCh38
rs552029214 Chr.9: 134434268-134434268 on Build GRCh38
rs772712511 Chr.9: 134434176-134434176 on Build GRCh38
rs750236508 Chr.9: 134434124-134434124 on Build GRCh38
rs551097827 Chr.9: 134434368-134434368 on Build GRCh38
rs775612621 Chr.9: 134434220-134434220 on Build GRCh38
rs771014431 Chr.9: 134434201-134434201 on Build GRCh38
rs1014355615 Chr.9: 134434455-134434455 on Build GRCh38
rs376910846 Chr.9: 134434060-134434060 on Build GRCh38
rs35341799 Chr.9: 134434118-134434118 on Build GRCh38
rs976523092 Chr.9: 134434402-134434402 on Build GRCh38
rs748854470 Chr.9: 134434054-134434054 on Build GRCh38
rs772554977 Chr.9: 134434065-134434065 on Build GRCh38
rs771200755 Chr.9: 134434083-134434083 on Build GRCh38
rs1047093232 Chr.9: 134434421-134434421 on Build GRCh38
rs769848478 Chr.9: 134434217-134434217 on Build GRCh38
rs760744198 Chr.9: 134434081-134434081 on Build GRCh38
rs569208250 Chr.9: 134434382-134434382 on Build GRCh38
rs1023146805 Chr.9: 134434284-134434284 on Build GRCh38
rs764775603 Chr.9: 134434247-134434247 on Build GRCh38
rs773784147 Chr.9: 134434068-134434068 on Build GRCh38
rs995119165 Chr.9: 134434341-134434341 on Build GRCh38
rs11542209 Chr.9: 134434159-134434159 on Build GRCh38
rs1043425285 Chr.9: 134434233-134434233 on Build GRCh38
rs970676936 Chr.9: 134434300-134434300 on Build GRCh38
rs146783648 Chr.9: 134434256-134434256 on Build GRCh38
rs778625488 Chr.9: 134434160-134434160 on Build GRCh38
rs1027038441 Chr.9: 134434344-134434344 on Build GRCh38
rs34254499 Chr.9: 134434271-134434271 on Build GRCh38
rs375446455 Chr.9: 134434242-134434242 on Build GRCh38
rs937627971 Chr.9: 134434020-134434020 on Build GRCh38
rs942525582 Chr.9: 134434403-134434403 on Build GRCh38
rs949042901 Chr.9: 134434044-134434044 on Build GRCh38
rs748664039 Chr.9: 134434163-134434163 on Build GRCh38
rs1026026986 Chr.9: 134434221-134434221 on Build GRCh38
rs540374192 Chr.9: 134434128-134434128 on Build GRCh38
rs1034383220 Chr.9: 134434435-134434435 on Build GRCh38
rs901093038 Chr.9: 134434409-134434409 on Build GRCh38
rs778229173 Chr.9: 134434180-134434180 on Build GRCh38
rs747426420 Chr.9: 134434197-134434197 on Build GRCh38
rs755448808 Chr.9: 134434178-134434178 on Build GRCh38
rs567174354 Chr.9: 134434440-134434440 on Build GRCh38
rs150767145 Chr.9: 134434105-134434105 on Build GRCh38
rs35551025 Chr.9: 134434012-134434012 on Build GRCh38
rs540248051 Chr.9: 134434023-134434023 on Build GRCh38
rs888441084 Chr.9: 134434422-134434422 on Build GRCh38
rs1005658471 Chr.9: 134434434-134434434 on Build GRCh38
rs766139154 Chr.9: 134434237-134434237 on Build GRCh38
rs952434255 Chr.9: 134434346-134434346 on Build GRCh38
rs755663805 Chr.9: 134434125-134434125 on Build GRCh38
rs760488487 Chr.9: 134434235-134434235 on Build GRCh38
rs1006838045 Chr.9: 134434117-134434117 on Build GRCh38
rs746123251 Chr.9: 134434216-134434216 on Build GRCh38
rs761669692 Chr.9: 134434222-134434222 on Build GRCh38
rs548043017 Chr.9: 134434249-134434249 on Build GRCh38
rs776522092 Chr.9: 134434087-134434087 on Build GRCh38
rs908371905 Chr.9: 134434130-134434130 on Build GRCh38
rs536863536 Chr.9: 134434415-134434415 on Build GRCh38
rs779356437 Chr.9: 134434200-134434200 on Build GRCh38
rs921122373 Chr.9: 134434405-134434405 on Build GRCh38
rs35272031 Chr.9: 134434053-134434053 on Build GRCh38
rs190758119 Chr.9: 134434139-134434139 on Build GRCh38
rs994199952 Chr.9: 134434181-134434181 on Build GRCh38
rs901061760 Chr.9: 134434156-134434156 on Build GRCh38
rs895864421 Chr.9: 134434447-134434447 on Build GRCh38
rs977785869 Chr.9: 134434328-134434328 on Build GRCh38
rs367888804 Chr.9: 134434308-134434308 on Build GRCh38
rs779531935 Chr.9: 134434061-134434061 on Build GRCh38
rs368586400 Chr.9: 134434113-134434113 on Build GRCh38
rs527556712 Chr.9: 134434260-134434260 on Build GRCh38
rs554829061 Chr.9: 134434439-134434439 on Build GRCh38
rs917510385 Chr.9: 134434291-134434291 on Build GRCh38
rs889622629 Chr.9: 134434082-134434082 on Build GRCh38
rs753474465 Chr.9: 134434138-134434138 on Build GRCh38
rs765336165 Chr.9: 134434097-134434097 on Build GRCh38
rs34630192 Chr.9: 134434026-134434026 on Build GRCh38
rs1043003741 Chr.9: 134434413-134434413 on Build GRCh38

View Full Product Details