Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 49323875-49324373 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C10orf71
Entrez Gene ID
118461
Gene Name
chromosome 10 open reading frame 71
Gene Aliases
-
UniGene
Hs.585480
Gene Chromosome Location
Chr.10: 49297009-49330145 on Build GRCh38
Transcripts
NM_001135196.1 XM_005269476.4 XM_005269477.3 XM_005269478.4 XM_017015619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5441489 c.1735G>C p.D579H Chr.10: 49324280-49324280 on Build GRCh38
918622 c.1339C>A p.L447M Chr.10: 49323884-49323884 on Build GRCh38
5605571 c.1603C>G p.Q535E Chr.10: 49324148-49324148 on Build GRCh38
3867341 c.1406C>T p.P469L Chr.10: 49323951-49323951 on Build GRCh38
3438353 c.1646A>G p.E549G Chr.10: 49324191-49324191 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs45554335 Chr.10: 49323927-49323927 on Build GRCh38
rs749103332 Chr.10: 49324042-49324042 on Build GRCh38
rs143544986 Chr.10: 49324159-49324159 on Build GRCh38
rs774837674 Chr.10: 49324224-49324224 on Build GRCh38
rs763325120 Chr.10: 49324175-49324175 on Build GRCh38
rs376094825 Chr.10: 49324299-49324299 on Build GRCh38
rs762918373 Chr.10: 49323945-49323945 on Build GRCh38
rs746515860 Chr.10: 49324237-49324237 on Build GRCh38
rs760760407 Chr.10: 49324300-49324300 on Build GRCh38
rs766893593 Chr.10: 49324182-49324182 on Build GRCh38
rs772077867 Chr.10: 49324134-49324134 on Build GRCh38
rs568594834 Chr.10: 49324093-49324093 on Build GRCh38
rs201857646 Chr.10: 49324325-49324325 on Build GRCh38
rs141342139 Chr.10: 49323974-49323974 on Build GRCh38
rs1024869493 Chr.10: 49324350-49324350 on Build GRCh38
rs550545516 Chr.10: 49324065-49324065 on Build GRCh38
rs760558125 Chr.10: 49324133-49324144 on Build GRCh38
rs1009712254 Chr.10: 49324270-49324270 on Build GRCh38
rs770803940 Chr.10: 49323948-49323948 on Build GRCh38
rs371550755 Chr.10: 49324330-49324330 on Build GRCh38
rs771359282 Chr.10: 49324277-49324277 on Build GRCh38
rs746534437 Chr.10: 49324137-49324137 on Build GRCh38
rs767531345 Chr.10: 49324261-49324261 on Build GRCh38
rs554031001 Chr.10: 49324064-49324064 on Build GRCh38
rs776686854 Chr.10: 49324273-49324274 on Build GRCh38
rs767196896 Chr.10: 49323958-49323958 on Build GRCh38
rs138877908 Chr.10: 49323910-49323910 on Build GRCh38
rs767625364 Chr.10: 49324297-49324297 on Build GRCh38
rs765766954 Chr.10: 49324081-49324081 on Build GRCh38
rs775096407 Chr.10: 49324136-49324136 on Build GRCh38
rs1057168979 Chr.10: 49324051-49324051 on Build GRCh38
rs774488701 Chr.10: 49324215-49324215 on Build GRCh38
rs758338800 Chr.10: 49324319-49324319 on Build GRCh38
rs757046894 Chr.10: 49324203-49324203 on Build GRCh38
rs759592630 Chr.10: 49324294-49324294 on Build GRCh38
rs750453040 Chr.10: 49324004-49324004 on Build GRCh38
rs992105882 Chr.10: 49324343-49324343 on Build GRCh38
rs770842902 Chr.10: 49324060-49324060 on Build GRCh38
rs755143970 Chr.10: 49324100-49324100 on Build GRCh38
rs867354143 Chr.10: 49323954-49323954 on Build GRCh38
rs895506574 Chr.10: 49324271-49324271 on Build GRCh38
rs889329877 Chr.10: 49323946-49323946 on Build GRCh38
rs924394600 Chr.10: 49324205-49324205 on Build GRCh38
rs752456405 Chr.10: 49323963-49323963 on Build GRCh38
rs569703214 Chr.10: 49324030-49324030 on Build GRCh38
rs778532590 Chr.10: 49324225-49324225 on Build GRCh38
rs972167545 Chr.10: 49324120-49324120 on Build GRCh38
rs114069700 Chr.10: 49324167-49324167 on Build GRCh38
rs376302106 Chr.10: 49324084-49324084 on Build GRCh38
rs374214345 Chr.10: 49323973-49323973 on Build GRCh38
rs764484472 Chr.10: 49324218-49324218 on Build GRCh38
rs755054038 Chr.10: 49324190-49324190 on Build GRCh38
rs972450410 Chr.10: 49323913-49323913 on Build GRCh38
rs116202086 Chr.10: 49324336-49324336 on Build GRCh38
rs780129738 Chr.10: 49324320-49324320 on Build GRCh38
rs201064215 Chr.10: 49323992-49323992 on Build GRCh38
rs947599528 Chr.10: 49324183-49324183 on Build GRCh38
rs777485597 Chr.10: 49324346-49324346 on Build GRCh38
rs764393943 Chr.10: 49324308-49324308 on Build GRCh38
rs923314523 Chr.10: 49324147-49324147 on Build GRCh38
rs181600390 Chr.10: 49324061-49324061 on Build GRCh38
rs148337814 Chr.10: 49324244-49324244 on Build GRCh38
rs777081340 Chr.10: 49324079-49324079 on Build GRCh38
rs774903024 Chr.10: 49324286-49324286 on Build GRCh38
rs910744966 Chr.10: 49323968-49323968 on Build GRCh38
rs377285836 Chr.10: 49324040-49324040 on Build GRCh38
rs200257260 Chr.10: 49324106-49324106 on Build GRCh38
rs762091617 Chr.10: 49324080-49324080 on Build GRCh38
rs772911645 Chr.10: 49323956-49323956 on Build GRCh38
rs758472555 Chr.10: 49324011-49324011 on Build GRCh38
rs775875033 Chr.10: 49324071-49324071 on Build GRCh38
rs768337422 Chr.10: 49324241-49324241 on Build GRCh38
rs1005768469 Chr.10: 49324088-49324088 on Build GRCh38
rs201677101 Chr.10: 49324070-49324070 on Build GRCh38
rs750947211 Chr.10: 49324083-49324083 on Build GRCh38
rs747276832 Chr.10: 49324032-49324032 on Build GRCh38
rs758980501 Chr.10: 49323951-49323951 on Build GRCh38
rs773106985 Chr.10: 49323944-49323944 on Build GRCh38
rs749458818 Chr.10: 49324165-49324165 on Build GRCh38
rs994013054 Chr.10: 49324344-49324344 on Build GRCh38
rs557712493 Chr.10: 49324246-49324246 on Build GRCh38
rs754504230 Chr.10: 49324329-49324329 on Build GRCh38
rs61453891 Chr.10: 49324110-49324110 on Build GRCh38
rs778888907 Chr.10: 49324118-49324118 on Build GRCh38
rs903901860 Chr.10: 49324323-49324323 on Build GRCh38
rs750258938 Chr.10: 49324228-49324228 on Build GRCh38
rs751670172 Chr.10: 49324099-49324099 on Build GRCh38
rs866295870 Chr.10: 49324245-49324245 on Build GRCh38
rs902909346 Chr.10: 49323989-49323989 on Build GRCh38
rs781404544 Chr.10: 49324105-49324105 on Build GRCh38
rs769272748 Chr.10: 49324337-49324337 on Build GRCh38
rs751580350 Chr.10: 49324189-49324189 on Build GRCh38
rs766482465 Chr.10: 49324098-49324098 on Build GRCh38
rs780148185 Chr.10: 49324232-49324232 on Build GRCh38
rs763388167 Chr.10: 49324259-49324259 on Build GRCh38
rs559200171 Chr.10: 49323931-49323931 on Build GRCh38
rs889444095 Chr.10: 49324242-49324242 on Build GRCh38
rs766204439 Chr.10: 49324211-49324214 on Build GRCh38
rs758357160 Chr.10: 49324229-49324229 on Build GRCh38
rs185555584 Chr.10: 49324257-49324257 on Build GRCh38
rs201075807 Chr.10: 49323911-49323911 on Build GRCh38
rs757737048 Chr.10: 49323986-49323986 on Build GRCh38
rs368697466 Chr.10: 49324119-49324119 on Build GRCh38
rs752912795 Chr.10: 49324197-49324197 on Build GRCh38
rs767721099 Chr.10: 49324193-49324193 on Build GRCh38

View Full Product Details