Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 9021636-9022072 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCUBE2
Entrez Gene ID
57758
Gene Name
signal peptide, CUB domain and EGF like domain containing 2
Gene Aliases
CEGB1, CEGF1, CEGP1
UniGene
Hs.523468
Gene Chromosome Location
Chr.11: 9019500-9091603 on Build GRCh38
Transcripts
NM_001170690.1 NM_020974.2 XM_005253032.4 XM_005253033.4 XM_005253034.4 XM_005253035.4 XM_005253036.4 XM_005253037.4 XM_011520246.2 XM_011520247.2 XM_017018080.1 XM_017018081.1 XM_017018082.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
251118 c.2846A>G p.K949R Chr.11: 9021877-9021877 on Build GRCh38
6409594 c.2894G>T p.G965V Chr.11: 9021916-9021916 on Build GRCh38
467538 c.2800C>A p.R934R Chr.11: 9021923-9021923 on Build GRCh38
6409593 c.2807G>T p.G936V Chr.11: 9021916-9021916 on Build GRCh38
5349380 c.2933A>G p.K978R Chr.11: 9021877-9021877 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769404173 Chr.11: 9021980-9021980 on Build GRCh38
rs931660609 Chr.11: 9022031-9022031 on Build GRCh38
rs866848221 Chr.11: 9021895-9021895 on Build GRCh38
rs75002200 Chr.11: 9021971-9021971 on Build GRCh38
rs752650702 Chr.11: 9021912-9021912 on Build GRCh38
rs757661901 Chr.11: 9021942-9021942 on Build GRCh38
rs150107896 Chr.11: 9021951-9021951 on Build GRCh38
rs762889564 Chr.11: 9021879-9021879 on Build GRCh38
rs762447560 Chr.11: 9021876-9021877 on Build GRCh38
rs373064208 Chr.11: 9021858-9021858 on Build GRCh38
rs766055704 Chr.11: 9021888-9021888 on Build GRCh38
rs756184692 Chr.11: 9021922-9021922 on Build GRCh38
rs777897025 Chr.11: 9021989-9021989 on Build GRCh38
rs760519067 Chr.11: 9021992-9021992 on Build GRCh38
rs922446549 Chr.11: 9021765-9021765 on Build GRCh38
rs768421970 Chr.11: 9021922-9021925 on Build GRCh38
rs1053355961 Chr.11: 9021734-9021734 on Build GRCh38
rs188492769 Chr.11: 9021827-9021827 on Build GRCh38
rs749197879 Chr.11: 9021991-9021991 on Build GRCh38
rs751247297 Chr.11: 9021962-9021962 on Build GRCh38
rs533128958 Chr.11: 9021805-9021805 on Build GRCh38
rs749030840 Chr.11: 9021795-9021795 on Build GRCh38
rs907453241 Chr.11: 9021862-9021862 on Build GRCh38
rs571508363 Chr.11: 9021773-9021773 on Build GRCh38
rs961428492 Chr.11: 9021797-9021797 on Build GRCh38
rs370986866 Chr.11: 9021923-9021923 on Build GRCh38
rs780760329 Chr.11: 9021972-9021972 on Build GRCh38
rs754324991 Chr.11: 9021932-9021932 on Build GRCh38
rs774662035 Chr.11: 9021903-9021903 on Build GRCh38
rs575610034 Chr.11: 9021719-9021719 on Build GRCh38
rs202120189 Chr.11: 9021828-9021828 on Build GRCh38
rs376369922 Chr.11: 9021907-9021907 on Build GRCh38
rs900207224 Chr.11: 9022008-9022008 on Build GRCh38
rs761241578 Chr.11: 9022017-9022017 on Build GRCh38
rs768244262 Chr.11: 9021843-9021843 on Build GRCh38
rs779107141 Chr.11: 9021961-9021961 on Build GRCh38
rs773721450 Chr.11: 9022007-9022010 on Build GRCh38
rs773267262 Chr.11: 9021864-9021864 on Build GRCh38
rs367701167 Chr.11: 9021976-9021976 on Build GRCh38
rs1017026339 Chr.11: 9021974-9021974 on Build GRCh38
rs933834638 Chr.11: 9021770-9021770 on Build GRCh38
rs893396516 Chr.11: 9021739-9021739 on Build GRCh38
rs974080329 Chr.11: 9021806-9021806 on Build GRCh38
rs745886978 Chr.11: 9021687-9021687 on Build GRCh38
rs370979204 Chr.11: 9021829-9021829 on Build GRCh38
rs185397645 Chr.11: 9021772-9021772 on Build GRCh38
rs909511579 Chr.11: 9021690-9021690 on Build GRCh38
rs140739403 Chr.11: 9021908-9021908 on Build GRCh38
rs997469252 Chr.11: 9021743-9021743 on Build GRCh38
rs544550537 Chr.11: 9021720-9021720 on Build GRCh38
rs1036922310 Chr.11: 9021760-9021760 on Build GRCh38
rs769918400 Chr.11: 9021861-9021861 on Build GRCh38
rs775957120 Chr.11: 9021851-9021851 on Build GRCh38

View Full Product Details