Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 67367382-67367889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCF1
Entrez Gene ID
23529
Gene Name
cardiotrophin-like cytokine factor 1
Gene Aliases
BSF-3, BSF3, CISS2, CLC, NNT-1, NNT1, NR6
UniGene
Hs.502977
Gene Chromosome Location
Chr.11: 67364168-67374177 on Build GRCh38
Transcripts
NM_001166212.1 NM_013246.2 NR_024469.1

Gene Symbol
LOC100130987
Entrez Gene ID
100130987
Gene Name
uncharacterized LOC100130987
Gene Aliases
-
UniGene
Hs.664585
Gene Chromosome Location
Chr.11: 67317839-67391687 on Build GRCh38
Transcripts
NM_001166212.1 NM_013246.2 NR_024469.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1356381 c.172G>A p.A58T Chr.11: 67367471-67367471 on Build GRCh38
4146196 c.138C>A p.L46L Chr.11: 67367505-67367505 on Build GRCh38
7275352 c.103G>T p.G35W Chr.11: 67367540-67367540 on Build GRCh38
4167596 c.31A>G p.M11V Chr.11: 67367612-67367612 on Build GRCh38
4165926 c.55C>T p.L19F Chr.11: 67367588-67367588 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765171910 Chr.11: 67367649-67367649 on Build GRCh38
rs959399691 Chr.11: 67367841-67367841 on Build GRCh38
rs752740163 Chr.11: 67367589-67367589 on Build GRCh38
rs140222264 Chr.11: 67367614-67367614 on Build GRCh38
rs753589757 Chr.11: 67367568-67367568 on Build GRCh38
rs777940053 Chr.11: 67367492-67367492 on Build GRCh38
rs958190824 Chr.11: 67367576-67367576 on Build GRCh38
rs982233902 Chr.11: 67367484-67367484 on Build GRCh38
rs927990126 Chr.11: 67367817-67367817 on Build GRCh38
rs757479653 Chr.11: 67367499-67367499 on Build GRCh38
rs759391139 Chr.11: 67367646-67367646 on Build GRCh38
rs137879055 Chr.11: 67367464-67367464 on Build GRCh38
rs909719937 Chr.11: 67367794-67367794 on Build GRCh38
rs141954460 Chr.11: 67367619-67367619 on Build GRCh38
rs751692316 Chr.11: 67367494-67367494 on Build GRCh38
rs113591556 Chr.11: 67367432-67367432 on Build GRCh38
rs758844365 Chr.11: 67367663-67367663 on Build GRCh38
rs576667161 Chr.11: 67367409-67367409 on Build GRCh38
rs1018584537 Chr.11: 67367534-67367534 on Build GRCh38
rs765259754 Chr.11: 67367580-67367580 on Build GRCh38
rs140759579 Chr.11: 67367531-67367531 on Build GRCh38
rs982207756 Chr.11: 67367799-67367802 on Build GRCh38
rs541825951 Chr.11: 67367469-67367469 on Build GRCh38
rs116231318 Chr.11: 67367665-67367665 on Build GRCh38
rs184518539 Chr.11: 67367539-67367539 on Build GRCh38
rs532257038 Chr.11: 67367745-67367745 on Build GRCh38
rs369594545 Chr.11: 67367500-67367500 on Build GRCh38
rs761910171 Chr.11: 67367674-67367674 on Build GRCh38
rs200379623 Chr.11: 67367592-67367592 on Build GRCh38
rs749650966 Chr.11: 67367613-67367613 on Build GRCh38
rs776491928 Chr.11: 67367644-67367644 on Build GRCh38
rs1033768746 Chr.11: 67367709-67367709 on Build GRCh38
rs748605837 Chr.11: 67367660-67367660 on Build GRCh38
rs201269564 Chr.11: 67367503-67367503 on Build GRCh38
rs780404936 Chr.11: 67367610-67367610 on Build GRCh38
rs576604539 Chr.11: 67367664-67367664 on Build GRCh38
rs201618590 Chr.11: 67367658-67367658 on Build GRCh38
rs750104329 Chr.11: 67367450-67367450 on Build GRCh38
rs770492996 Chr.11: 67367643-67367643 on Build GRCh38
rs781167028 Chr.11: 67367598-67367598 on Build GRCh38
rs143166489 Chr.11: 67367414-67367414 on Build GRCh38
rs766150197 Chr.11: 67367561-67367561 on Build GRCh38
rs145282707 Chr.11: 67367615-67367615 on Build GRCh38
rs766128849 Chr.11: 67367466-67367466 on Build GRCh38
rs369972181 Chr.11: 67367673-67367673 on Build GRCh38
rs560558450 Chr.11: 67367480-67367480 on Build GRCh38
rs369213496 Chr.11: 67367605-67367605 on Build GRCh38
rs574363993 Chr.11: 67367712-67367712 on Build GRCh38
rs562468314 Chr.11: 67367428-67367428 on Build GRCh38
rs767157151 Chr.11: 67367447-67367447 on Build GRCh38
rs867193 Chr.11: 67367535-67367535 on Build GRCh38
rs760318893 Chr.11: 67367637-67367637 on Build GRCh38
rs76168219 Chr.11: 67367555-67367555 on Build GRCh38
rs137853934 Chr.11: 67367597-67367597 on Build GRCh38
rs772890565 Chr.11: 67367554-67367554 on Build GRCh38
rs775609571 Chr.11: 67367442-67367443 on Build GRCh38
rs763405129 Chr.11: 67367423-67367423 on Build GRCh38
rs759478800 Chr.11: 67367577-67367577 on Build GRCh38
rs112879451 Chr.11: 67367666-67367666 on Build GRCh38
rs569006019 Chr.11: 67367419-67367419 on Build GRCh38
rs775534310 Chr.11: 67367652-67367652 on Build GRCh38
rs756771030 Chr.11: 67367805-67367805 on Build GRCh38
rs547690023 Chr.11: 67367789-67367789 on Build GRCh38
rs753771467 Chr.11: 67367479-67367479 on Build GRCh38
rs749743060 Chr.11: 67367510-67367510 on Build GRCh38
rs767583156 Chr.11: 67367655-67367655 on Build GRCh38
rs376831170 Chr.11: 67367674-67367674 on Build GRCh38
rs566471248 Chr.11: 67367623-67367623 on Build GRCh38
rs35361219 Chr.11: 67367698-67367698 on Build GRCh38
rs771570860 Chr.11: 67367621-67367621 on Build GRCh38
rs376191009 Chr.11: 67367604-67367604 on Build GRCh38
rs776038583 Chr.11: 67367421-67367421 on Build GRCh38
rs774478596 Chr.11: 67367627-67367628 on Build GRCh38
rs982545383 Chr.11: 67367695-67367695 on Build GRCh38
rs763011438 Chr.11: 67367653-67367653 on Build GRCh38
rs763515623 Chr.11: 67367765-67367765 on Build GRCh38
rs770489191 Chr.11: 67367659-67367659 on Build GRCh38
rs761391507 Chr.11: 67367446-67367446 on Build GRCh38
rs770160336 Chr.11: 67367502-67367502 on Build GRCh38
rs756287540 Chr.11: 67367656-67367656 on Build GRCh38
rs199499449 Chr.11: 67367657-67367657 on Build GRCh38
rs768669679 Chr.11: 67367619-67367624 on Build GRCh38
rs749497999 Chr.11: 67367447-67367447 on Build GRCh38
rs771535281 Chr.11: 67367544-67367544 on Build GRCh38
rs372255151 Chr.11: 67367501-67367501 on Build GRCh38

View Full Product Details