Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50769029-50769590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RABL2B
Entrez Gene ID
11158
Gene Name
RAB, member of RAS oncogene family-like 2B
Gene Aliases
-
UniGene
Hs.584862
Gene Chromosome Location
Chr.22: 50767492-50783705 on Build GRCh38
Transcripts
NM_001003789.1 NM_001130919.1 NM_001130920.1 NM_001130921.1 NM_001130922.1 NM_001130923.1 NM_007081.2 XM_005261899.2 XM_011530668.1 XM_011530669.2 XM_011530670.1 XM_011530671.1 XM_011530673.1 XM_011530675.1 XM_017028543.1 XM_017028544.1 XM_017028545.1 XM_017028546.1 XM_017028547.1 XM_017028548.1 XM_017028549.1 XM_017028550.1 XM_017028551.1 XM_017028552.1 XM_017028553.1 XM_017028554.1 XM_017028555.1 XM_017028556.1 XM_017028557.1 XM_017028558.1 NR_026981.1

Gene Symbol
RPL23AP82
Entrez Gene ID
284942
Gene Name
ribosomal protein L23a pseudogene 82
Gene Aliases
RPL23A_43_1761
UniGene
Hs.406135
Gene Chromosome Location
Chr.22: 50757086-50799637 on Build GRCh38
Transcripts
NM_001003789.1 NM_001130919.1 NM_001130920.1 NM_001130921.1 NM_001130922.1 NM_001130923.1 NM_007081.2 XM_005261899.2 XM_011530668.1 XM_011530669.2 XM_011530670.1 XM_011530671.1 XM_011530673.1 XM_011530675.1 XM_017028543.1 XM_017028544.1 XM_017028545.1 XM_017028546.1 XM_017028547.1 XM_017028548.1 XM_017028549.1 XM_017028550.1 XM_017028551.1 XM_017028552.1 XM_017028553.1 XM_017028554.1 XM_017028555.1 XM_017028556.1 XM_017028557.1 XM_017028558.1 NR_026981.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3681474 c.504+4T>G p.? Chr.22: 50769451-50769451 on Build GRCh38
5155085 c.430delA p.S144fs*24 Chr.22: 50769529-50769529 on Build GRCh38
3681473 c.504+7T>G p.? Chr.22: 50769448-50769448 on Build GRCh38
4719930 c.418G>A p.V140M Chr.22: 50769541-50769541 on Build GRCh38
1035574 c.524G>A p.R175Q Chr.22: 50769105-50769105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138665583 Chr.22: 50769482-50769482 on Build GRCh38
rs530899616 Chr.22: 50769258-50769258 on Build GRCh38
rs781940096 Chr.22: 50769529-50769529 on Build GRCh38
rs782046766 Chr.22: 50769476-50769476 on Build GRCh38
rs782017790 Chr.22: 50769100-50769100 on Build GRCh38
rs369512280 Chr.22: 50769071-50769071 on Build GRCh38
rs782540820 Chr.22: 50769389-50769389 on Build GRCh38
rs782790899 Chr.22: 50769463-50769463 on Build GRCh38
rs782327033 Chr.22: 50769433-50769433 on Build GRCh38
rs139609882 Chr.22: 50769542-50769542 on Build GRCh38
rs782252555 Chr.22: 50769416-50769416 on Build GRCh38
rs530264176 Chr.22: 50769158-50769158 on Build GRCh38
rs781998539 Chr.22: 50769067-50769067 on Build GRCh38
rs782025466 Chr.22: 50769531-50769531 on Build GRCh38
rs371770888 Chr.22: 50769417-50769417 on Build GRCh38
rs370028333 Chr.22: 50769488-50769488 on Build GRCh38
rs782395656 Chr.22: 50769444-50769444 on Build GRCh38
rs782651550 Chr.22: 50769521-50769521 on Build GRCh38
rs782773645 Chr.22: 50769562-50769562 on Build GRCh38
rs781812282 Chr.22: 50769465-50769465 on Build GRCh38
rs782235529 Chr.22: 50769517-50769517 on Build GRCh38
rs542295368 Chr.22: 50769204-50769204 on Build GRCh38
rs782176113 Chr.22: 50769403-50769403 on Build GRCh38
rs781900215 Chr.22: 50769496-50769496 on Build GRCh38
rs782118071 Chr.22: 50769074-50769074 on Build GRCh38
rs782217658 Chr.22: 50769051-50769051 on Build GRCh38
rs571543803 Chr.22: 50769408-50769408 on Build GRCh38
rs375510798 Chr.22: 50769558-50769558 on Build GRCh38
rs781990792 Chr.22: 50769446-50769446 on Build GRCh38
rs782084698 Chr.22: 50769437-50769437 on Build GRCh38
rs782514294 Chr.22: 50769379-50769379 on Build GRCh38
rs782169155 Chr.22: 50769451-50769451 on Build GRCh38
rs147376674 Chr.22: 50769522-50769522 on Build GRCh38
rs782783535 Chr.22: 50769381-50769381 on Build GRCh38
rs549827645 Chr.22: 50769305-50769305 on Build GRCh38
rs782069543 Chr.22: 50769541-50769541 on Build GRCh38
rs142730268 Chr.22: 50769487-50769487 on Build GRCh38
rs559977293 Chr.22: 50769106-50769106 on Build GRCh38
rs782591141 Chr.22: 50769395-50769395 on Build GRCh38
rs782748475 Chr.22: 50769145-50769145 on Build GRCh38
rs376723096 Chr.22: 50769436-50769436 on Build GRCh38
rs781961192 Chr.22: 50769555-50769555 on Build GRCh38
rs782277299 Chr.22: 50769426-50769426 on Build GRCh38
rs782542291 Chr.22: 50769503-50769503 on Build GRCh38
rs781823725 Chr.22: 50769174-50769174 on Build GRCh38
rs781908870 Chr.22: 50769385-50769385 on Build GRCh38
rs541646021 Chr.22: 50769054-50769054 on Build GRCh38
rs782574342 Chr.22: 50769508-50769508 on Build GRCh38
rs782585369 Chr.22: 50769414-50769414 on Build GRCh38
rs563680639 Chr.22: 50769208-50769208 on Build GRCh38

View Full Product Details