Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60799896-60800114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A10
Entrez Gene ID
341116
Gene Name
membrane spanning 4-domains A10
Gene Aliases
CD20L7, MS4A9
UniGene
Hs.591956
Gene Chromosome Location
Chr.11: 60785332-60801305 on Build GRCh38
Transcripts
NM_206893.3 XM_011544989.1 XM_011544991.2

SNP Detail

SNP ID Chromosome Location
rs74413514 Chr.11: 60800013-60800013 on Build GRCh38
rs745503586 Chr.11: 60800052-60800053 on Build GRCh38
rs971114484 Chr.11: 60799973-60799973 on Build GRCh38
rs748450357 Chr.11: 60799916-60799916 on Build GRCh38
rs986237014 Chr.11: 60800041-60800041 on Build GRCh38
rs1022745828 Chr.11: 60799967-60799967 on Build GRCh38
rs767495207 Chr.11: 60799952-60799952 on Build GRCh38
rs71997819 Chr.11: 60800053-60800057 on Build GRCh38
rs1031303470 Chr.11: 60799986-60799986 on Build GRCh38
rs371159537 Chr.11: 60799945-60799945 on Build GRCh38
rs957183371 Chr.11: 60800001-60800001 on Build GRCh38
rs977699432 Chr.11: 60800017-60800017 on Build GRCh38
rs770038922 Chr.11: 60799932-60799932 on Build GRCh38
rs759581290 Chr.11: 60799948-60799948 on Build GRCh38
rs991858279 Chr.11: 60799955-60799955 on Build GRCh38
rs911048931 Chr.11: 60800060-60800060 on Build GRCh38
rs771094536 Chr.11: 60799936-60799936 on Build GRCh38
rs559711834 Chr.11: 60799984-60799984 on Build GRCh38
rs368751612 Chr.11: 60799915-60799915 on Build GRCh38
rs74932478 Chr.11: 60800061-60800061 on Build GRCh38
rs946907609 Chr.11: 60800033-60800034 on Build GRCh38
rs556364512 Chr.11: 60799939-60799939 on Build GRCh38
rs910648272 Chr.11: 60800049-60800049 on Build GRCh38
rs112381946 Chr.11: 60799975-60799975 on Build GRCh38
rs1050064642 Chr.11: 60800050-60800050 on Build GRCh38
rs763056970 Chr.11: 60799935-60799935 on Build GRCh38
rs773546459 Chr.11: 60799934-60799934 on Build GRCh38
rs117619803 Chr.11: 60800018-60800018 on Build GRCh38
rs936562797 Chr.11: 60800038-60800038 on Build GRCh38
rs917608785 Chr.11: 60799987-60799987 on Build GRCh38
rs981529752 Chr.11: 60799974-60799974 on Build GRCh38
rs539890395 Chr.11: 60799977-60799977 on Build GRCh38

View Full Product Details