Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 42165630-42166129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VPS39
Entrez Gene ID
23339
Gene Name
VPS39, HOPS complex subunit
Gene Aliases
TLP, VAM6, hVam6p
UniGene
Hs.88025
Gene Chromosome Location
Chr.15: 42158701-42208331 on Build GRCh38
Transcripts
NM_001301138.1 NM_015289.3 XM_011521403.2 XM_011521404.2 XM_017022036.1 XM_017022037.1 XM_017022038.1 XM_017022039.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6318855 c.1717C>A p.P573T Chr.15: 42165780-42165780 on Build GRCh38
5546111 c.1771C>T p.P591S Chr.15: 42165726-42165726 on Build GRCh38
5987376 c.1690G>A p.E564K Chr.15: 42165807-42165807 on Build GRCh38
5971406 c.1753T>A p.F585I Chr.15: 42165744-42165744 on Build GRCh38
6318856 c.1750C>A p.P584T Chr.15: 42165780-42165780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014983529 Chr.15: 42166077-42166077 on Build GRCh38
rs944145784 Chr.15: 42165964-42165964 on Build GRCh38
rs747152122 Chr.15: 42165770-42165770 on Build GRCh38
rs780029284 Chr.15: 42165672-42165672 on Build GRCh38
rs187191900 Chr.15: 42165762-42165762 on Build GRCh38
rs570223268 Chr.15: 42165908-42165908 on Build GRCh38
rs781315267 Chr.15: 42165696-42165696 on Build GRCh38
rs200469742 Chr.15: 42165702-42165702 on Build GRCh38
rs754802687 Chr.15: 42165688-42165688 on Build GRCh38
rs781184895 Chr.15: 42165901-42165901 on Build GRCh38
rs541014371 Chr.15: 42165832-42165832 on Build GRCh38
rs748127127 Chr.15: 42165796-42165796 on Build GRCh38
rs182526317 Chr.15: 42165681-42165681 on Build GRCh38
rs529405366 Chr.15: 42165912-42165912 on Build GRCh38
rs561738448 Chr.15: 42165797-42165797 on Build GRCh38
rs767864831 Chr.15: 42165739-42165739 on Build GRCh38
rs777373404 Chr.15: 42165800-42165800 on Build GRCh38
rs371879544 Chr.15: 42165735-42165735 on Build GRCh38
rs1025087511 Chr.15: 42165794-42165794 on Build GRCh38
rs902693481 Chr.15: 42165994-42165994 on Build GRCh38
rs768998826 Chr.15: 42165840-42165840 on Build GRCh38
rs758594711 Chr.15: 42165668-42165668 on Build GRCh38
rs948285144 Chr.15: 42165857-42165857 on Build GRCh38
rs765608707 Chr.15: 42165776-42165776 on Build GRCh38
rs774057291 Chr.15: 42165813-42165813 on Build GRCh38
rs61761618 Chr.15: 42165774-42165774 on Build GRCh38
rs902642158 Chr.15: 42165669-42165669 on Build GRCh38
rs891210547 Chr.15: 42165919-42165919 on Build GRCh38
rs979735716 Chr.15: 42165877-42165877 on Build GRCh38
rs770406403 Chr.15: 42165811-42165811 on Build GRCh38
rs961164260 Chr.15: 42165884-42165884 on Build GRCh38
rs534741944 Chr.15: 42166020-42166020 on Build GRCh38
rs754882709 Chr.15: 42165778-42165778 on Build GRCh38
rs375967273 Chr.15: 42165880-42165880 on Build GRCh38
rs566142029 Chr.15: 42166011-42166011 on Build GRCh38
rs762028798 Chr.15: 42165856-42165856 on Build GRCh38
rs935361928 Chr.15: 42165883-42165883 on Build GRCh38
rs1039540757 Chr.15: 42165971-42165971 on Build GRCh38
rs781117382 Chr.15: 42165793-42165793 on Build GRCh38
rs149704041 Chr.15: 42165763-42165763 on Build GRCh38
rs1052478121 Chr.15: 42165890-42165890 on Build GRCh38
rs932347081 Chr.15: 42165918-42165918 on Build GRCh38
rs943788239 Chr.15: 42165960-42165960 on Build GRCh38
rs1026923477 Chr.15: 42166019-42166019 on Build GRCh38
rs1003891082 Chr.15: 42166053-42166053 on Build GRCh38
rs1042105440 Chr.15: 42165992-42165993 on Build GRCh38
rs201390887 Chr.15: 42165805-42165805 on Build GRCh38
rs539299110 Chr.15: 42166026-42166026 on Build GRCh38
rs555946593 Chr.15: 42166066-42166066 on Build GRCh38
rs549181971 Chr.15: 42165926-42165926 on Build GRCh38
rs916785810 Chr.15: 42165822-42165822 on Build GRCh38
rs1025395540 Chr.15: 42166045-42166045 on Build GRCh38
rs372539690 Chr.15: 42165854-42165854 on Build GRCh38
rs550401543 Chr.15: 42165899-42165899 on Build GRCh38
rs1012289371 Chr.15: 42166030-42166030 on Build GRCh38
rs145146659 Chr.15: 42166021-42166021 on Build GRCh38
rs773500663 Chr.15: 42166071-42166071 on Build GRCh38
rs761277845 Chr.15: 42165833-42165833 on Build GRCh38
rs375035999 Chr.15: 42165771-42165771 on Build GRCh38
rs138951752 Chr.15: 42166022-42166022 on Build GRCh38
rs770529523 Chr.15: 42165732-42165732 on Build GRCh38
rs748634920 Chr.15: 42166006-42166006 on Build GRCh38
rs12595000 Chr.15: 42165881-42165881 on Build GRCh38
rs191703716 Chr.15: 42165777-42165777 on Build GRCh38
rs759934227 Chr.15: 42165736-42165736 on Build GRCh38
rs199516643 Chr.15: 42165729-42165729 on Build GRCh38
rs745507648 Chr.15: 42165827-42165827 on Build GRCh38

View Full Product Details