Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 25822474-25823014 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC646471
Entrez Gene ID
646471
Gene Name
uncharacterized LOC646471
Gene Aliases
-
UniGene
Hs.727271
Gene Chromosome Location
Chr.1: 25819954-25823606 on Build GRCh38
Transcripts
NR_024498.1 NM_001099625.1 NM_001099626.1 NM_001099627.1 NM_019557.5

Gene Symbol
MTFR1L
Entrez Gene ID
56181
Gene Name
mitochondrial fission regulator 1 like
Gene Aliases
FAM54B, HYST1888, MST116, MSTP116
UniGene
Hs.149305
Gene Chromosome Location
Chr.1: 25819906-25832942 on Build GRCh38
Transcripts
NR_024498.1 NM_001099625.1 NM_001099626.1 NM_001099627.1 NM_019557.5

SNP Detail

SNP ID Chromosome Location
rs138804319 Chr.1: 25822540-25822544 on Build GRCh38
rs761204575 Chr.1: 25822826-25822826 on Build GRCh38
rs577835553 Chr.1: 25822943-25822943 on Build GRCh38
rs902871306 Chr.1: 25822762-25822762 on Build GRCh38
rs559800008 Chr.1: 25822639-25822639 on Build GRCh38
rs762845147 Chr.1: 25822744-25822745 on Build GRCh38
rs767207259 Chr.1: 25822522-25822526 on Build GRCh38
rs201965603 Chr.1: 25822864-25822865 on Build GRCh38
rs989598615 Chr.1: 25822949-25822949 on Build GRCh38
rs910391341 Chr.1: 25822956-25822956 on Build GRCh38
rs188942435 Chr.1: 25822769-25822769 on Build GRCh38
rs762811104 Chr.1: 25822972-25822972 on Build GRCh38
rs905692032 Chr.1: 25822726-25822726 on Build GRCh38
rs965535113 Chr.1: 25822889-25822889 on Build GRCh38
rs1044304235 Chr.1: 25822715-25822715 on Build GRCh38
rs869079514 Chr.1: 25822523-25822527 on Build GRCh38
rs776355810 Chr.1: 25822627-25822627 on Build GRCh38
rs920442132 Chr.1: 25822519-25822519 on Build GRCh38
rs915198623 Chr.1: 25822965-25822965 on Build GRCh38
rs1032740499 Chr.1: 25822614-25822614 on Build GRCh38
rs200226491 Chr.1: 25822865-25822866 on Build GRCh38
rs568582783 Chr.1: 25822521-25822525 on Build GRCh38
rs2142624 Chr.1: 25822573-25822573 on Build GRCh38
rs768605369 Chr.1: 25822523-25822528 on Build GRCh38
rs145195575 Chr.1: 25822895-25822895 on Build GRCh38
rs768539938 Chr.1: 25822802-25822802 on Build GRCh38
rs182730972 Chr.1: 25822647-25822647 on Build GRCh38
rs910369474 Chr.1: 25822748-25822748 on Build GRCh38
rs866897577 Chr.1: 25822846-25822846 on Build GRCh38
rs930463892 Chr.1: 25822549-25822549 on Build GRCh38
rs571799856 Chr.1: 25822747-25822747 on Build GRCh38
rs1007117778 Chr.1: 25822778-25822778 on Build GRCh38
rs577978502 Chr.1: 25822673-25822673 on Build GRCh38
rs552059419 Chr.1: 25822736-25822736 on Build GRCh38
rs201004913 Chr.1: 25822699-25822700 on Build GRCh38
rs892924092 Chr.1: 25822777-25822777 on Build GRCh38
rs200467130 Chr.1: 25822608-25822608 on Build GRCh38
rs924063601 Chr.1: 25822504-25822504 on Build GRCh38
rs749583804 Chr.1: 25822521-25822526 on Build GRCh38
rs572194596 Chr.1: 25822894-25822894 on Build GRCh38
rs535584739 Chr.1: 25822734-25822734 on Build GRCh38
rs34239512 Chr.1: 25822767-25822767 on Build GRCh38
rs1013071372 Chr.1: 25822779-25822779 on Build GRCh38
rs902819021 Chr.1: 25822587-25822587 on Build GRCh38
rs559877511 Chr.1: 25822714-25822714 on Build GRCh38
rs184216888 Chr.1: 25822699-25822699 on Build GRCh38
rs998610370 Chr.1: 25822596-25822596 on Build GRCh38
rs192266789 Chr.1: 25822825-25822825 on Build GRCh38
rs972134685 Chr.1: 25822713-25822713 on Build GRCh38
rs184840222 Chr.1: 25822896-25822896 on Build GRCh38
rs889063075 Chr.1: 25822557-25822557 on Build GRCh38
rs3033467 Chr.1: 25822866-25822867 on Build GRCh38
rs188262402 Chr.1: 25822675-25822675 on Build GRCh38
rs755646300 Chr.1: 25822524-25822528 on Build GRCh38
rs1035205419 Chr.1: 25822864-25822864 on Build GRCh38
rs796345322 Chr.1: 25822865-25822866 on Build GRCh38
rs938542389 Chr.1: 25822730-25822730 on Build GRCh38
rs146766336 Chr.1: 25822510-25822510 on Build GRCh38
rs572275124 Chr.1: 25822601-25822601 on Build GRCh38
rs1040278681 Chr.1: 25822758-25822758 on Build GRCh38
rs766272979 Chr.1: 25822978-25822978 on Build GRCh38
rs1031185367 Chr.1: 25822899-25822899 on Build GRCh38
rs555188081 Chr.1: 25822877-25822877 on Build GRCh38
rs976011768 Chr.1: 25822976-25822976 on Build GRCh38
rs2142623 Chr.1: 25822692-25822692 on Build GRCh38
rs376628457 Chr.1: 25822768-25822768 on Build GRCh38
rs867761932 Chr.1: 25822618-25822618 on Build GRCh38
rs780828834 Chr.1: 25822984-25822984 on Build GRCh38
rs940784394 Chr.1: 25822495-25822495 on Build GRCh38
rs1011096580 Chr.1: 25822632-25822632 on Build GRCh38
rs944604098 Chr.1: 25822558-25822558 on Build GRCh38
rs935340498 Chr.1: 25822667-25822667 on Build GRCh38
rs965847120 Chr.1: 25822973-25822973 on Build GRCh38
rs1050671997 Chr.1: 25822550-25822550 on Build GRCh38
rs758904111 Chr.1: 25822525-25822528 on Build GRCh38
rs751516627 Chr.1: 25822801-25822801 on Build GRCh38
rs555124733 Chr.1: 25822590-25822590 on Build GRCh38
rs190537866 Chr.1: 25822983-25822983 on Build GRCh38
rs192545096 Chr.1: 25822677-25822677 on Build GRCh38
rs962255135 Chr.1: 25822872-25822872 on Build GRCh38
rs34408316 Chr.1: 25822983-25822984 on Build GRCh38
rs569444499 Chr.1: 25822818-25822818 on Build GRCh38
rs775124819 Chr.1: 25822716-25822716 on Build GRCh38
rs944613883 Chr.1: 25822749-25822749 on Build GRCh38
rs1023810228 Chr.1: 25822805-25822805 on Build GRCh38
rs972083665 Chr.1: 25822499-25822499 on Build GRCh38
rs200192608 Chr.1: 25822867-25822867 on Build GRCh38
rs145662884 Chr.1: 25822810-25822810 on Build GRCh38

View Full Product Details