Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 40026541-40027035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MED24
Entrez Gene ID
9862
Gene Name
mediator complex subunit 24
Gene Aliases
ARC100, CRSP100, CRSP4, DRIP100, MED5, THRAP4, TRAP100
UniGene
Hs.462983
Gene Chromosome Location
Chr.17: 40019097-40054636 on Build GRCh38
Transcripts
NM_001079518.1 NM_001267797.1 NM_014815.3 NR_052017.1 XM_005257874.2 XM_006722204.2 XM_006722206.2 XM_006722207.2 XM_011525529.2 XM_011525530.2 XM_011525531.2 XM_011525532.2 XM_011525533.2 XM_011525534.2 XM_011525535.2 XM_011525536.2 XM_011525538.2 XM_017025458.1 XM_017025459.1 XM_017025460.1 XM_017025461.1 XM_017025462.1 XM_017025463.1 XM_017025464.1 XM_017025465.1 XM_017025466.1 XM_017025467.1 XM_017025468.1 XM_017025469.1 XM_017025470.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5824458 c.1732T>A p.C578S Chr.17: 40026724-40026724 on Build GRCh38
7075983 c.1643G>C p.R548P Chr.17: 40026922-40026922 on Build GRCh38
4640099 c.1792G>A p.A598T Chr.17: 40026664-40026664 on Build GRCh38
78074 c.1802C>T p.S601F Chr.17: 40026654-40026654 on Build GRCh38
2838543 c.1797C>T p.F599F Chr.17: 40026659-40026659 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757828252 Chr.17: 40026973-40026973 on Build GRCh38
rs144413662 Chr.17: 40026574-40026574 on Build GRCh38
rs764229846 Chr.17: 40026810-40026810 on Build GRCh38
rs749833557 Chr.17: 40026750-40026750 on Build GRCh38
rs759572786 Chr.17: 40026790-40026790 on Build GRCh38
rs756400726 Chr.17: 40026821-40026821 on Build GRCh38
rs776882718 Chr.17: 40026601-40026601 on Build GRCh38
rs762994793 Chr.17: 40027016-40027016 on Build GRCh38
rs956291260 Chr.17: 40026592-40026592 on Build GRCh38
rs191409548 Chr.17: 40027015-40027015 on Build GRCh38
rs756011087 Chr.17: 40026638-40026638 on Build GRCh38
rs767673387 Chr.17: 40026916-40026916 on Build GRCh38
rs11555252 Chr.17: 40026928-40026928 on Build GRCh38
rs765803564 Chr.17: 40026736-40026736 on Build GRCh38
rs746346700 Chr.17: 40026602-40026602 on Build GRCh38
rs781259172 Chr.17: 40026817-40026817 on Build GRCh38
rs138398755 Chr.17: 40026663-40026663 on Build GRCh38
rs2302775 Chr.17: 40026591-40026591 on Build GRCh38
rs138072890 Chr.17: 40026922-40026922 on Build GRCh38
rs757444295 Chr.17: 40026816-40026816 on Build GRCh38
rs779376836 Chr.17: 40026940-40026940 on Build GRCh38
rs745646252 Chr.17: 40026998-40026998 on Build GRCh38
rs972501635 Chr.17: 40026995-40026995 on Build GRCh38
rs998474520 Chr.17: 40026814-40026814 on Build GRCh38
rs770208959 Chr.17: 40026604-40026604 on Build GRCh38
rs985217134 Chr.17: 40026838-40026838 on Build GRCh38
rs773869689 Chr.17: 40026753-40026753 on Build GRCh38
rs201006975 Chr.17: 40026625-40026625 on Build GRCh38
rs370375402 Chr.17: 40026681-40026681 on Build GRCh38
rs2302774 Chr.17: 40026837-40026837 on Build GRCh38
rs377622090 Chr.17: 40026918-40026918 on Build GRCh38
rs147382251 Chr.17: 40026877-40026877 on Build GRCh38
rs765508362 Chr.17: 40026767-40026769 on Build GRCh38
rs142736212 Chr.17: 40026715-40026715 on Build GRCh38
rs773522997 Chr.17: 40026768-40026768 on Build GRCh38
rs780418870 Chr.17: 40026725-40026725 on Build GRCh38
rs777369383 Chr.17: 40026977-40026977 on Build GRCh38
rs762434715 Chr.17: 40026608-40026608 on Build GRCh38
rs11555253 Chr.17: 40026704-40026704 on Build GRCh38
rs756379864 Chr.17: 40026917-40026917 on Build GRCh38
rs751913595 Chr.17: 40027006-40027006 on Build GRCh38
rs576634349 Chr.17: 40026801-40026801 on Build GRCh38
rs766238553 Chr.17: 40026787-40026787 on Build GRCh38
rs761411330 Chr.17: 40026762-40026762 on Build GRCh38
rs760385490 Chr.17: 40026777-40026777 on Build GRCh38
rs770587060 Chr.17: 40026852-40026852 on Build GRCh38
rs750723310 Chr.17: 40026819-40026819 on Build GRCh38
rs772711836 Chr.17: 40026845-40026845 on Build GRCh38
rs778053085 Chr.17: 40026659-40026659 on Build GRCh38
rs771625386 Chr.17: 40026774-40026774 on Build GRCh38
rs749745106 Chr.17: 40026823-40026823 on Build GRCh38
rs751786358 Chr.17: 40026665-40026665 on Build GRCh38
rs201473936 Chr.17: 40026605-40026605 on Build GRCh38
rs773799134 Chr.17: 40026989-40026989 on Build GRCh38
rs1044995681 Chr.17: 40026575-40026575 on Build GRCh38
rs772999759 Chr.17: 40026775-40026775 on Build GRCh38
rs773407700 Chr.17: 40026815-40026817 on Build GRCh38
rs755417753 Chr.17: 40026933-40026933 on Build GRCh38
rs142434171 Chr.17: 40026990-40026990 on Build GRCh38
rs918451858 Chr.17: 40027004-40027004 on Build GRCh38
rs1006690309 Chr.17: 40026668-40026668 on Build GRCh38
rs904617202 Chr.17: 40026786-40026786 on Build GRCh38
rs763318220 Chr.17: 40026796-40026796 on Build GRCh38
rs759480952 Chr.17: 40026873-40026873 on Build GRCh38
rs776603016 Chr.17: 40026866-40026866 on Build GRCh38
rs1031844172 Chr.17: 40026800-40026800 on Build GRCh38
rs781553190 Chr.17: 40026690-40026690 on Build GRCh38
rs780446078 Chr.17: 40026822-40026822 on Build GRCh38
rs773961718 Chr.17: 40026621-40026621 on Build GRCh38
rs543207953 Chr.17: 40026656-40026656 on Build GRCh38
rs370938302 Chr.17: 40026971-40026971 on Build GRCh38
rs376702531 Chr.17: 40026628-40026628 on Build GRCh38
rs752740552 Chr.17: 40026903-40026903 on Build GRCh38
rs771666982 Chr.17: 40026841-40026841 on Build GRCh38
rs760427361 Chr.17: 40026849-40026849 on Build GRCh38
rs770712637 Chr.17: 40026996-40026996 on Build GRCh38
rs148797154 Chr.17: 40026716-40026716 on Build GRCh38
rs768130849 Chr.17: 40026751-40026751 on Build GRCh38
rs769781486 Chr.17: 40027010-40027010 on Build GRCh38
rs926133816 Chr.17: 40026923-40026923 on Build GRCh38
rs763658379 Chr.17: 40026614-40026614 on Build GRCh38
rs769179547 Chr.17: 40026830-40026830 on Build GRCh38
rs377238436 Chr.17: 40026639-40026639 on Build GRCh38
rs763105153 Chr.17: 40026874-40026875 on Build GRCh38
rs750212516 Chr.17: 40026632-40026632 on Build GRCh38
rs376105540 Chr.17: 40026840-40026840 on Build GRCh38
rs962511455 Chr.17: 40026847-40026847 on Build GRCh38
rs746233458 Chr.17: 40026706-40026706 on Build GRCh38
rs559819787 Chr.17: 40026912-40026912 on Build GRCh38
rs751694334 Chr.17: 40026815-40026815 on Build GRCh38
rs769640357 Chr.17: 40026724-40026725 on Build GRCh38
rs199995954 Chr.17: 40026641-40026641 on Build GRCh38
rs972605478 Chr.17: 40026689-40026689 on Build GRCh38
rs918481023 Chr.17: 40026707-40026707 on Build GRCh38

View Full Product Details