Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 3033268-3033765 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARSH
Entrez Gene ID
347527
Gene Name
arylsulfatase family member H
Gene Aliases
sulfatase
UniGene
Hs.351533
Gene Chromosome Location
Chr.X: 3006613-3033385 on Build GRCh38
Transcripts
NM_001011719.1

SNP Detail

SNP ID Chromosome Location
rs774118629 Chr.X: 3033401-3033401 on Build GRCh38
rs1000380164 Chr.X: 3033484-3033484 on Build GRCh38
rs755599706 Chr.X: 3033368-3033368 on Build GRCh38
rs970396921 Chr.X: 3033547-3033547 on Build GRCh38
rs904753804 Chr.X: 3033442-3033442 on Build GRCh38
rs751829033 Chr.X: 3033595-3033595 on Build GRCh38
rs770502968 Chr.X: 3033394-3033394 on Build GRCh38
rs935402576 Chr.X: 3033624-3033624 on Build GRCh38
rs568718357 Chr.X: 3033671-3033671 on Build GRCh38
rs760554820 Chr.X: 3033437-3033437 on Build GRCh38
rs138812517 Chr.X: 3033306-3033306 on Build GRCh38
rs139634852 Chr.X: 3033361-3033361 on Build GRCh38
rs1001809405 Chr.X: 3033571-3033571 on Build GRCh38
rs146321466 Chr.X: 3033355-3033355 on Build GRCh38
rs748842703 Chr.X: 3033629-3033629 on Build GRCh38
rs766702626 Chr.X: 3033330-3033330 on Build GRCh38
rs893916564 Chr.X: 3033730-3033730 on Build GRCh38
rs754761382 Chr.X: 3033492-3033492 on Build GRCh38
rs752649661 Chr.X: 3033578-3033578 on Build GRCh38
rs921174058 Chr.X: 3033553-3033553 on Build GRCh38
rs932658237 Chr.X: 3033717-3033717 on Build GRCh38
rs1045239985 Chr.X: 3033737-3033737 on Build GRCh38
rs745728178 Chr.X: 3033409-3033409 on Build GRCh38
rs371265926 Chr.X: 3033372-3033372 on Build GRCh38
rs751990184 Chr.X: 3033346-3033346 on Build GRCh38
rs780920387 Chr.X: 3033543-3033543 on Build GRCh38
rs750609595 Chr.X: 3033308-3033308 on Build GRCh38
rs61978642 Chr.X: 3033307-3033307 on Build GRCh38
rs56954982 Chr.X: 3033606-3033606 on Build GRCh38
rs911915808 Chr.X: 3033612-3033612 on Build GRCh38
rs754036441 Chr.X: 3033317-3033317 on Build GRCh38
rs146403199 Chr.X: 3033424-3033424 on Build GRCh38
rs1003152740 Chr.X: 3033400-3033400 on Build GRCh38
rs922667620 Chr.X: 3033702-3033702 on Build GRCh38
rs982955605 Chr.X: 3033414-3033414 on Build GRCh38
rs775457220 Chr.X: 3033431-3033431 on Build GRCh38
rs777174842 Chr.X: 3033390-3033390 on Build GRCh38
rs908332490 Chr.X: 3033619-3033619 on Build GRCh38
rs1052937657 Chr.X: 3033690-3033690 on Build GRCh38
rs781714677 Chr.X: 3033535-3033535 on Build GRCh38
rs5939155 Chr.X: 3033393-3033393 on Build GRCh38
rs778458964 Chr.X: 3033734-3033734 on Build GRCh38
rs1053662349 Chr.X: 3033527-3033527 on Build GRCh38
rs892218185 Chr.X: 3033533-3033533 on Build GRCh38
rs977276018 Chr.X: 3033670-3033670 on Build GRCh38
rs765429274 Chr.X: 3033424-3033424 on Build GRCh38

View Full Product Details