Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 133360178-133360680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SURF2
Entrez Gene ID
6835
Gene Name
surfeit 2
Gene Aliases
SURF-2
UniGene
Hs.159448
Gene Chromosome Location
Chr.9: 133356545-133361165 on Build GRCh38
Transcripts
NM_001278928.1 NM_017503.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3905269 c.646G>A p.E216K Chr.9: 133360393-133360393 on Build GRCh38
4949530 c.661C>G p.R221G Chr.9: 133360408-133360408 on Build GRCh38
3929894 c.551C>T p.T184M Chr.9: 133360298-133360298 on Build GRCh38
6425954 c.679C>A p.R227S Chr.9: 133360426-133360426 on Build GRCh38
3213356 c.638G>A p.S213N Chr.9: 133360385-133360385 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782681873 Chr.9: 133360370-133360370 on Build GRCh38
rs141313898 Chr.9: 133360337-133360337 on Build GRCh38
rs1002135603 Chr.9: 133360607-133360607 on Build GRCh38
rs375185119 Chr.9: 133360218-133360218 on Build GRCh38
rs975774084 Chr.9: 133360549-133360549 on Build GRCh38
rs1034517776 Chr.9: 133360653-133360653 on Build GRCh38
rs781972573 Chr.9: 133360473-133360473 on Build GRCh38
rs782773644 Chr.9: 133360229-133360229 on Build GRCh38
rs961663844 Chr.9: 133360314-133360314 on Build GRCh38
rs745823928 Chr.9: 133360329-133360332 on Build GRCh38
rs781857591 Chr.9: 133360358-133360358 on Build GRCh38
rs1025016473 Chr.9: 133360432-133360432 on Build GRCh38
rs935964333 Chr.9: 133360559-133360559 on Build GRCh38
rs367625058 Chr.9: 133360379-133360379 on Build GRCh38
rs926043611 Chr.9: 133360548-133360548 on Build GRCh38
rs935692481 Chr.9: 133360197-133360197 on Build GRCh38
rs143679153 Chr.9: 133360360-133360360 on Build GRCh38
rs375240778 Chr.9: 133360555-133360555 on Build GRCh38
rs782635332 Chr.9: 133360427-133360427 on Build GRCh38
rs782631692 Chr.9: 133360537-133360537 on Build GRCh38
rs587679179 Chr.9: 133360407-133360407 on Build GRCh38
rs976008010 Chr.9: 133360516-133360516 on Build GRCh38
rs371730978 Chr.9: 133360429-133360429 on Build GRCh38
rs782419050 Chr.9: 133360441-133360441 on Build GRCh38
rs781822748 Chr.9: 133360505-133360505 on Build GRCh38
rs782052522 Chr.9: 133360274-133360274 on Build GRCh38
rs1017335332 Chr.9: 133360309-133360309 on Build GRCh38
rs139626160 Chr.9: 133360245-133360245 on Build GRCh38
rs969834849 Chr.9: 133360199-133360199 on Build GRCh38
rs782117114 Chr.9: 133360402-133360402 on Build GRCh38
rs782014664 Chr.9: 133360459-133360459 on Build GRCh38
rs931887322 Chr.9: 133360570-133360570 on Build GRCh38
rs781329837 Chr.9: 133360250-133360251 on Build GRCh38
rs782227511 Chr.9: 133360265-133360266 on Build GRCh38
rs201253644 Chr.9: 133360331-133360331 on Build GRCh38
rs587775331 Chr.9: 133360655-133360655 on Build GRCh38
rs369306757 Chr.9: 133360336-133360336 on Build GRCh38
rs781910805 Chr.9: 133360419-133360419 on Build GRCh38
rs782517068 Chr.9: 133360519-133360519 on Build GRCh38
rs587626466 Chr.9: 133360401-133360401 on Build GRCh38
rs912907081 Chr.9: 133360533-133360533 on Build GRCh38
rs782642385 Chr.9: 133360389-133360390 on Build GRCh38
rs782403751 Chr.9: 133360292-133360292 on Build GRCh38
rs12763 Chr.9: 133360384-133360384 on Build GRCh38
rs989825142 Chr.9: 133360363-133360363 on Build GRCh38
rs7863933 Chr.9: 133360409-133360409 on Build GRCh38
rs768411905 Chr.9: 133360526-133360526 on Build GRCh38
rs1057411 Chr.9: 133360375-133360375 on Build GRCh38
rs782258896 Chr.9: 133360462-133360462 on Build GRCh38
rs60003745 Chr.9: 133360217-133360217 on Build GRCh38
rs782088433 Chr.9: 133360475-133360475 on Build GRCh38
rs782281339 Chr.9: 133360248-133360248 on Build GRCh38
rs782492337 Chr.9: 133360359-133360359 on Build GRCh38
rs782663158 Chr.9: 133360367-133360369 on Build GRCh38
rs782568061 Chr.9: 133360647-133360647 on Build GRCh38
rs782343859 Chr.9: 133360550-133360551 on Build GRCh38
rs782741319 Chr.9: 133360317-133360317 on Build GRCh38
rs146539596 Chr.9: 133360300-133360300 on Build GRCh38
rs781959450 Chr.9: 133360381-133360381 on Build GRCh38
rs901635380 Chr.9: 133360240-133360240 on Build GRCh38
rs587730522 Chr.9: 133360386-133360386 on Build GRCh38
rs782542115 Chr.9: 133360600-133360600 on Build GRCh38
rs782180121 Chr.9: 133360431-133360431 on Build GRCh38
rs906060059 Chr.9: 133360422-133360422 on Build GRCh38
rs377182275 Chr.9: 133360430-133360430 on Build GRCh38
rs781889297 Chr.9: 133360426-133360426 on Build GRCh38
rs975905070 Chr.9: 133360311-133360311 on Build GRCh38
rs1045719638 Chr.9: 133360604-133360604 on Build GRCh38
rs782439907 Chr.9: 133360450-133360451 on Build GRCh38
rs782063410 Chr.9: 133360326-133360326 on Build GRCh38
rs587709365 Chr.9: 133360621-133360621 on Build GRCh38
rs781804323 Chr.9: 133360241-133360241 on Build GRCh38
rs887544441 Chr.9: 133360641-133360641 on Build GRCh38
rs62637582 Chr.9: 133360397-133360397 on Build GRCh38
rs782237808 Chr.9: 133360260-133360260 on Build GRCh38
rs74713084 Chr.9: 133360428-133360428 on Build GRCh38
rs28516839 Chr.9: 133360576-133360576 on Build GRCh38
rs782396281 Chr.9: 133360252-133360252 on Build GRCh38
rs782760849 Chr.9: 133360471-133360471 on Build GRCh38
rs950272157 Chr.9: 133360580-133360580 on Build GRCh38
rs1046441767 Chr.9: 133360593-133360593 on Build GRCh38
rs1014770158 Chr.9: 133360491-133360491 on Build GRCh38
rs782795928 Chr.9: 133360324-133360324 on Build GRCh38
rs1053113181 Chr.9: 133360569-133360569 on Build GRCh38
rs781973263 Chr.9: 133360214-133360214 on Build GRCh38
rs941452868 Chr.9: 133360588-133360588 on Build GRCh38
rs781827181 Chr.9: 133360408-133360408 on Build GRCh38
rs906013151 Chr.9: 133360594-133360594 on Build GRCh38
rs372069635 Chr.9: 133360258-133360258 on Build GRCh38
rs931834371 Chr.9: 133360524-133360524 on Build GRCh38
rs587600299 Chr.9: 133360298-133360298 on Build GRCh38
rs781992111 Chr.9: 133360299-133360299 on Build GRCh38
rs186385572 Chr.9: 133360246-133360246 on Build GRCh38
rs140549826 Chr.9: 133360398-133360398 on Build GRCh38
rs782248643 Chr.9: 133360434-133360434 on Build GRCh38
rs897235125 Chr.9: 133360590-133360590 on Build GRCh38
rs782530420 Chr.9: 133360223-133360223 on Build GRCh38
rs782079415 Chr.9: 133360383-133360383 on Build GRCh38
rs782316400 Chr.9: 133360369-133360369 on Build GRCh38
rs782649923 Chr.9: 133360353-133360353 on Build GRCh38
rs587696398 Chr.9: 133360513-133360513 on Build GRCh38
rs782197024 Chr.9: 133360368-133360368 on Build GRCh38
rs200846373 Chr.9: 133360262-133360262 on Build GRCh38
rs782717544 Chr.9: 133360476-133360476 on Build GRCh38
rs199647229 Chr.9: 133360235-133360235 on Build GRCh38
rs782015994 Chr.9: 133360654-133360654 on Build GRCh38
rs748978322 Chr.9: 133360518-133360519 on Build GRCh38
rs1038894143 Chr.9: 133360589-133360589 on Build GRCh38
rs879183000 Chr.9: 133360264-133360264 on Build GRCh38
rs781919822 Chr.9: 133360388-133360388 on Build GRCh38
rs781854301 Chr.9: 133360232-133360232 on Build GRCh38
rs782250665 Chr.9: 133360372-133360372 on Build GRCh38
rs782369344 Chr.9: 133360465-133360465 on Build GRCh38
rs781947332 Chr.9: 133360271-133360271 on Build GRCh38
rs782116986 Chr.9: 133360403-133360403 on Build GRCh38
rs202019985 Chr.9: 133360315-133360315 on Build GRCh38
rs782705626 Chr.9: 133360411-133360411 on Build GRCh38

View Full Product Details