Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 62388368-62388917 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF365
Entrez Gene ID
22891
Gene Name
zinc finger protein 365
Gene Aliases
Su48, UAN, ZNF365D
UniGene
Hs.22653
Gene Chromosome Location
Chr.10: 62373609-62672011 on Build GRCh38
Transcripts
NM_014951.2 NM_199450.2 NM_199451.2 XM_017015937.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2158219 c.842G>A p.R281Q Chr.10: 62388494-62388494 on Build GRCh38
1505942 c.809G>A p.R270Q Chr.10: 62388461-62388461 on Build GRCh38
3439546 c.843G>A p.R281R Chr.10: 62388495-62388495 on Build GRCh38
1171705 c.841C>T p.R281W Chr.10: 62388493-62388493 on Build GRCh38
4918640 c.850C>G p.L284V Chr.10: 62388502-62388502 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745478861 Chr.10: 62388394-62388394 on Build GRCh38
rs908323441 Chr.10: 62388787-62388787 on Build GRCh38
rs1042739378 Chr.10: 62388443-62388443 on Build GRCh38
rs750454816 Chr.10: 62388596-62388596 on Build GRCh38
rs928090498 Chr.10: 62388777-62388777 on Build GRCh38
rs1032902374 Chr.10: 62388653-62388653 on Build GRCh38
rs376625531 Chr.10: 62388579-62388579 on Build GRCh38
rs946722995 Chr.10: 62388396-62388396 on Build GRCh38
rs754614222 Chr.10: 62388616-62388616 on Build GRCh38
rs551952528 Chr.10: 62388798-62388798 on Build GRCh38
rs771265677 Chr.10: 62388569-62388569 on Build GRCh38
rs751767621 Chr.10: 62388450-62388450 on Build GRCh38
rs941069478 Chr.10: 62388492-62388492 on Build GRCh38
rs763271481 Chr.10: 62388568-62388568 on Build GRCh38
rs2893897 Chr.10: 62388610-62388610 on Build GRCh38
rs964013510 Chr.10: 62388654-62388654 on Build GRCh38
rs74156188 Chr.10: 62388601-62388601 on Build GRCh38
rs938150188 Chr.10: 62388784-62388784 on Build GRCh38
rs367827531 Chr.10: 62388431-62388431 on Build GRCh38
rs766562467 Chr.10: 62388449-62388449 on Build GRCh38
rs757292372 Chr.10: 62388481-62388481 on Build GRCh38
rs889549507 Chr.10: 62388539-62388539 on Build GRCh38
rs531423000 Chr.10: 62388404-62388404 on Build GRCh38
rs140880170 Chr.10: 62388815-62388815 on Build GRCh38
rs1037899238 Chr.10: 62388535-62388535 on Build GRCh38
rs374943096 Chr.10: 62388618-62388618 on Build GRCh38
rs941016956 Chr.10: 62388805-62388805 on Build GRCh38
rs1000227635 Chr.10: 62388648-62388648 on Build GRCh38
rs116297631 Chr.10: 62388556-62388556 on Build GRCh38
rs746701226 Chr.10: 62388532-62388532 on Build GRCh38
rs937609219 Chr.10: 62388847-62388847 on Build GRCh38
rs191326652 Chr.10: 62388576-62388576 on Build GRCh38
rs770073528 Chr.10: 62388429-62388429 on Build GRCh38
rs767595424 Chr.10: 62388587-62388587 on Build GRCh38
rs955418481 Chr.10: 62388752-62388752 on Build GRCh38
rs200153426 Chr.10: 62388567-62388567 on Build GRCh38
rs752742970 Chr.10: 62388588-62388588 on Build GRCh38
rs997980356 Chr.10: 62388480-62388480 on Build GRCh38
rs975613271 Chr.10: 62388673-62388673 on Build GRCh38
rs920819166 Chr.10: 62388825-62388825 on Build GRCh38
rs1029410527 Chr.10: 62388521-62388521 on Build GRCh38
rs567008368 Chr.10: 62388527-62388527 on Build GRCh38
rs148035675 Chr.10: 62388408-62388408 on Build GRCh38
rs374076744 Chr.10: 62388557-62388557 on Build GRCh38
rs775055367 Chr.10: 62388401-62388401 on Build GRCh38
rs763110946 Chr.10: 62388445-62388445 on Build GRCh38
rs192211282 Chr.10: 62388421-62388421 on Build GRCh38
rs765237370 Chr.10: 62388592-62388592 on Build GRCh38
rs758310261 Chr.10: 62388493-62388493 on Build GRCh38
rs760887858 Chr.10: 62388589-62388589 on Build GRCh38
rs577733385 Chr.10: 62388742-62388742 on Build GRCh38
rs750221846 Chr.10: 62388489-62388489 on Build GRCh38
rs778803179 Chr.10: 62388484-62388484 on Build GRCh38
rs778856479 Chr.10: 62388392-62388392 on Build GRCh38
rs368278289 Chr.10: 62388566-62388566 on Build GRCh38
rs758257255 Chr.10: 62388600-62388600 on Build GRCh38
rs71508919 Chr.10: 62388816-62388816 on Build GRCh38
rs547383793 Chr.10: 62388461-62388461 on Build GRCh38
rs767551232 Chr.10: 62388469-62388469 on Build GRCh38
rs369288154 Chr.10: 62388619-62388619 on Build GRCh38
rs774619216 Chr.10: 62388570-62388570 on Build GRCh38
rs777186162 Chr.10: 62388413-62388413 on Build GRCh38
rs899442977 Chr.10: 62388546-62388546 on Build GRCh38
rs184098438 Chr.10: 62388460-62388460 on Build GRCh38
rs771875693 Chr.10: 62388400-62388400 on Build GRCh38
rs572179220 Chr.10: 62388428-62388428 on Build GRCh38
rs1056332012 Chr.10: 62388882-62388882 on Build GRCh38
rs745708419 Chr.10: 62388476-62388476 on Build GRCh38
rs1029506175 Chr.10: 62388735-62388735 on Build GRCh38
rs982492683 Chr.10: 62388765-62388765 on Build GRCh38

View Full Product Details