Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 95245834-95246366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf37
Entrez Gene ID
157657
Gene Name
chromosome 8 open reading frame 37
Gene Aliases
BBS21, CORD16, RP64, smalltalk
UniGene
Hs.548157
Gene Chromosome Location
Chr.8: 95244913-95270602 on Build GRCh38
Transcripts
NM_177965.3 XM_005250799.3

SNP Detail

SNP ID Chromosome Location
rs998647818 Chr.8: 95246116-95246116 on Build GRCh38
rs62522657 Chr.8: 95246013-95246013 on Build GRCh38
rs959375255 Chr.8: 95245930-95245930 on Build GRCh38
rs752842895 Chr.8: 95245939-95245939 on Build GRCh38
rs975959163 Chr.8: 95246219-95246219 on Build GRCh38
rs747749238 Chr.8: 95246238-95246238 on Build GRCh38
rs1033137959 Chr.8: 95246119-95246119 on Build GRCh38
rs368518141 Chr.8: 95246146-95246146 on Build GRCh38
rs886063192 Chr.8: 95245942-95245942 on Build GRCh38
rs778569217 Chr.8: 95245978-95245978 on Build GRCh38
rs559314854 Chr.8: 95246105-95246105 on Build GRCh38
rs938037709 Chr.8: 95246336-95246336 on Build GRCh38
rs984894963 Chr.8: 95246279-95246279 on Build GRCh38
rs1032715440 Chr.8: 95245900-95245900 on Build GRCh38
rs1042601009 Chr.8: 95246113-95246113 on Build GRCh38
rs1003978116 Chr.8: 95246067-95246067 on Build GRCh38
rs998584333 Chr.8: 95245862-95245862 on Build GRCh38
rs926399359 Chr.8: 95245913-95245913 on Build GRCh38
rs532315210 Chr.8: 95245988-95245988 on Build GRCh38
rs886063191 Chr.8: 95245930-95245930 on Build GRCh38
rs758354515 Chr.8: 95246094-95246094 on Build GRCh38
rs746561645 Chr.8: 95246114-95246114 on Build GRCh38
rs1054831782 Chr.8: 95246025-95246025 on Build GRCh38
rs929421093 Chr.8: 95245953-95245953 on Build GRCh38
rs909253969 Chr.8: 95246317-95246317 on Build GRCh38
rs965958623 Chr.8: 95246218-95246218 on Build GRCh38
rs919125915 Chr.8: 95246268-95246268 on Build GRCh38
rs372353759 Chr.8: 95246042-95246042 on Build GRCh38
rs548195674 Chr.8: 95246144-95246144 on Build GRCh38
rs779926352 Chr.8: 95246106-95246106 on Build GRCh38
rs937969592 Chr.8: 95245989-95245989 on Build GRCh38
rs748612624 Chr.8: 95246050-95246050 on Build GRCh38
rs775094115 Chr.8: 95245992-95245993 on Build GRCh38
rs147952646 Chr.8: 95246143-95246143 on Build GRCh38
rs189282313 Chr.8: 95246286-95246286 on Build GRCh38
rs62522656 Chr.8: 95245950-95245950 on Build GRCh38
rs1055387174 Chr.8: 95246041-95246041 on Build GRCh38
rs967093325 Chr.8: 95245861-95245861 on Build GRCh38
rs890972035 Chr.8: 95246047-95246047 on Build GRCh38
rs1024704383 Chr.8: 95246307-95246307 on Build GRCh38
rs995215831 Chr.8: 95246126-95246126 on Build GRCh38
rs1007609906 Chr.8: 95246145-95246145 on Build GRCh38
rs977548262 Chr.8: 95246318-95246318 on Build GRCh38
rs1015467276 Chr.8: 95246082-95246082 on Build GRCh38
rs962371730 Chr.8: 95245890-95245890 on Build GRCh38
rs974106770 Chr.8: 95245903-95245903 on Build GRCh38
rs562105277 Chr.8: 95245899-95245899 on Build GRCh38
rs934365003 Chr.8: 95246095-95246095 on Build GRCh38
rs184820378 Chr.8: 95245947-95245947 on Build GRCh38
rs536667071 Chr.8: 95246278-95246278 on Build GRCh38
rs561198464 Chr.8: 95245993-95245993 on Build GRCh38
rs192783069 Chr.8: 95245897-95245897 on Build GRCh38
rs992302252 Chr.8: 95246220-95246220 on Build GRCh38
rs965863896 Chr.8: 95245942-95245942 on Build GRCh38
rs957261696 Chr.8: 95245911-95245911 on Build GRCh38
rs986070254 Chr.8: 95245934-95245934 on Build GRCh38
rs950551435 Chr.8: 95246269-95246269 on Build GRCh38
rs564592768 Chr.8: 95245979-95245979 on Build GRCh38
rs10109660 Chr.8: 95246159-95246159 on Build GRCh38
rs769417563 Chr.8: 95246280-95246280 on Build GRCh38

View Full Product Details