Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 34418283-34418779 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS10
Entrez Gene ID
6204
Gene Name
ribosomal protein S10
Gene Aliases
DBA9, S10
UniGene
Hs.406620
Gene Chromosome Location
Chr.6: 34417454-34426125 on Build GRCh38
Transcripts
NM_001014.4 NM_001203245.2 NM_001204091.1 NM_001202470.2

Gene Symbol
RPS10-NUDT3
Entrez Gene ID
100529239
Gene Name
RPS10-NUDT3 readthrough
Gene Aliases
-
UniGene
Hs.188882
Gene Chromosome Location
Chr.6: 34287194-34426125 on Build GRCh38
Transcripts
NM_001014.4 NM_001203245.2 NM_001204091.1 NM_001202470.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
742272 c.438A>T p.S146S Chr.6: 34418387-34418387 on Build GRCh38
5445263 c.445G>A p.E149K Chr.6: 34418380-34418380 on Build GRCh38
461787 c.414G>C p.K138N Chr.6: 34418411-34418411 on Build GRCh38
6768897 c.409G>A p.D137N Chr.6: 34418416-34418416 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs987926862 Chr.6: 34418559-34418559 on Build GRCh38
rs754412473 Chr.6: 34418439-34418449 on Build GRCh38
rs774151484 Chr.6: 34418530-34418530 on Build GRCh38
rs1013247110 Chr.6: 34418532-34418532 on Build GRCh38
rs969196290 Chr.6: 34418318-34418318 on Build GRCh38
rs368791535 Chr.6: 34418388-34418388 on Build GRCh38
rs972066033 Chr.6: 34418588-34418588 on Build GRCh38
rs918484370 Chr.6: 34418618-34418618 on Build GRCh38
rs41269040 Chr.6: 34418442-34418442 on Build GRCh38
rs141985920 Chr.6: 34418639-34418640 on Build GRCh38
rs999179400 Chr.6: 34418740-34418740 on Build GRCh38
rs568496534 Chr.6: 34418635-34418635 on Build GRCh38
rs551244017 Chr.6: 34418512-34418512 on Build GRCh38
rs9394213 Chr.6: 34418513-34418513 on Build GRCh38
rs1011058953 Chr.6: 34418504-34418504 on Build GRCh38
rs368006253 Chr.6: 34418465-34418465 on Build GRCh38
rs180930706 Chr.6: 34418714-34418714 on Build GRCh38
rs766816705 Chr.6: 34418440-34418440 on Build GRCh38
rs557394189 Chr.6: 34418351-34418351 on Build GRCh38
rs770796118 Chr.6: 34418354-34418354 on Build GRCh38
rs753421531 Chr.6: 34418441-34418441 on Build GRCh38
rs1029554231 Chr.6: 34418527-34418527 on Build GRCh38
rs775548399 Chr.6: 34418380-34418380 on Build GRCh38
rs911980760 Chr.6: 34418579-34418579 on Build GRCh38
rs191872288 Chr.6: 34418308-34418308 on Build GRCh38
rs777660016 Chr.6: 34418343-34418343 on Build GRCh38
rs781105913 Chr.6: 34418472-34418472 on Build GRCh38
rs760917852 Chr.6: 34418433-34418433 on Build GRCh38
rs560780193 Chr.6: 34418540-34418540 on Build GRCh38
rs762252535 Chr.6: 34418404-34418404 on Build GRCh38
rs962017111 Chr.6: 34418586-34418586 on Build GRCh38
rs73744897 Chr.6: 34418626-34418626 on Build GRCh38
rs766928999 Chr.6: 34418434-34418435 on Build GRCh38
rs548914871 Chr.6: 34418627-34418627 on Build GRCh38
rs947913170 Chr.6: 34418688-34418688 on Build GRCh38
rs1024731746 Chr.6: 34418455-34418455 on Build GRCh38
rs961907367 Chr.6: 34418605-34418605 on Build GRCh38
rs147566753 Chr.6: 34418417-34418417 on Build GRCh38
rs984572600 Chr.6: 34418563-34418563 on Build GRCh38
rs971912115 Chr.6: 34418620-34418621 on Build GRCh38
rs149556277 Chr.6: 34418697-34418697 on Build GRCh38
rs999471503 Chr.6: 34418471-34418471 on Build GRCh38
rs778507034 Chr.6: 34418444-34418444 on Build GRCh38
rs755895934 Chr.6: 34418687-34418690 on Build GRCh38
rs559654960 Chr.6: 34418329-34418329 on Build GRCh38
rs903303625 Chr.6: 34418749-34418752 on Build GRCh38
rs1050361610 Chr.6: 34418650-34418650 on Build GRCh38
rs1043589926 Chr.6: 34418452-34418452 on Build GRCh38
rs199758134 Chr.6: 34418393-34418393 on Build GRCh38
rs532802045 Chr.6: 34418483-34418483 on Build GRCh38
rs562137217 Chr.6: 34418476-34418476 on Build GRCh38
rs752276242 Chr.6: 34418456-34418456 on Build GRCh38
rs781063077 Chr.6: 34418479-34418479 on Build GRCh38
rs906157481 Chr.6: 34418457-34418457 on Build GRCh38
rs754065393 Chr.6: 34418335-34418335 on Build GRCh38
rs747013389 Chr.6: 34418461-34418461 on Build GRCh38
rs149897629 Chr.6: 34418381-34418381 on Build GRCh38
rs768038583 Chr.6: 34418416-34418416 on Build GRCh38
rs535947692 Chr.6: 34418669-34418669 on Build GRCh38
rs1048320517 Chr.6: 34418661-34418661 on Build GRCh38
rs138756023 Chr.6: 34418474-34418474 on Build GRCh38
rs758371386 Chr.6: 34418460-34418460 on Build GRCh38
rs909068219 Chr.6: 34418350-34418350 on Build GRCh38

View Full Product Details