Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 74586605-74587144 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RLIM
Entrez Gene ID
51132
Gene Name
ring finger protein, LIM domain interacting
Gene Aliases
NY-REN-43, RNF12
UniGene
Hs.653288
Gene Chromosome Location
Chr.X: 74582976-74614626 on Build GRCh38
Transcripts
NM_016120.3 NM_183353.2

SNP Detail

SNP ID Chromosome Location
rs756024078 Chr.X: 74586717-74586717 on Build GRCh38
rs1052868174 Chr.X: 74586911-74586911 on Build GRCh38
rs996145932 Chr.X: 74587095-74587099 on Build GRCh38
rs939579059 Chr.X: 74586758-74586758 on Build GRCh38
rs948426254 Chr.X: 74586832-74586832 on Build GRCh38
rs1027669796 Chr.X: 74587097-74587099 on Build GRCh38
rs964988710 Chr.X: 74587079-74587079 on Build GRCh38
rs1044040660 Chr.X: 74586889-74586889 on Build GRCh38
rs987220008 Chr.X: 74587023-74587023 on Build GRCh38
rs954539611 Chr.X: 74586679-74586679 on Build GRCh38
rs183003640 Chr.X: 74586780-74586780 on Build GRCh38
rs1042452176 Chr.X: 74586792-74586792 on Build GRCh38
rs943285853 Chr.X: 74586768-74586768 on Build GRCh38
rs555247847 Chr.X: 74586944-74586944 on Build GRCh38
rs904254867 Chr.X: 74586901-74586901 on Build GRCh38
rs5937851 Chr.X: 74587024-74587024 on Build GRCh38
rs983590450 Chr.X: 74586648-74586648 on Build GRCh38
rs910430213 Chr.X: 74586729-74586729 on Build GRCh38
rs945393410 Chr.X: 74586785-74586785 on Build GRCh38
rs906624386 Chr.X: 74586809-74586809 on Build GRCh38
rs1026028030 Chr.X: 74586963-74586963 on Build GRCh38
rs1009222749 Chr.X: 74586922-74586922 on Build GRCh38
rs767939308 Chr.X: 74586914-74586914 on Build GRCh38
rs999922195 Chr.X: 74586904-74586904 on Build GRCh38
rs908053098 Chr.X: 74586718-74586718 on Build GRCh38
rs915232396 Chr.X: 74586782-74586782 on Build GRCh38
rs41303703 Chr.X: 74586920-74586920 on Build GRCh38
rs895356835 Chr.X: 74586843-74586843 on Build GRCh38
rs987272679 Chr.X: 74586683-74586683 on Build GRCh38
rs1055304778 Chr.X: 74586836-74586836 on Build GRCh38
rs762185681 Chr.X: 74586846-74586846 on Build GRCh38
rs5981651 Chr.X: 74586933-74586933 on Build GRCh38
rs775020802 Chr.X: 74586824-74586824 on Build GRCh38
rs954616253 Chr.X: 74586978-74586978 on Build GRCh38
rs7066577 Chr.X: 74586915-74586915 on Build GRCh38
rs752618586 Chr.X: 74586646-74586646 on Build GRCh38

View Full Product Details