Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 74587135-74587680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RLIM
Entrez Gene ID
51132
Gene Name
ring finger protein, LIM domain interacting
Gene Aliases
NY-REN-43, RNF12
UniGene
Hs.653288
Gene Chromosome Location
Chr.X: 74582976-74614626 on Build GRCh38
Transcripts
NM_016120.3 NM_183353.2

SNP Detail

SNP ID Chromosome Location
rs1029827610 Chr.X: 74587275-74587275 on Build GRCh38
rs778928501 Chr.X: 74587506-74587506 on Build GRCh38
rs992814915 Chr.X: 74587210-74587210 on Build GRCh38
rs967033334 Chr.X: 74587549-74587549 on Build GRCh38
rs1027637206 Chr.X: 74587508-74587508 on Build GRCh38
rs955710060 Chr.X: 74587519-74587519 on Build GRCh38
rs1038587109 Chr.X: 74587377-74587377 on Build GRCh38
rs1040619169 Chr.X: 74587463-74587464 on Build GRCh38
rs865992780 Chr.X: 74587319-74587319 on Build GRCh38
rs928033059 Chr.X: 74587238-74587238 on Build GRCh38
rs754040763 Chr.X: 74587162-74587162 on Build GRCh38
rs944516198 Chr.X: 74587404-74587404 on Build GRCh38
rs766052857 Chr.X: 74587565-74587565 on Build GRCh38
rs770171853 Chr.X: 74587433-74587434 on Build GRCh38
rs748650079 Chr.X: 74587192-74587193 on Build GRCh38
rs939416417 Chr.X: 74587247-74587247 on Build GRCh38
rs891621476 Chr.X: 74587388-74587388 on Build GRCh38
rs993137876 Chr.X: 74587437-74587437 on Build GRCh38
rs960936694 Chr.X: 74587163-74587163 on Build GRCh38
rs1047846488 Chr.X: 74587464-74587464 on Build GRCh38
rs1055251736 Chr.X: 74587294-74587294 on Build GRCh38
rs571425948 Chr.X: 74587536-74587536 on Build GRCh38
rs897406942 Chr.X: 74587399-74587399 on Build GRCh38
rs945369887 Chr.X: 74587174-74587174 on Build GRCh38
rs935741113 Chr.X: 74587339-74587339 on Build GRCh38
rs755074710 Chr.X: 74587169-74587169 on Build GRCh38
rs762537618 Chr.X: 74587557-74587557 on Build GRCh38
rs900391326 Chr.X: 74587464-74587464 on Build GRCh38
rs996113354 Chr.X: 74587487-74587489 on Build GRCh38
rs948322261 Chr.X: 74587226-74587226 on Build GRCh38
rs916797593 Chr.X: 74587217-74587217 on Build GRCh38
rs773223009 Chr.X: 74587571-74587574 on Build GRCh38
rs978505680 Chr.X: 74587207-74587207 on Build GRCh38
rs890153237 Chr.X: 74587495-74587495 on Build GRCh38
rs757080836 Chr.X: 74587560-74587560 on Build GRCh38
rs895305868 Chr.X: 74587310-74587310 on Build GRCh38
rs964491563 Chr.X: 74587533-74587533 on Build GRCh38
rs751523159 Chr.X: 74587540-74587540 on Build GRCh38
rs781171855 Chr.X: 74587638-74587638 on Build GRCh38

View Full Product Details