Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 75785016-75785543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGEE2
Entrez Gene ID
139599
Gene Name
MAGE family member E2
Gene Aliases
HCA3
UniGene
Hs.356869
Gene Chromosome Location
Chr.X: 75782988-75785244 on Build GRCh38
Transcripts
NM_138703.4

SNP Detail

SNP ID Chromosome Location
rs763060005 Chr.X: 75785299-75785299 on Build GRCh38
rs1011485492 Chr.X: 75785223-75785223 on Build GRCh38
rs1052216459 Chr.X: 75785363-75785363 on Build GRCh38
rs1043373870 Chr.X: 75785258-75785258 on Build GRCh38
rs964262469 Chr.X: 75785075-75785075 on Build GRCh38
rs982922097 Chr.X: 75785112-75785112 on Build GRCh38
rs760354639 Chr.X: 75785077-75785077 on Build GRCh38
rs1007629101 Chr.X: 75785183-75785183 on Build GRCh38
rs756003829 Chr.X: 75785108-75785108 on Build GRCh38
rs73625865 Chr.X: 75785383-75785383 on Build GRCh38
rs991274248 Chr.X: 75785305-75785305 on Build GRCh38
rs774167769 Chr.X: 75785246-75785246 on Build GRCh38
rs933586708 Chr.X: 75785362-75785362 on Build GRCh38
rs752019042 Chr.X: 75785490-75785490 on Build GRCh38
rs765244377 Chr.X: 75785376-75785376 on Build GRCh38
rs947937428 Chr.X: 75785222-75785222 on Build GRCh38
rs939221516 Chr.X: 75785149-75785149 on Build GRCh38
rs1018826882 Chr.X: 75785500-75785500 on Build GRCh38
rs951358174 Chr.X: 75785111-75785111 on Build GRCh38
rs773264726 Chr.X: 75785071-75785071 on Build GRCh38
rs761781381 Chr.X: 75785039-75785039 on Build GRCh38
rs942292399 Chr.X: 75785045-75785045 on Build GRCh38
rs777702164 Chr.X: 75785130-75785130 on Build GRCh38
rs189489714 Chr.X: 75785165-75785165 on Build GRCh38
rs1054313756 Chr.X: 75785433-75785433 on Build GRCh38
rs892999949 Chr.X: 75785208-75785208 on Build GRCh38
rs776569114 Chr.X: 75785294-75785294 on Build GRCh38
rs992377682 Chr.X: 75785159-75785159 on Build GRCh38
rs373745004 Chr.X: 75785292-75785292 on Build GRCh38
rs776501592 Chr.X: 75785193-75785193 on Build GRCh38
rs903566997 Chr.X: 75785345-75785345 on Build GRCh38
rs761884657 Chr.X: 75785355-75785355 on Build GRCh38
rs766820454 Chr.X: 75785487-75785487 on Build GRCh38
rs766244475 Chr.X: 75785092-75785092 on Build GRCh38
rs943454416 Chr.X: 75785431-75785431 on Build GRCh38
rs1051701157 Chr.X: 75785375-75785375 on Build GRCh38
rs1051733169 Chr.X: 75785146-75785146 on Build GRCh38
rs981082607 Chr.X: 75785256-75785256 on Build GRCh38
rs958425083 Chr.X: 75785285-75785285 on Build GRCh38
rs890848250 Chr.X: 75785409-75785409 on Build GRCh38
rs909372677 Chr.X: 75785308-75785308 on Build GRCh38
rs774510705 Chr.X: 75785357-75785357 on Build GRCh38
rs747205431 Chr.X: 75785266-75785266 on Build GRCh38
rs768671695 Chr.X: 75785300-75785300 on Build GRCh38
rs972260946 Chr.X: 75785329-75785329 on Build GRCh38
rs1008411926 Chr.X: 75785420-75785420 on Build GRCh38
rs771887173 Chr.X: 75785070-75785070 on Build GRCh38
rs181601548 Chr.X: 75785414-75785414 on Build GRCh38
rs1018550148 Chr.X: 75785069-75785069 on Build GRCh38
rs963843480 Chr.X: 75785309-75785309 on Build GRCh38
rs140744282 Chr.X: 75785237-75785237 on Build GRCh38

View Full Product Details