Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20476313-20476912 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PNP
Entrez Gene ID
4860
Gene Name
purine nucleoside phosphorylase
Gene Aliases
NP, PRO1837, PUNP
UniGene
Hs.75514
Gene Chromosome Location
Chr.14: 20469379-20478006 on Build GRCh38
Transcripts
NM_000270.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5053845 c.679G>A p.V227I Chr.14: 20476410-20476410 on Build GRCh38
7160086 c.819G>C p.Q273H Chr.14: 20476550-20476550 on Build GRCh38
1262633 c.772_774delGAA p.E259delE Chr.14: 20476503-20476505 on Build GRCh38
5053844 c.653-2A>G p.? Chr.14: 20476382-20476382 on Build GRCh38
5517792 c.748G>A p.E250K Chr.14: 20476479-20476479 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866626659 Chr.14: 20476715-20476715 on Build GRCh38
rs778150398 Chr.14: 20476602-20476602 on Build GRCh38
rs779851923 Chr.14: 20476501-20476501 on Build GRCh38
rs898455107 Chr.14: 20476549-20476549 on Build GRCh38
rs755234109 Chr.14: 20476538-20476538 on Build GRCh38
rs775032909 Chr.14: 20476449-20476449 on Build GRCh38
rs987189241 Chr.14: 20476820-20476820 on Build GRCh38
rs104894451 Chr.14: 20476432-20476432 on Build GRCh38
rs569036203 Chr.14: 20476679-20476679 on Build GRCh38
rs752340519 Chr.14: 20476352-20476352 on Build GRCh38
rs375289878 Chr.14: 20476615-20476615 on Build GRCh38
rs753845133 Chr.14: 20476379-20476379 on Build GRCh38
rs949202515 Chr.14: 20476783-20476783 on Build GRCh38
rs112999633 Chr.14: 20476882-20476882 on Build GRCh38
rs781318202 Chr.14: 20476336-20476336 on Build GRCh38
rs763654890 Chr.14: 20476376-20476376 on Build GRCh38
rs367615281 Chr.14: 20476638-20476638 on Build GRCh38
rs748196799 Chr.14: 20476417-20476417 on Build GRCh38
rs7785 Chr.14: 20476855-20476855 on Build GRCh38
rs772541140 Chr.14: 20476701-20476701 on Build GRCh38
rs182309640 Chr.14: 20476836-20476836 on Build GRCh38
rs111742618 Chr.14: 20476826-20476826 on Build GRCh38
rs1019775744 Chr.14: 20476864-20476864 on Build GRCh38
rs972582182 Chr.14: 20476848-20476848 on Build GRCh38
rs768488828 Chr.14: 20476618-20476618 on Build GRCh38
rs755580654 Chr.14: 20476373-20476373 on Build GRCh38
rs749760621 Chr.14: 20476611-20476611 on Build GRCh38
rs566125117 Chr.14: 20476730-20476730 on Build GRCh38
rs1036184563 Chr.14: 20476769-20476769 on Build GRCh38
rs371184527 Chr.14: 20476460-20476460 on Build GRCh38
rs767136169 Chr.14: 20476344-20476344 on Build GRCh38
rs1000846561 Chr.14: 20476829-20476829 on Build GRCh38
rs370513543 Chr.14: 20476571-20476571 on Build GRCh38
rs758159693 Chr.14: 20476487-20476487 on Build GRCh38
rs761403833 Chr.14: 20476467-20476467 on Build GRCh38
rs940627229 Chr.14: 20476755-20476755 on Build GRCh38
rs965475737 Chr.14: 20476873-20476873 on Build GRCh38
rs375159414 Chr.14: 20476468-20476468 on Build GRCh38
rs754229427 Chr.14: 20476735-20476735 on Build GRCh38
rs758196911 Chr.14: 20476400-20476400 on Build GRCh38
rs886050390 Chr.14: 20476375-20476375 on Build GRCh38
rs1032567583 Chr.14: 20476479-20476479 on Build GRCh38
rs149957827 Chr.14: 20476424-20476424 on Build GRCh38
rs371711940 Chr.14: 20476639-20476639 on Build GRCh38
rs999050537 Chr.14: 20476722-20476722 on Build GRCh38
rs753438749 Chr.14: 20476466-20476466 on Build GRCh38
rs368488127 Chr.14: 20476511-20476511 on Build GRCh38
rs563086985 Chr.14: 20476409-20476409 on Build GRCh38
rs748289161 Chr.14: 20476550-20476550 on Build GRCh38
rs912675762 Chr.14: 20476824-20476824 on Build GRCh38
rs771220552 Chr.14: 20476612-20476612 on Build GRCh38
rs892708636 Chr.14: 20476841-20476841 on Build GRCh38
rs199866228 Chr.14: 20476380-20476380 on Build GRCh38
rs896550845 Chr.14: 20476431-20476431 on Build GRCh38
rs1000877435 Chr.14: 20476463-20476463 on Build GRCh38
rs886050391 Chr.14: 20476509-20476521 on Build GRCh38
rs201497504 Chr.14: 20476616-20476616 on Build GRCh38
rs771263911 Chr.14: 20476433-20476433 on Build GRCh38
rs139364283 Chr.14: 20476416-20476416 on Build GRCh38
rs748702029 Chr.14: 20476551-20476551 on Build GRCh38
rs763375783 Chr.14: 20476342-20476342 on Build GRCh38
rs763812491 Chr.14: 20476464-20476464 on Build GRCh38
rs535741893 Chr.14: 20476654-20476654 on Build GRCh38
rs764882608 Chr.14: 20476743-20476743 on Build GRCh38
rs776408969 Chr.14: 20476632-20476632 on Build GRCh38
rs1045141311 Chr.14: 20476811-20476811 on Build GRCh38
rs773751064 Chr.14: 20476546-20476546 on Build GRCh38
rs988309791 Chr.14: 20476579-20476579 on Build GRCh38
rs931229423 Chr.14: 20476605-20476605 on Build GRCh38
rs745716644 Chr.14: 20476396-20476396 on Build GRCh38
rs780151852 Chr.14: 20476404-20476404 on Build GRCh38
rs912415165 Chr.14: 20476477-20476477 on Build GRCh38
rs761420050 Chr.14: 20476633-20476633 on Build GRCh38
rs76571622 Chr.14: 20476779-20476779 on Build GRCh38
rs61755047 Chr.14: 20476410-20476410 on Build GRCh38
rs754595756 Chr.14: 20476740-20476740 on Build GRCh38
rs774308308 Chr.14: 20476341-20476341 on Build GRCh38
rs551176074 Chr.14: 20476393-20476393 on Build GRCh38
rs902244094 Chr.14: 20476719-20476719 on Build GRCh38
rs372200673 Chr.14: 20476652-20476652 on Build GRCh38
rs151283499 Chr.14: 20476814-20476814 on Build GRCh38
rs770355699 Chr.14: 20476429-20476429 on Build GRCh38
rs931895349 Chr.14: 20476371-20476371 on Build GRCh38
rs766791132 Chr.14: 20476681-20476681 on Build GRCh38
rs555762832 Chr.14: 20476875-20476875 on Build GRCh38
rs750316115 Chr.14: 20476472-20476472 on Build GRCh38
rs139143167 Chr.14: 20476881-20476881 on Build GRCh38

View Full Product Details