Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 32636175-32636771 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP11A
Entrez Gene ID
9824
Gene Name
Rho GTPase activating protein 11A
Gene Aliases
GAP (1-12)
UniGene
Hs.591130
Gene Chromosome Location
Chr.15: 32615144-32639949 on Build GRCh38
Transcripts
NM_001286479.1 NM_001286480.1 NM_014783.4 NM_199357.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
960463 c.1782G>T p.E594D Chr.15: 32636555-32636555 on Build GRCh38
4451938 c.1495A>G p.I499V Chr.15: 32636268-32636268 on Build GRCh38
6603025 c.1621G>T p.V541L Chr.15: 32636394-32636394 on Build GRCh38
4469966 c.1630C>T p.L544F Chr.15: 32636403-32636403 on Build GRCh38
5644963 c.1663A>G p.M555V Chr.15: 32636436-32636436 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780887436 Chr.15: 32636544-32636544 on Build GRCh38
rs146499716 Chr.15: 32636324-32636324 on Build GRCh38
rs61758695 Chr.15: 32636652-32636652 on Build GRCh38
rs745795797 Chr.15: 32636396-32636396 on Build GRCh38
rs559605810 Chr.15: 32636560-32636560 on Build GRCh38
rs760502227 Chr.15: 32636304-32636304 on Build GRCh38
rs775156342 Chr.15: 32636378-32636378 on Build GRCh38
rs780749897 Chr.15: 32636651-32636651 on Build GRCh38
rs372403826 Chr.15: 32636238-32636238 on Build GRCh38
rs186578850 Chr.15: 32636325-32636325 on Build GRCh38
rs755388866 Chr.15: 32636330-32636330 on Build GRCh38
rs769514202 Chr.15: 32636540-32636540 on Build GRCh38
rs561697586 Chr.15: 32636724-32636724 on Build GRCh38
rs746595137 Chr.15: 32636409-32636409 on Build GRCh38
rs1038225235 Chr.15: 32636451-32636451 on Build GRCh38
rs776223285 Chr.15: 32636417-32636417 on Build GRCh38
rs556950894 Chr.15: 32636319-32636319 on Build GRCh38
rs375487655 Chr.15: 32636272-32636272 on Build GRCh38
rs773263026 Chr.15: 32636455-32636455 on Build GRCh38
rs756643216 Chr.15: 32636358-32636358 on Build GRCh38
rs776655424 Chr.15: 32636311-32636311 on Build GRCh38
rs138580047 Chr.15: 32636270-32636270 on Build GRCh38
rs778239587 Chr.15: 32636363-32636363 on Build GRCh38
rs780424096 Chr.15: 32636216-32636216 on Build GRCh38
rs769722243 Chr.15: 32636656-32636656 on Build GRCh38
rs1047623150 Chr.15: 32636476-32636476 on Build GRCh38
rs765363769 Chr.15: 32636317-32636317 on Build GRCh38
rs771769961 Chr.15: 32636301-32636301 on Build GRCh38
rs115031896 Chr.15: 32636434-32636434 on Build GRCh38
rs773779981 Chr.15: 32636292-32636292 on Build GRCh38
rs373354756 Chr.15: 32636663-32636663 on Build GRCh38
rs764392576 Chr.15: 32636596-32636596 on Build GRCh38
rs531220822 Chr.15: 32636688-32636688 on Build GRCh38
rs949630215 Chr.15: 32636356-32636356 on Build GRCh38
rs557744320 Chr.15: 32636525-32636525 on Build GRCh38
rs755085144 Chr.15: 32636492-32636492 on Build GRCh38
rs771047764 Chr.15: 32636667-32636667 on Build GRCh38
rs868164508 Chr.15: 32636430-32636430 on Build GRCh38
rs140803286 Chr.15: 32636370-32636370 on Build GRCh38
rs775597191 Chr.15: 32636720-32636720 on Build GRCh38
rs369362748 Chr.15: 32636452-32636452 on Build GRCh38
rs906875073 Chr.15: 32636372-32636372 on Build GRCh38
rs779484378 Chr.15: 32636528-32636528 on Build GRCh38
rs776718156 Chr.15: 32636606-32636606 on Build GRCh38
rs1033200713 Chr.15: 32636654-32636654 on Build GRCh38
rs914090958 Chr.15: 32636725-32636725 on Build GRCh38
rs771137664 Chr.15: 32636563-32636563 on Build GRCh38
rs751092235 Chr.15: 32636632-32636632 on Build GRCh38
rs762923878 Chr.15: 32636561-32636561 on Build GRCh38
rs894259370 Chr.15: 32636335-32636335 on Build GRCh38
rs370442091 Chr.15: 32636526-32636526 on Build GRCh38
rs781121046 Chr.15: 32636456-32636456 on Build GRCh38
rs759644956 Chr.15: 32636571-32636571 on Build GRCh38
rs769682763 Chr.15: 32636549-32636549 on Build GRCh38
rs747946271 Chr.15: 32636547-32636547 on Build GRCh38
rs374926086 Chr.15: 32636326-32636326 on Build GRCh38
rs747437914 Chr.15: 32636230-32636230 on Build GRCh38
rs771537185 Chr.15: 32636374-32636374 on Build GRCh38
rs201097638 Chr.15: 32636511-32636511 on Build GRCh38
rs767680198 Chr.15: 32636577-32636577 on Build GRCh38
rs573976518 Chr.15: 32636497-32636497 on Build GRCh38
rs898994310 Chr.15: 32636458-32636458 on Build GRCh38
rs781351606 Chr.15: 32636331-32636331 on Build GRCh38
rs771829860 Chr.15: 32636632-32636633 on Build GRCh38
rs201457056 Chr.15: 32636286-32636286 on Build GRCh38
rs993313474 Chr.15: 32636471-32636471 on Build GRCh38
rs1045487941 Chr.15: 32636702-32636702 on Build GRCh38
rs931541429 Chr.15: 32636598-32636598 on Build GRCh38
rs761598791 Chr.15: 32636316-32636316 on Build GRCh38
rs756478051 Chr.15: 32636498-32636498 on Build GRCh38
rs763168810 Chr.15: 32636475-32636475 on Build GRCh38
rs972707513 Chr.15: 32636578-32636578 on Build GRCh38
rs202105243 Chr.15: 32636700-32636700 on Build GRCh38
rs781736910 Chr.15: 32636256-32636256 on Build GRCh38
rs779388959 Chr.15: 32636612-32636612 on Build GRCh38
rs112715874 Chr.15: 32636500-32636500 on Build GRCh38
rs775099347 Chr.15: 32636303-32636303 on Build GRCh38
rs755279399 Chr.15: 32636245-32636245 on Build GRCh38
rs199884674 Chr.15: 32636607-32636607 on Build GRCh38
rs762856943 Chr.15: 32636510-32636510 on Build GRCh38
rs760976558 Chr.15: 32636588-32636588 on Build GRCh38
rs545619705 Chr.15: 32636647-32636647 on Build GRCh38
rs373006108 Chr.15: 32636462-32636462 on Build GRCh38
rs748689204 Chr.15: 32636343-32636343 on Build GRCh38
rs746678098 Chr.15: 32636529-32636529 on Build GRCh38
rs751841609 Chr.15: 32636482-32636482 on Build GRCh38
rs772151113 Chr.15: 32636718-32636718 on Build GRCh38
rs764447695 Chr.15: 32636745-32636745 on Build GRCh38
rs954308130 Chr.15: 32636550-32636550 on Build GRCh38
rs376278433 Chr.15: 32636479-32636479 on Build GRCh38
rs986897972 Chr.15: 32636553-32636553 on Build GRCh38
rs926619183 Chr.15: 32636428-32636428 on Build GRCh38
rs374853485 Chr.15: 32636579-32636579 on Build GRCh38
rs751802895 Chr.15: 32636327-32636327 on Build GRCh38
rs774357269 Chr.15: 32636565-32636565 on Build GRCh38
rs988279997 Chr.15: 32636694-32636694 on Build GRCh38
rs369384014 Chr.15: 32636298-32636298 on Build GRCh38
rs774321509 Chr.15: 32636686-32636686 on Build GRCh38
rs768509850 Chr.15: 32636410-32636410 on Build GRCh38
rs373156350 Chr.15: 32636305-32636305 on Build GRCh38
rs199734120 Chr.15: 32636608-32636608 on Build GRCh38

View Full Product Details