Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48410236-48410807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBN1
Entrez Gene ID
2200
Gene Name
fibrillin 1
Gene Aliases
ACMICD, ECTOL1, FBN, GPHYSD2, MASS, MFLS, MFS1, OCTD, SGS, SSKS, WMS, WMS2
UniGene
Hs.591133
Gene Chromosome Location
Chr.15: 48408306-48645788 on Build GRCh38
Transcripts
NM_000138.4

SNP Detail

SNP ID Chromosome Location
rs140923867 Chr.15: 48410489-48410489 on Build GRCh38
rs755147629 Chr.15: 48410550-48410550 on Build GRCh38
rs1026526739 Chr.15: 48410387-48410387 on Build GRCh38
rs763450242 Chr.15: 48410658-48410658 on Build GRCh38
rs886686959 Chr.15: 48410422-48410422 on Build GRCh38
rs894774727 Chr.15: 48410679-48410679 on Build GRCh38
rs990506418 Chr.15: 48410470-48410470 on Build GRCh38
rs955018922 Chr.15: 48410509-48410509 on Build GRCh38
rs901981116 Chr.15: 48410280-48410280 on Build GRCh38
rs13598 Chr.15: 48410260-48410260 on Build GRCh38
rs1006865592 Chr.15: 48410675-48410675 on Build GRCh38
rs397702821 Chr.15: 48410261-48410261 on Build GRCh38
rs539383558 Chr.15: 48410737-48410737 on Build GRCh38
rs551258633 Chr.15: 48410453-48410453 on Build GRCh38
rs920215896 Chr.15: 48410281-48410281 on Build GRCh38
rs886051243 Chr.15: 48410736-48410736 on Build GRCh38
rs968026887 Chr.15: 48410463-48410463 on Build GRCh38
rs761985133 Chr.15: 48410487-48410487 on Build GRCh38
rs1022545724 Chr.15: 48410458-48410458 on Build GRCh38
rs773342320 Chr.15: 48410652-48410652 on Build GRCh38
rs963748087 Chr.15: 48410644-48410644 on Build GRCh38
rs886051241 Chr.15: 48410704-48410704 on Build GRCh38
rs375866380 Chr.15: 48410268-48410268 on Build GRCh38
rs571869753 Chr.15: 48410735-48410735 on Build GRCh38
rs886051242 Chr.15: 48410722-48410722 on Build GRCh38
rs1011876253 Chr.15: 48410690-48410690 on Build GRCh38
rs186005462 Chr.15: 48410604-48410604 on Build GRCh38
rs548674669 Chr.15: 48410602-48410602 on Build GRCh38
rs780110628 Chr.15: 48410356-48410356 on Build GRCh38
rs923838873 Chr.15: 48410523-48410523 on Build GRCh38
rs933429195 Chr.15: 48410649-48410649 on Build GRCh38
rs936579323 Chr.15: 48410572-48410572 on Build GRCh38
rs970449151 Chr.15: 48410725-48410725 on Build GRCh38
rs376410505 Chr.15: 48410744-48410744 on Build GRCh38
rs886051240 Chr.15: 48410584-48410584 on Build GRCh38
rs1055858503 Chr.15: 48410429-48410429 on Build GRCh38
rs986526476 Chr.15: 48410539-48410539 on Build GRCh38
rs1047093313 Chr.15: 48410347-48410347 on Build GRCh38
rs914541392 Chr.15: 48410576-48410576 on Build GRCh38
rs1002975429 Chr.15: 48410417-48410417 on Build GRCh38
rs930220028 Chr.15: 48410301-48410301 on Build GRCh38
rs920082241 Chr.15: 48410671-48410671 on Build GRCh38
rs1042078 Chr.15: 48410676-48410676 on Build GRCh38
rs200718684 Chr.15: 48410724-48410724 on Build GRCh38
rs989782818 Chr.15: 48410748-48410748 on Build GRCh38
rs533522969 Chr.15: 48410536-48410536 on Build GRCh38
rs533061153 Chr.15: 48410340-48410340 on Build GRCh38
rs1047737230 Chr.15: 48410714-48410714 on Build GRCh38
rs977898891 Chr.15: 48410498-48410498 on Build GRCh38
rs943371991 Chr.15: 48410601-48410601 on Build GRCh38
rs1006505865 Chr.15: 48410444-48410444 on Build GRCh38
rs974310310 Chr.15: 48410660-48410660 on Build GRCh38
rs752081445 Chr.15: 48410763-48410763 on Build GRCh38
rs997690529 Chr.15: 48410372-48410372 on Build GRCh38
rs977846549 Chr.15: 48410736-48410736 on Build GRCh38
rs989443401 Chr.15: 48410573-48410573 on Build GRCh38
rs368918344 Chr.15: 48410327-48410327 on Build GRCh38
rs959092536 Chr.15: 48410460-48410460 on Build GRCh38
rs901928828 Chr.15: 48410636-48410636 on Build GRCh38
rs748707223 Chr.15: 48410718-48410718 on Build GRCh38
rs537505904 Chr.15: 48410673-48410673 on Build GRCh38
rs886051239 Chr.15: 48410266-48410266 on Build GRCh38
rs16960899 Chr.15: 48410767-48410767 on Build GRCh38
rs537909430 Chr.15: 48410665-48410665 on Build GRCh38

View Full Product Details