Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77061526-77061999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYCBP2
Entrez Gene ID
23077
Gene Name
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases
Myc-bp2, PAM, Phr
UniGene
Hs.591221
Gene Chromosome Location
Chr.13: 77044657-77327066 on Build GRCh38
Transcripts
NM_015057.4 XM_005266299.2 XM_006719779.2 XM_006719780.2 XM_006719781.2 XM_006719782.2 XM_006719783.2 XM_006719784.2 XM_006719785.2 XM_006719786.2 XM_006719787.2 XM_006719788.2 XM_006719789.2 XM_011535004.2 XM_011535005.2 XM_017020454.1 XM_017020455.1 XM_017020456.1 XM_017020457.1 XM_017020458.1 XM_017020459.1 XM_017020460.1 XM_017020461.1 XM_017020462.1 XM_017020463.1 XM_017020464.1 XM_017020465.1 XM_017020466.1 XM_017020467.1 XM_017020468.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1949702 c.12826A>C p.N4276H Chr.13: 77061739-77061739 on Build GRCh38
1367725 c.12761A>G p.H4254R Chr.13: 77061690-77061690 on Build GRCh38
1607223 c.12793G>A p.D4265N Chr.13: 77061772-77061772 on Build GRCh38
1367727 c.12677A>G p.H4226R Chr.13: 77061774-77061774 on Build GRCh38
7193167 c.12774A>C p.K4258N Chr.13: 77061677-77061677 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901815405 Chr.13: 77061640-77061640 on Build GRCh38
rs183650084 Chr.13: 77061882-77061882 on Build GRCh38
rs77195856 Chr.13: 77061783-77061783 on Build GRCh38
rs936706307 Chr.13: 77061966-77061966 on Build GRCh38
rs933151238 Chr.13: 77061943-77061943 on Build GRCh38
rs958425965 Chr.13: 77061888-77061888 on Build GRCh38
rs908861846 Chr.13: 77061920-77061920 on Build GRCh38
rs560842375 Chr.13: 77061877-77061877 on Build GRCh38
rs187556810 Chr.13: 77061555-77061555 on Build GRCh38
rs970854280 Chr.13: 77061859-77061859 on Build GRCh38
rs776488528 Chr.13: 77061833-77061833 on Build GRCh38
rs985374770 Chr.13: 77061925-77061925 on Build GRCh38
rs764878975 Chr.13: 77061682-77061682 on Build GRCh38
rs930844767 Chr.13: 77061569-77061569 on Build GRCh38
rs776176852 Chr.13: 77061647-77061647 on Build GRCh38
rs371362078 Chr.13: 77061734-77061734 on Build GRCh38
rs757587384 Chr.13: 77061689-77061689 on Build GRCh38
rs760708049 Chr.13: 77061616-77061616 on Build GRCh38
rs772154789 Chr.13: 77061810-77061810 on Build GRCh38
rs889173859 Chr.13: 77061573-77061573 on Build GRCh38
rs374652237 Chr.13: 77061750-77061750 on Build GRCh38
rs759046938 Chr.13: 77061679-77061679 on Build GRCh38
rs780689984 Chr.13: 77061746-77061746 on Build GRCh38
rs1050227034 Chr.13: 77061944-77061944 on Build GRCh38
rs748330803 Chr.13: 77061808-77061808 on Build GRCh38
rs147087914 Chr.13: 77061551-77061551 on Build GRCh38
rs750851054 Chr.13: 77061852-77061852 on Build GRCh38
rs770969533 Chr.13: 77061826-77061826 on Build GRCh38
rs1010227956 Chr.13: 77061671-77061671 on Build GRCh38
rs984957147 Chr.13: 77061575-77061575 on Build GRCh38
rs369373750 Chr.13: 77061811-77061811 on Build GRCh38
rs141978651 Chr.13: 77061902-77061902 on Build GRCh38
rs960394596 Chr.13: 77061876-77061876 on Build GRCh38
rs756688977 Chr.13: 77061743-77061743 on Build GRCh38
rs1053648913 Chr.13: 77061655-77061655 on Build GRCh38
rs527367393 Chr.13: 77061764-77061764 on Build GRCh38
rs150275330 Chr.13: 77061933-77061933 on Build GRCh38
rs758788277 Chr.13: 77061932-77061932 on Build GRCh38
rs192546735 Chr.13: 77061562-77061562 on Build GRCh38
rs752291996 Chr.13: 77061687-77061687 on Build GRCh38
rs56274460 Chr.13: 77061586-77061586 on Build GRCh38
rs1040374081 Chr.13: 77061605-77061605 on Build GRCh38
rs766206724 Chr.13: 77061633-77061633 on Build GRCh38
rs923126083 Chr.13: 77061940-77061940 on Build GRCh38
rs779289700 Chr.13: 77061790-77061790 on Build GRCh38
rs767880319 Chr.13: 77061707-77061707 on Build GRCh38
rs549129732 Chr.13: 77061817-77061817 on Build GRCh38
rs757566485 Chr.13: 77061584-77061584 on Build GRCh38
rs79231543 Chr.13: 77061557-77061557 on Build GRCh38
rs1026506001 Chr.13: 77061889-77061889 on Build GRCh38
rs892728332 Chr.13: 77061638-77061638 on Build GRCh38
rs1002081981 Chr.13: 77061850-77061850 on Build GRCh38
rs750719931 Chr.13: 77061740-77061740 on Build GRCh38
rs755041001 Chr.13: 77061765-77061765 on Build GRCh38
rs889205093 Chr.13: 77061952-77061952 on Build GRCh38
rs981185512 Chr.13: 77061873-77061873 on Build GRCh38
rs754521731 Chr.13: 77061769-77061770 on Build GRCh38

View Full Product Details