Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 130350550-130351026 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A14
Entrez Gene ID
9016
Gene Name
solute carrier family 25 member 14
Gene Aliases
BMCP1, UCP5
UniGene
Hs.194686
Gene Chromosome Location
Chr.X: 130339888-130373361 on Build GRCh38
Transcripts
NM_001282195.1 NM_001282196.1 NM_001282197.1 NM_001282198.1 NR_104107.1 XM_005262489.4 XM_011531402.2 XM_011531403.1 XM_017029936.1 XM_017029937.1 XM_017029938.1 XM_017029939.1 XM_017029940.1 XM_017029941.1 XM_017029942.1 XM_017029943.1

SNP Detail

SNP ID Chromosome Location
rs145005879 Chr.X: 130350654-130350654 on Build GRCh38
rs753206105 Chr.X: 130350630-130350630 on Build GRCh38
rs145386095 Chr.X: 130350719-130350719 on Build GRCh38
rs950482453 Chr.X: 130350927-130350927 on Build GRCh38
rs747189546 Chr.X: 130350667-130350667 on Build GRCh38
rs753428332 Chr.X: 130350598-130350598 on Build GRCh38
rs779024534 Chr.X: 130350805-130350820 on Build GRCh38
rs5930401 Chr.X: 130350677-130350677 on Build GRCh38
rs184749242 Chr.X: 130350804-130350804 on Build GRCh38
rs376543159 Chr.X: 130350762-130350762 on Build GRCh38
rs774366850 Chr.X: 130350748-130350748 on Build GRCh38
rs765769260 Chr.X: 130350595-130350595 on Build GRCh38
rs367623353 Chr.X: 130350611-130350611 on Build GRCh38
rs776383756 Chr.X: 130350883-130350883 on Build GRCh38
rs138798145 Chr.X: 130350657-130350657 on Build GRCh38
rs770903156 Chr.X: 130350829-130350829 on Build GRCh38
rs1018853044 Chr.X: 130350588-130350588 on Build GRCh38
rs781105686 Chr.X: 130350703-130350703 on Build GRCh38
rs746021349 Chr.X: 130350714-130350714 on Build GRCh38
rs753568681 Chr.X: 130350775-130350776 on Build GRCh38
rs932079114 Chr.X: 130350918-130350918 on Build GRCh38
rs755066251 Chr.X: 130350788-130350788 on Build GRCh38
rs1051754733 Chr.X: 130350940-130350940 on Build GRCh38
rs939263436 Chr.X: 130350957-130350957 on Build GRCh38
rs745706721 Chr.X: 130350969-130350969 on Build GRCh38
rs899423686 Chr.X: 130350999-130350999 on Build GRCh38
rs766148420 Chr.X: 130350741-130350742 on Build GRCh38
rs927673317 Chr.X: 130350956-130350956 on Build GRCh38
rs779540618 Chr.X: 130350616-130350616 on Build GRCh38
rs377089664 Chr.X: 130350873-130350873 on Build GRCh38
rs759612683 Chr.X: 130350961-130350961 on Build GRCh38
rs983064518 Chr.X: 130350942-130350942 on Build GRCh38

View Full Product Details